Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11555239-11555776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLK
Entrez Gene ID
640
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
MODY11
UniGene
Hs.146591
Gene Chromosome Location
Chr.8: 11493991-11564599 on Build GRCh38
Transcripts
NM_001715.2 XM_011543824.1 XM_011543825.2 XM_011543827.1 XM_011543828.2 XM_011543829.2 XM_017013730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6994541 c.667C>A p.P223T Chr.8: 11555379-11555379 on Build GRCh38
485988 c.723C>G p.L241L Chr.8: 11555435-11555435 on Build GRCh38
6900389 c.710C>A p.P237H Chr.8: 11555422-11555422 on Build GRCh38
3665306 c.665G>A p.R222H Chr.8: 11555377-11555377 on Build GRCh38
5873766 c.758G>A p.G253D Chr.8: 11555470-11555470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906238475 Chr.8: 11555669-11555669 on Build GRCh38
rs937821418 Chr.8: 11555302-11555302 on Build GRCh38
rs773963245 Chr.8: 11555471-11555471 on Build GRCh38
rs201252364 Chr.8: 11555384-11555384 on Build GRCh38
rs1042519713 Chr.8: 11555658-11555658 on Build GRCh38
rs771406613 Chr.8: 11555444-11555444 on Build GRCh38
rs763397385 Chr.8: 11555510-11555510 on Build GRCh38
rs779263158 Chr.8: 11555351-11555351 on Build GRCh38
rs377381294 Chr.8: 11555326-11555326 on Build GRCh38
rs1007905229 Chr.8: 11555498-11555498 on Build GRCh38
rs945378807 Chr.8: 11555436-11555436 on Build GRCh38
rs200515443 Chr.8: 11555472-11555472 on Build GRCh38
rs373560683 Chr.8: 11555391-11555391 on Build GRCh38
rs764915190 Chr.8: 11555321-11555321 on Build GRCh38
rs551088779 Chr.8: 11555389-11555389 on Build GRCh38
rs756089529 Chr.8: 11555532-11555532 on Build GRCh38
rs775121719 Chr.8: 11555381-11555381 on Build GRCh38
rs945241219 Chr.8: 11555567-11555567 on Build GRCh38
rs772086626 Chr.8: 11555398-11555398 on Build GRCh38
rs979577130 Chr.8: 11555399-11555399 on Build GRCh38
rs759506858 Chr.8: 11555483-11555484 on Build GRCh38
rs779861022 Chr.8: 11555473-11555473 on Build GRCh38
rs764391136 Chr.8: 11555456-11555456 on Build GRCh38
rs533412555 Chr.8: 11555360-11555360 on Build GRCh38
rs372042909 Chr.8: 11555725-11555725 on Build GRCh38
rs755201619 Chr.8: 11555348-11555348 on Build GRCh38
rs565575821 Chr.8: 11555469-11555469 on Build GRCh38
rs919845038 Chr.8: 11555590-11555590 on Build GRCh38
rs979909261 Chr.8: 11555260-11555260 on Build GRCh38
rs761815633 Chr.8: 11555521-11555521 on Build GRCh38
rs757191370 Chr.8: 11555411-11555411 on Build GRCh38
rs777866039 Chr.8: 11555367-11555367 on Build GRCh38
rs202028021 Chr.8: 11555380-11555380 on Build GRCh38
rs773896490 Chr.8: 11555431-11555431 on Build GRCh38
rs752889602 Chr.8: 11555356-11555356 on Build GRCh38
rs1001859848 Chr.8: 11555677-11555677 on Build GRCh38
rs563038217 Chr.8: 11555503-11555503 on Build GRCh38
rs766608059 Chr.8: 11555336-11555336 on Build GRCh38
rs746358606 Chr.8: 11555534-11555534 on Build GRCh38
rs551362684 Chr.8: 11555529-11555529 on Build GRCh38
rs931222410 Chr.8: 11555593-11555593 on Build GRCh38
rs759831348 Chr.8: 11555452-11555452 on Build GRCh38
rs111642844 Chr.8: 11555405-11555405 on Build GRCh38
rs753696674 Chr.8: 11555535-11555535 on Build GRCh38
rs1036711228 Chr.8: 11555362-11555362 on Build GRCh38
rs577734831 Chr.8: 11555726-11555726 on Build GRCh38
rs146907175 Chr.8: 11555614-11555614 on Build GRCh38
rs754196779 Chr.8: 11555344-11555344 on Build GRCh38
rs777128635 Chr.8: 11555447-11555447 on Build GRCh38
rs533113642 Chr.8: 11555515-11555515 on Build GRCh38
rs569138316 Chr.8: 11555313-11555313 on Build GRCh38
rs985543278 Chr.8: 11555598-11555598 on Build GRCh38
rs201698663 Chr.8: 11555501-11555501 on Build GRCh38
rs201671994 Chr.8: 11555453-11555453 on Build GRCh38
rs369356288 Chr.8: 11555308-11555308 on Build GRCh38
rs373207102 Chr.8: 11555312-11555312 on Build GRCh38
rs904118683 Chr.8: 11555659-11555659 on Build GRCh38
rs774629622 Chr.8: 11555317-11555318 on Build GRCh38
rs775031751 Chr.8: 11555322-11555322 on Build GRCh38
rs746873052 Chr.8: 11555370-11555370 on Build GRCh38
rs1025055505 Chr.8: 11555694-11555694 on Build GRCh38
rs750867843 Chr.8: 11555333-11555333 on Build GRCh38
rs2254959 Chr.8: 11555624-11555624 on Build GRCh38
rs972997368 Chr.8: 11555709-11555709 on Build GRCh38
rs12550718 Chr.8: 11555700-11555700 on Build GRCh38
rs535184490 Chr.8: 11555402-11555402 on Build GRCh38
rs144592409 Chr.8: 11555386-11555386 on Build GRCh38
rs143699141 Chr.8: 11555423-11555423 on Build GRCh38
rs766782778 Chr.8: 11555638-11555638 on Build GRCh38
rs549762171 Chr.8: 11555744-11555744 on Build GRCh38
rs200129390 Chr.8: 11555500-11555500 on Build GRCh38
rs897417171 Chr.8: 11555657-11555657 on Build GRCh38
rs148146433 Chr.8: 11555494-11555494 on Build GRCh38
rs771552059 Chr.8: 11555495-11555495 on Build GRCh38
rs966280631 Chr.8: 11555388-11555388 on Build GRCh38
rs765133469 Chr.8: 11555514-11555515 on Build GRCh38
rs901235103 Chr.8: 11555323-11555323 on Build GRCh38
rs761805613 Chr.8: 11555378-11555378 on Build GRCh38
rs968453155 Chr.8: 11555397-11555397 on Build GRCh38
rs778896633 Chr.8: 11555300-11555300 on Build GRCh38
rs748214310 Chr.8: 11555304-11555304 on Build GRCh38
rs898140517 Chr.8: 11555377-11555377 on Build GRCh38
rs1050297062 Chr.8: 11555597-11555597 on Build GRCh38
rs368103800 Chr.8: 11555531-11555531 on Build GRCh38
rs551647338 Chr.8: 11555379-11555379 on Build GRCh38
rs571695391 Chr.8: 11555302-11555306 on Build GRCh38
rs976698938 Chr.8: 11555736-11555736 on Build GRCh38
rs768101243 Chr.8: 11555544-11555544 on Build GRCh38
rs147111193 Chr.8: 11555517-11555517 on Build GRCh38
rs754679320 Chr.8: 11555490-11555490 on Build GRCh38
rs761120131 Chr.8: 11555335-11555335 on Build GRCh38
rs770572857 Chr.8: 11555309-11555309 on Build GRCh38
rs567736573 Chr.8: 11555607-11555607 on Build GRCh38
rs761122806 Chr.8: 11555387-11555387 on Build GRCh38
rs139300133 Chr.8: 11555731-11555731 on Build GRCh38
rs572913609 Chr.8: 11555670-11555670 on Build GRCh38
rs527672973 Chr.8: 11555740-11555740 on Build GRCh38
rs748894201 Chr.8: 11555429-11555429 on Build GRCh38
rs888518777 Chr.8: 11555613-11555613 on Build GRCh38
rs1040770562 Chr.8: 11555621-11555621 on Build GRCh38
rs141865425 Chr.8: 11555425-11555425 on Build GRCh38
rs777960256 Chr.8: 11555406-11555406 on Build GRCh38
rs200893448 Chr.8: 11555390-11555390 on Build GRCh38
rs772192692 Chr.8: 11555307-11555307 on Build GRCh38
rs149756811 Chr.8: 11555435-11555435 on Build GRCh38
rs943699818 Chr.8: 11555752-11555752 on Build GRCh38
rs922498703 Chr.8: 11555743-11555743 on Build GRCh38
rs985452806 Chr.8: 11555749-11555749 on Build GRCh38
rs749057360 Chr.8: 11555476-11555476 on Build GRCh38
rs775132094 Chr.8: 11555376-11555376 on Build GRCh38
rs755112854 Chr.8: 11555290-11555290 on Build GRCh38
rs993134221 Chr.8: 11555499-11555499 on Build GRCh38
rs1021619836 Chr.8: 11555520-11555520 on Build GRCh38
rs544403485 Chr.8: 11555723-11555723 on Build GRCh38
rs992229071 Chr.8: 11555536-11555536 on Build GRCh38
rs1033707757 Chr.8: 11555674-11555674 on Build GRCh38
rs376429865 Chr.8: 11555493-11555493 on Build GRCh38
rs534061256 Chr.8: 11555678-11555678 on Build GRCh38
rs751627957 Chr.8: 11555409-11555409 on Build GRCh38
rs549300104 Chr.8: 11555602-11555602 on Build GRCh38
rs73195298 Chr.8: 11555683-11555683 on Build GRCh38
rs377446062 Chr.8: 11555284-11555284 on Build GRCh38
rs1041038076 Chr.8: 11555442-11555442 on Build GRCh38
rs575123730 Chr.8: 11555424-11555424 on Build GRCh38
rs776575167 Chr.8: 11555420-11555421 on Build GRCh38
rs768081709 Chr.8: 11555327-11555327 on Build GRCh38
rs201158722 Chr.8: 11555468-11555468 on Build GRCh38
rs76563369 Chr.8: 11555465-11555465 on Build GRCh38
rs531541706 Chr.8: 11555660-11555660 on Build GRCh38
rs889551238 Chr.8: 11555496-11555496 on Build GRCh38
rs527993791 Chr.8: 11555542-11555542 on Build GRCh38
rs976751392 Chr.8: 11555577-11555577 on Build GRCh38
rs539412706 Chr.8: 11555645-11555645 on Build GRCh38
rs778620889 Chr.8: 11555492-11555492 on Build GRCh38
rs568937594 Chr.8: 11555627-11555627 on Build GRCh38
rs776047429 Chr.8: 11555507-11555507 on Build GRCh38
rs568112977 Chr.8: 11555394-11555394 on Build GRCh38
rs972709896 Chr.8: 11555546-11555546 on Build GRCh38
rs767782748 Chr.8: 11555579-11555579 on Build GRCh38
rs781177993 Chr.8: 11555421-11555421 on Build GRCh38

View Full Product Details