Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11561179-11561598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLK
Entrez Gene ID
640
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
MODY11
UniGene
Hs.146591
Gene Chromosome Location
Chr.8: 11493991-11564599 on Build GRCh38
Transcripts
NM_001715.2 XM_011543824.1 XM_011543825.2 XM_011543827.1 XM_017013730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1095196 c.1155C>T p.I385I Chr.8: 11561427-11561427 on Build GRCh38
5596353 c.1030-7C>T p.? Chr.8: 11561295-11561295 on Build GRCh38
5408699 c.1078G>A p.D360N Chr.8: 11561350-11561350 on Build GRCh38
1095194 c.1058G>A p.R353H Chr.8: 11561330-11561330 on Build GRCh38
3644432 c.1094A>G p.N365S Chr.8: 11561366-11561366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781408045 Chr.8: 11561384-11561384 on Build GRCh38
rs768486674 Chr.8: 11561277-11561277 on Build GRCh38
rs779019154 Chr.8: 11561203-11561207 on Build GRCh38
rs756818797 Chr.8: 11561247-11561247 on Build GRCh38
rs559281535 Chr.8: 11561543-11561543 on Build GRCh38
rs758750492 Chr.8: 11561320-11561320 on Build GRCh38
rs763350654 Chr.8: 11561354-11561354 on Build GRCh38
rs150788249 Chr.8: 11561442-11561442 on Build GRCh38
rs542670447 Chr.8: 11561416-11561416 on Build GRCh38
rs1010367356 Chr.8: 11561572-11561572 on Build GRCh38
rs907705035 Chr.8: 11561400-11561400 on Build GRCh38
rs139923395 Chr.8: 11561404-11561404 on Build GRCh38
rs146505280 Chr.8: 11561347-11561347 on Build GRCh38
rs1000547640 Chr.8: 11561563-11561563 on Build GRCh38
rs746550281 Chr.8: 11561423-11561423 on Build GRCh38
rs746047142 Chr.8: 11561556-11561556 on Build GRCh38
rs916969505 Chr.8: 11561519-11561519 on Build GRCh38
rs371390075 Chr.8: 11561495-11561495 on Build GRCh38
rs537449632 Chr.8: 11561221-11561221 on Build GRCh38
rs532290959 Chr.8: 11561518-11561518 on Build GRCh38
rs4841561 Chr.8: 11561264-11561264 on Build GRCh38
rs772087915 Chr.8: 11561238-11561238 on Build GRCh38
rs1044662933 Chr.8: 11561539-11561539 on Build GRCh38
rs182344018 Chr.8: 11561463-11561463 on Build GRCh38
rs759424446 Chr.8: 11561231-11561231 on Build GRCh38
rs267601734 Chr.8: 11561229-11561229 on Build GRCh38
rs774129378 Chr.8: 11561283-11561283 on Build GRCh38
rs749903162 Chr.8: 11561312-11561312 on Build GRCh38
rs898831886 Chr.8: 11561260-11561260 on Build GRCh38
rs766903237 Chr.8: 11561297-11561297 on Build GRCh38
rs761432905 Chr.8: 11561289-11561289 on Build GRCh38
rs778909298 Chr.8: 11561476-11561476 on Build GRCh38
rs554284389 Chr.8: 11561387-11561387 on Build GRCh38
rs762231914 Chr.8: 11561445-11561445 on Build GRCh38
rs763779683 Chr.8: 11561244-11561244 on Build GRCh38
rs369728833 Chr.8: 11561484-11561484 on Build GRCh38
rs771692863 Chr.8: 11561419-11561419 on Build GRCh38
rs758190809 Chr.8: 11561485-11561485 on Build GRCh38
rs927397531 Chr.8: 11561386-11561386 on Build GRCh38
rs747954528 Chr.8: 11561483-11561483 on Build GRCh38
rs751751964 Chr.8: 11561323-11561323 on Build GRCh38
rs753914596 Chr.8: 11561471-11561471 on Build GRCh38
rs931001952 Chr.8: 11561261-11561261 on Build GRCh38
rs55836779 Chr.8: 11561361-11561361 on Build GRCh38
rs770895584 Chr.8: 11561267-11561267 on Build GRCh38
rs762715079 Chr.8: 11561240-11561240 on Build GRCh38
rs1008427991 Chr.8: 11561288-11561288 on Build GRCh38
rs756299584 Chr.8: 11561452-11561452 on Build GRCh38
rs1008823268 Chr.8: 11561570-11561570 on Build GRCh38
rs747027477 Chr.8: 11561209-11561209 on Build GRCh38
rs749204324 Chr.8: 11561393-11561393 on Build GRCh38
rs931653094 Chr.8: 11561515-11561515 on Build GRCh38
rs778868587 Chr.8: 11561398-11561398 on Build GRCh38
rs374531191 Chr.8: 11561262-11561262 on Build GRCh38
rs771699695 Chr.8: 11561348-11561348 on Build GRCh38
rs142803179 Chr.8: 11561414-11561414 on Build GRCh38
rs73545881 Chr.8: 11561516-11561516 on Build GRCh38
rs780185322 Chr.8: 11561391-11561391 on Build GRCh38
rs368588162 Chr.8: 11561256-11561256 on Build GRCh38
rs770398225 Chr.8: 11561427-11561427 on Build GRCh38
rs769720795 Chr.8: 11561271-11561271 on Build GRCh38
rs139119999 Chr.8: 11561337-11561337 on Build GRCh38
rs746313966 Chr.8: 11561524-11561524 on Build GRCh38
rs751115445 Chr.8: 11561245-11561245 on Build GRCh38
rs748458653 Chr.8: 11561259-11561259 on Build GRCh38
rs746708387 Chr.8: 11561494-11561494 on Build GRCh38
rs776422416 Chr.8: 11561228-11561228 on Build GRCh38
rs886062595 Chr.8: 11561392-11561392 on Build GRCh38
rs753130362 Chr.8: 11561319-11561319 on Build GRCh38
rs144349989 Chr.8: 11561333-11561333 on Build GRCh38
rs575586576 Chr.8: 11561443-11561443 on Build GRCh38
rs769000604 Chr.8: 11561201-11561202 on Build GRCh38
rs748325634 Chr.8: 11561210-11561210 on Build GRCh38
rs760725491 Chr.8: 11561429-11561439 on Build GRCh38
rs904346710 Chr.8: 11561541-11561541 on Build GRCh38
rs908229455 Chr.8: 11561511-11561511 on Build GRCh38
rs149605952 Chr.8: 11561330-11561330 on Build GRCh38
rs776671995 Chr.8: 11561269-11561269 on Build GRCh38
rs891977539 Chr.8: 11561569-11561569 on Build GRCh38
rs773865077 Chr.8: 11561420-11561420 on Build GRCh38
rs773325977 Chr.8: 11561293-11561293 on Build GRCh38
rs189980189 Chr.8: 11561248-11561248 on Build GRCh38
rs757431553 Chr.8: 11561380-11561380 on Build GRCh38
rs773296708 Chr.8: 11561205-11561205 on Build GRCh38
rs141714152 Chr.8: 11561568-11561568 on Build GRCh38
rs62490888 Chr.8: 11561257-11561257 on Build GRCh38
rs940498822 Chr.8: 11561428-11561428 on Build GRCh38
rs778018964 Chr.8: 11561322-11561322 on Build GRCh38
rs745309214 Chr.8: 11561504-11561504 on Build GRCh38
rs751901837 Chr.8: 11561367-11561367 on Build GRCh38
rs368828603 Chr.8: 11561276-11561276 on Build GRCh38
rs199696853 Chr.8: 11561329-11561329 on Build GRCh38
rs759976637 Chr.8: 11561318-11561318 on Build GRCh38
rs750632024 Chr.8: 11561450-11561450 on Build GRCh38
rs768547419 Chr.8: 11561340-11561340 on Build GRCh38
rs770969777 Chr.8: 11561224-11561224 on Build GRCh38
rs767686222 Chr.8: 11561448-11561448 on Build GRCh38
rs542354024 Chr.8: 11561352-11561352 on Build GRCh38
rs749149481 Chr.8: 11561339-11561339 on Build GRCh38
rs980105504 Chr.8: 11561356-11561356 on Build GRCh38
rs774184424 Chr.8: 11561345-11561345 on Build GRCh38
rs756202005 Chr.8: 11561389-11561389 on Build GRCh38
rs147935690 Chr.8: 11561536-11561536 on Build GRCh38
rs772607284 Chr.8: 11561349-11561349 on Build GRCh38
rs775438519 Chr.8: 11561272-11561272 on Build GRCh38
rs769622708 Chr.8: 11561336-11561336 on Build GRCh38
rs112637820 Chr.8: 11561567-11561567 on Build GRCh38
rs777330405 Chr.8: 11561486-11561486 on Build GRCh38
rs776130008 Chr.8: 11561501-11561501 on Build GRCh38
rs754973341 Chr.8: 11561474-11561474 on Build GRCh38
rs768672215 Chr.8: 11561394-11561394 on Build GRCh38
rs776080933 Chr.8: 11561434-11561434 on Build GRCh38
rs763474991 Chr.8: 11561441-11561441 on Build GRCh38
rs772174625 Chr.8: 11561212-11561217 on Build GRCh38

View Full Product Details