Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11759334-11759829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA4
Entrez Gene ID
2626
Gene Name
GATA binding protein 4
Gene Aliases
ASD2, TACHD, TOF, VSD1
UniGene
Hs.243987
Gene Chromosome Location
Chr.8: 11676919-11760002 on Build GRCh38
Transcripts
NM_001308093.1 NM_001308094.1 NM_002052.4 XM_005272385.4 XM_005272386.1 XM_006716248.1 XM_011543817.2 XM_011543818.2 XM_017013312.1

SNP Detail

SNP ID Chromosome Location
rs971833228 Chr.8: 11759494-11759494 on Build GRCh38
rs574121823 Chr.8: 11759436-11759436 on Build GRCh38
rs566823371 Chr.8: 11759367-11759367 on Build GRCh38
rs1015995588 Chr.8: 11759475-11759475 on Build GRCh38
rs961569540 Chr.8: 11759641-11759641 on Build GRCh38
rs114395528 Chr.8: 11759611-11759611 on Build GRCh38
rs1050210292 Chr.8: 11759785-11759785 on Build GRCh38
rs1041146697 Chr.8: 11759597-11759597 on Build GRCh38
rs997258893 Chr.8: 11759617-11759617 on Build GRCh38
rs1005797125 Chr.8: 11759805-11759805 on Build GRCh38
rs1009499939 Chr.8: 11759414-11759414 on Build GRCh38
rs76841597 Chr.8: 11759743-11759743 on Build GRCh38
rs556537569 Chr.8: 11759682-11759682 on Build GRCh38
rs758617309 Chr.8: 11759495-11759495 on Build GRCh38
rs932642678 Chr.8: 11759784-11759784 on Build GRCh38
rs144191832 Chr.8: 11759514-11759514 on Build GRCh38
rs770994309 Chr.8: 11759616-11759616 on Build GRCh38
rs922687788 Chr.8: 11759775-11759775 on Build GRCh38
rs905020993 Chr.8: 11759363-11759363 on Build GRCh38
rs534277520 Chr.8: 11759640-11759640 on Build GRCh38
rs528811175 Chr.8: 11759541-11759541 on Build GRCh38
rs1019555645 Chr.8: 11759424-11759424 on Build GRCh38
rs888480324 Chr.8: 11759798-11759798 on Build GRCh38
rs936487956 Chr.8: 11759549-11759549 on Build GRCh38
rs370376188 Chr.8: 11759467-11759467 on Build GRCh38
rs1025145812 Chr.8: 11759649-11759649 on Build GRCh38
rs146304341 Chr.8: 11759361-11759361 on Build GRCh38
rs191938369 Chr.8: 11759672-11759672 on Build GRCh38
rs892563848 Chr.8: 11759413-11759413 on Build GRCh38
rs989385856 Chr.8: 11759685-11759685 on Build GRCh38
rs567765903 Chr.8: 11759657-11759657 on Build GRCh38
rs534316677 Chr.8: 11759376-11759376 on Build GRCh38
rs945451194 Chr.8: 11759574-11759574 on Build GRCh38
rs772859891 Chr.8: 11759796-11759796 on Build GRCh38
rs139566390 Chr.8: 11759487-11759487 on Build GRCh38
rs182365313 Chr.8: 11759454-11759454 on Build GRCh38
rs949109930 Chr.8: 11759510-11759510 on Build GRCh38
rs11785481 Chr.8: 11759633-11759633 on Build GRCh38
rs12458 Chr.8: 11759731-11759731 on Build GRCh38
rs139741267 Chr.8: 11759683-11759683 on Build GRCh38
rs945251884 Chr.8: 11759750-11759750 on Build GRCh38
rs1041070552 Chr.8: 11759755-11759755 on Build GRCh38
rs780370506 Chr.8: 11759429-11759429 on Build GRCh38
rs901171843 Chr.8: 11759610-11759610 on Build GRCh38
rs756676481 Chr.8: 11759362-11759362 on Build GRCh38
rs533275667 Chr.8: 11759631-11759631 on Build GRCh38
rs952873138 Chr.8: 11759450-11759450 on Build GRCh38
rs1037324956 Chr.8: 11759806-11759806 on Build GRCh38
rs115161018 Chr.8: 11759636-11759636 on Build GRCh38
rs557944704 Chr.8: 11759366-11759366 on Build GRCh38
rs528154800 Chr.8: 11759518-11759518 on Build GRCh38
rs1029032619 Chr.8: 11759437-11759437 on Build GRCh38
rs561339130 Chr.8: 11759527-11759527 on Build GRCh38
rs746685830 Chr.8: 11759728-11759728 on Build GRCh38
rs187432043 Chr.8: 11759513-11759513 on Build GRCh38
rs747073469 Chr.8: 11759595-11759595 on Build GRCh38
rs780176836 Chr.8: 11759624-11759624 on Build GRCh38
rs1053568436 Chr.8: 11759562-11759562 on Build GRCh38
rs565131524 Chr.8: 11759519-11759519 on Build GRCh38
rs146102045 Chr.8: 11759573-11759573 on Build GRCh38
rs569133968 Chr.8: 11759357-11759357 on Build GRCh38
rs970384118 Chr.8: 11759654-11759654 on Build GRCh38
rs555781562 Chr.8: 11759430-11759430 on Build GRCh38
rs549543886 Chr.8: 11759643-11759643 on Build GRCh38

View Full Product Details