Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11830151-11830698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDFT1
Entrez Gene ID
2222
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
DGPT, ERG9, SQS, SS
UniGene
Hs.593928
Gene Chromosome Location
Chr.8: 11795573-11839309 on Build GRCh38
Transcripts
NM_001287742.1 NM_001287743.1 NM_001287744.1 NM_001287745.1 NM_001287747.1 NM_001287748.1 NM_001287749.1 NM_001287750.1 NM_001287751.1 NM_001287756.1 NM_004462.4 XM_017013217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3164739 c.735delT p.F246fs*14 Chr.8: 11830276-11830276 on Build GRCh38
3164738 c.726G>T p.K242N Chr.8: 11830267-11830267 on Build GRCh38
7425257 c.751A>T p.N251Y Chr.8: 11830292-11830292 on Build GRCh38
5976607 c.775C>T p.L259L Chr.8: 11830316-11830316 on Build GRCh38
5873780 c.766G>A p.V256M Chr.8: 11830307-11830307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887463788 Chr.8: 11830563-11830563 on Build GRCh38
rs746271702 Chr.8: 11830358-11830358 on Build GRCh38
rs768326489 Chr.8: 11830275-11830275 on Build GRCh38
rs1046005664 Chr.8: 11830660-11830660 on Build GRCh38
rs375875514 Chr.8: 11830235-11830235 on Build GRCh38
rs780767601 Chr.8: 11830301-11830301 on Build GRCh38
rs377059690 Chr.8: 11830599-11830599 on Build GRCh38
rs528044182 Chr.8: 11830559-11830559 on Build GRCh38
rs763288176 Chr.8: 11830539-11830539 on Build GRCh38
rs75019066 Chr.8: 11830546-11830546 on Build GRCh38
rs79569566 Chr.8: 11830628-11830628 on Build GRCh38
rs754736055 Chr.8: 11830226-11830226 on Build GRCh38
rs776673503 Chr.8: 11830420-11830420 on Build GRCh38
rs562183536 Chr.8: 11830221-11830221 on Build GRCh38
rs977851929 Chr.8: 11830481-11830481 on Build GRCh38
rs746546484 Chr.8: 11830289-11830289 on Build GRCh38
rs762956536 Chr.8: 11830430-11830430 on Build GRCh38
rs1005772435 Chr.8: 11830466-11830466 on Build GRCh38
rs73207296 Chr.8: 11830414-11830414 on Build GRCh38
rs370010461 Chr.8: 11830209-11830209 on Build GRCh38
rs757283079 Chr.8: 11830640-11830640 on Build GRCh38
rs915871465 Chr.8: 11830652-11830652 on Build GRCh38
rs767233482 Chr.8: 11830385-11830385 on Build GRCh38
rs757547316 Chr.8: 11830465-11830465 on Build GRCh38
rs563016550 Chr.8: 11830282-11830282 on Build GRCh38
rs1017074355 Chr.8: 11830568-11830568 on Build GRCh38
rs140063318 Chr.8: 11830296-11830296 on Build GRCh38
rs2294137 Chr.8: 11830450-11830450 on Build GRCh38
rs982088584 Chr.8: 11830475-11830475 on Build GRCh38
rs769233178 Chr.8: 11830355-11830355 on Build GRCh38
rs776988972 Chr.8: 11830468-11830468 on Build GRCh38
rs964324352 Chr.8: 11830606-11830606 on Build GRCh38
rs192025259 Chr.8: 11830181-11830181 on Build GRCh38
rs765247257 Chr.8: 11830373-11830373 on Build GRCh38
rs553048453 Chr.8: 11830554-11830554 on Build GRCh38
rs564455236 Chr.8: 11830203-11830203 on Build GRCh38
rs17153904 Chr.8: 11830489-11830489 on Build GRCh38
rs1051511506 Chr.8: 11830638-11830638 on Build GRCh38
rs142337685 Chr.8: 11830649-11830649 on Build GRCh38
rs781487529 Chr.8: 11830467-11830467 on Build GRCh38
rs753718991 Chr.8: 11830280-11830280 on Build GRCh38
rs78698440 Chr.8: 11830434-11830434 on Build GRCh38
rs150271048 Chr.8: 11830375-11830375 on Build GRCh38
rs763358396 Chr.8: 11830207-11830207 on Build GRCh38
rs568313214 Chr.8: 11830565-11830565 on Build GRCh38
rs1055163073 Chr.8: 11830323-11830323 on Build GRCh38
rs770570736 Chr.8: 11830341-11830342 on Build GRCh38
rs745961881 Chr.8: 11830424-11830424 on Build GRCh38
rs948595578 Chr.8: 11830516-11830516 on Build GRCh38
rs922717128 Chr.8: 11830479-11830479 on Build GRCh38
rs778078365 Chr.8: 11830394-11830394 on Build GRCh38
rs750525000 Chr.8: 11830273-11830273 on Build GRCh38
rs760115135 Chr.8: 11830197-11830197 on Build GRCh38
rs756896439 Chr.8: 11830351-11830351 on Build GRCh38
rs747225690 Chr.8: 11830365-11830365 on Build GRCh38
rs368325351 Chr.8: 11830359-11830359 on Build GRCh38
rs369706172 Chr.8: 11830300-11830300 on Build GRCh38
rs150179680 Chr.8: 11830345-11830345 on Build GRCh38
rs867323246 Chr.8: 11830619-11830619 on Build GRCh38
rs946763489 Chr.8: 11830525-11830525 on Build GRCh38
rs575734226 Chr.8: 11830184-11830184 on Build GRCh38
rs550711143 Chr.8: 11830515-11830515 on Build GRCh38
rs752555247 Chr.8: 11830337-11830337 on Build GRCh38
rs535588750 Chr.8: 11830177-11830177 on Build GRCh38
rs1029815357 Chr.8: 11830616-11830616 on Build GRCh38
rs759481784 Chr.8: 11830370-11830370 on Build GRCh38
rs367718004 Chr.8: 11830393-11830393 on Build GRCh38
rs770442846 Chr.8: 11830240-11830240 on Build GRCh38
rs959260923 Chr.8: 11830618-11830618 on Build GRCh38
rs570109609 Chr.8: 11830307-11830307 on Build GRCh38
rs1026478577 Chr.8: 11830586-11830586 on Build GRCh38
rs773358438 Chr.8: 11830325-11830325 on Build GRCh38
rs975599144 Chr.8: 11830478-11830478 on Build GRCh38
rs77318013 Chr.8: 11830567-11830567 on Build GRCh38
rs749987862 Chr.8: 11830439-11830439 on Build GRCh38
rs183344780 Chr.8: 11830560-11830560 on Build GRCh38
rs200193052 Chr.8: 11830210-11830210 on Build GRCh38
rs530566147 Chr.8: 11830513-11830513 on Build GRCh38
rs753067410 Chr.8: 11830528-11830528 on Build GRCh38
rs779248530 Chr.8: 11830356-11830356 on Build GRCh38
rs767547038 Chr.8: 11830270-11830270 on Build GRCh38
rs777100951 Chr.8: 11830329-11830329 on Build GRCh38
rs919193674 Chr.8: 11830254-11830254 on Build GRCh38
rs780820890 Chr.8: 11830352-11830352 on Build GRCh38
rs756951488 Chr.8: 11830381-11830381 on Build GRCh38
rs113954339 Chr.8: 11830598-11830598 on Build GRCh38
rs941705353 Chr.8: 11830233-11830233 on Build GRCh38
rs753837696 Chr.8: 11830332-11830332 on Build GRCh38
rs566753367 Chr.8: 11830654-11830654 on Build GRCh38
rs746422066 Chr.8: 11830236-11830236 on Build GRCh38
rs779791237 Chr.8: 11830200-11830200 on Build GRCh38
rs766401728 Chr.8: 11830659-11830659 on Build GRCh38
rs771550389 Chr.8: 11830195-11830195 on Build GRCh38
rs749812596 Chr.8: 11830306-11830306 on Build GRCh38
rs4841600 Chr.8: 11830639-11830639 on Build GRCh38
rs555978554 Chr.8: 11830464-11830464 on Build GRCh38
rs574173181 Chr.8: 11830471-11830471 on Build GRCh38
rs151238023 Chr.8: 11830311-11830311 on Build GRCh38
rs915799176 Chr.8: 11830502-11830502 on Build GRCh38
rs984277152 Chr.8: 11830266-11830266 on Build GRCh38
rs1042491355 Chr.8: 11830630-11830630 on Build GRCh38
rs137899191 Chr.8: 11830480-11830480 on Build GRCh38
rs757341476 Chr.8: 11830213-11830213 on Build GRCh38
rs776669194 Chr.8: 11830366-11830366 on Build GRCh38
rs747279017 Chr.8: 11830396-11830396 on Build GRCh38
rs761245301 Chr.8: 11830460-11830460 on Build GRCh38
rs993248318 Chr.8: 11830547-11830547 on Build GRCh38
rs924617761 Chr.8: 11830485-11830485 on Build GRCh38
rs997446418 Chr.8: 11830612-11830612 on Build GRCh38
rs779489540 Chr.8: 11830388-11830388 on Build GRCh38
rs370875302 Chr.8: 11830276-11830276 on Build GRCh38
rs749811488 Chr.8: 11830353-11830353 on Build GRCh38
rs765804226 Chr.8: 11830451-11830451 on Build GRCh38
rs772371816 Chr.8: 11830316-11830316 on Build GRCh38
rs780917785 Chr.8: 11830455-11830455 on Build GRCh38
rs762068425 Chr.8: 11830262-11830262 on Build GRCh38
rs1017173872 Chr.8: 11830601-11830601 on Build GRCh38
rs769055412 Chr.8: 11830249-11830249 on Build GRCh38
rs754785762 Chr.8: 11830281-11830281 on Build GRCh38
rs764033347 Chr.8: 11830431-11830431 on Build GRCh38
rs763412017 Chr.8: 11830244-11830244 on Build GRCh38
rs1054374778 Chr.8: 11830494-11830494 on Build GRCh38
rs540577739 Chr.8: 11830457-11830457 on Build GRCh38
rs777062389 Chr.8: 11830196-11830196 on Build GRCh38
rs1293313 Chr.8: 11830429-11830429 on Build GRCh38
rs762034617 Chr.8: 11830315-11830315 on Build GRCh38
rs146586993 Chr.8: 11830288-11830288 on Build GRCh38
rs541562716 Chr.8: 11830255-11830255 on Build GRCh38
rs374045749 Chr.8: 11830229-11830229 on Build GRCh38
rs371623501 Chr.8: 11830374-11830374 on Build GRCh38
rs753285291 Chr.8: 11830455-11830455 on Build GRCh38
rs760759849 Chr.8: 11830328-11830328 on Build GRCh38
rs202060379 Chr.8: 11830287-11830287 on Build GRCh38
rs748591914 Chr.8: 11830389-11830389 on Build GRCh38
rs774313830 Chr.8: 11830432-11830432 on Build GRCh38
rs934019461 Chr.8: 11830634-11830634 on Build GRCh38
rs902547404 Chr.8: 11830533-11830533 on Build GRCh38
rs773483339 Chr.8: 11830363-11830363 on Build GRCh38
rs570595272 Chr.8: 11830520-11830520 on Build GRCh38
rs747821827 Chr.8: 11830231-11830231 on Build GRCh38
rs994621343 Chr.8: 11830582-11830582 on Build GRCh38
rs756024887 Chr.8: 11830218-11830218 on Build GRCh38
rs950438097 Chr.8: 11830593-11830593 on Build GRCh38
rs201339661 Chr.8: 11830339-11830339 on Build GRCh38
rs767284179 Chr.8: 11830436-11830436 on Build GRCh38
rs1029792449 Chr.8: 11830188-11830188 on Build GRCh38
rs766310256 Chr.8: 11830278-11830278 on Build GRCh38
rs540916846 Chr.8: 11830215-11830215 on Build GRCh38
rs147848089 Chr.8: 11830387-11830387 on Build GRCh38
rs778203058 Chr.8: 11830460-11830460 on Build GRCh38
rs546186641 Chr.8: 11830190-11830190 on Build GRCh38
rs963129435 Chr.8: 11830573-11830573 on Build GRCh38
rs528416963 Chr.8: 11830330-11830330 on Build GRCh38
rs140885465 Chr.8: 11830335-11830335 on Build GRCh38
rs746820922 Chr.8: 11830251-11830252 on Build GRCh38
rs111283886 Chr.8: 11830428-11830429 on Build GRCh38
rs569067917 Chr.8: 11830631-11830631 on Build GRCh38

View Full Product Details