Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11838152-11838648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDFT1
Entrez Gene ID
2222
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
DGPT, ERG9, SQS, SS
UniGene
Hs.593928
Gene Chromosome Location
Chr.8: 11795573-11839309 on Build GRCh38
Transcripts
NM_001287742.1 NM_001287743.1 NM_001287744.1 NM_001287745.1 NM_001287747.1 NM_001287748.1 NM_001287749.1 NM_001287750.1 NM_001287751.1 NM_001287756.1 NM_004462.4 XM_017013217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6283250 c.1115C>T p.P372L Chr.8: 11838470-11838470 on Build GRCh38
5184359 c.1099C>A p.R367R Chr.8: 11838454-11838454 on Build GRCh38
6260283 c.1164G>T p.S388S Chr.8: 11838519-11838519 on Build GRCh38
7303130 c.1051G>A p.D351N Chr.8: 11838406-11838406 on Build GRCh38
3748947 c.1055C>T p.S352L Chr.8: 11838410-11838410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029344031 Chr.8: 11838347-11838347 on Build GRCh38
rs775446844 Chr.8: 11838475-11838475 on Build GRCh38
rs772766586 Chr.8: 11838269-11838271 on Build GRCh38
rs745911401 Chr.8: 11838519-11838519 on Build GRCh38
rs1024349943 Chr.8: 11838180-11838180 on Build GRCh38
rs373360995 Chr.8: 11838455-11838455 on Build GRCh38
rs572243364 Chr.8: 11838406-11838406 on Build GRCh38
rs62495675 Chr.8: 11838226-11838226 on Build GRCh38
rs369149569 Chr.8: 11838304-11838304 on Build GRCh38
rs1037298400 Chr.8: 11838257-11838257 on Build GRCh38
rs376098828 Chr.8: 11838611-11838611 on Build GRCh38
rs144392308 Chr.8: 11838191-11838191 on Build GRCh38
rs950146538 Chr.8: 11838279-11838279 on Build GRCh38
rs527379865 Chr.8: 11838283-11838283 on Build GRCh38
rs376391117 Chr.8: 11838479-11838479 on Build GRCh38
rs780359576 Chr.8: 11838409-11838409 on Build GRCh38
rs751852979 Chr.8: 11838568-11838568 on Build GRCh38
rs1041965347 Chr.8: 11838534-11838534 on Build GRCh38
rs1010913317 Chr.8: 11838427-11838427 on Build GRCh38
rs554020851 Chr.8: 11838188-11838188 on Build GRCh38
rs936567324 Chr.8: 11838438-11838438 on Build GRCh38
rs773305066 Chr.8: 11838430-11838430 on Build GRCh38
rs757575333 Chr.8: 11838613-11838613 on Build GRCh38
rs80310078 Chr.8: 11838606-11838606 on Build GRCh38
rs984934049 Chr.8: 11838239-11838239 on Build GRCh38
rs751246035 Chr.8: 11838483-11838483 on Build GRCh38
rs754659198 Chr.8: 11838391-11838391 on Build GRCh38
rs981300579 Chr.8: 11838495-11838495 on Build GRCh38
rs146112670 Chr.8: 11838488-11838488 on Build GRCh38
rs148786056 Chr.8: 11838248-11838248 on Build GRCh38
rs764954065 Chr.8: 11838395-11838395 on Build GRCh38
rs565608299 Chr.8: 11838451-11838451 on Build GRCh38
rs764102515 Chr.8: 11838365-11838366 on Build GRCh38
rs10089896 Chr.8: 11838552-11838552 on Build GRCh38
rs75684569 Chr.8: 11838504-11838504 on Build GRCh38
rs781251270 Chr.8: 11838405-11838405 on Build GRCh38
rs1010468874 Chr.8: 11838312-11838312 on Build GRCh38
rs894895604 Chr.8: 11838319-11838319 on Build GRCh38
rs962708897 Chr.8: 11838623-11838623 on Build GRCh38
rs374221730 Chr.8: 11838540-11838540 on Build GRCh38
rs771646816 Chr.8: 11838364-11838364 on Build GRCh38
rs932281441 Chr.8: 11838228-11838228 on Build GRCh38
rs146404720 Chr.8: 11838453-11838453 on Build GRCh38
rs755672538 Chr.8: 11838458-11838458 on Build GRCh38
rs755037036 Chr.8: 11838363-11838363 on Build GRCh38
rs769935220 Chr.8: 11838572-11838572 on Build GRCh38
rs936496910 Chr.8: 11838309-11838309 on Build GRCh38
rs1055368069 Chr.8: 11838440-11838440 on Build GRCh38
rs925182892 Chr.8: 11838429-11838429 on Build GRCh38
rs559977336 Chr.8: 11838490-11838490 on Build GRCh38
rs760167733 Chr.8: 11838548-11838548 on Build GRCh38
rs923301469 Chr.8: 11838415-11838415 on Build GRCh38
rs966149763 Chr.8: 11838322-11838322 on Build GRCh38
rs1046214777 Chr.8: 11838529-11838529 on Build GRCh38
rs896982153 Chr.8: 11838273-11838273 on Build GRCh38
rs756665662 Chr.8: 11838447-11838447 on Build GRCh38
rs150943942 Chr.8: 11838431-11838431 on Build GRCh38
rs749937003 Chr.8: 11838546-11838546 on Build GRCh38
rs556396786 Chr.8: 11838308-11838308 on Build GRCh38
rs200967001 Chr.8: 11838560-11838560 on Build GRCh38
rs548744798 Chr.8: 11838292-11838292 on Build GRCh38
rs758044689 Chr.8: 11838402-11838402 on Build GRCh38
rs766907168 Chr.8: 11838487-11838487 on Build GRCh38
rs769678905 Chr.8: 11838474-11838474 on Build GRCh38
rs1046115192 Chr.8: 11838327-11838327 on Build GRCh38
rs762205085 Chr.8: 11838454-11838454 on Build GRCh38
rs10903343 Chr.8: 11838363-11838363 on Build GRCh38
rs549487939 Chr.8: 11838578-11838578 on Build GRCh38
rs761939298 Chr.8: 11838384-11838384 on Build GRCh38
rs748342272 Chr.8: 11838426-11838426 on Build GRCh38
rs1804473 Chr.8: 11838530-11838530 on Build GRCh38
rs553546119 Chr.8: 11838382-11838382 on Build GRCh38
rs181746559 Chr.8: 11838498-11838498 on Build GRCh38
rs571369651 Chr.8: 11838333-11838333 on Build GRCh38
rs746372741 Chr.8: 11838366-11838366 on Build GRCh38
rs745937609 Chr.8: 11838570-11838570 on Build GRCh38
rs763841409 Chr.8: 11838482-11838482 on Build GRCh38
rs748801966 Chr.8: 11838586-11838587 on Build GRCh38
rs763202642 Chr.8: 11838337-11838337 on Build GRCh38
rs375962292 Chr.8: 11838375-11838375 on Build GRCh38
rs758101680 Chr.8: 11838443-11838443 on Build GRCh38
rs755974027 Chr.8: 11838357-11838357 on Build GRCh38
rs202175915 Chr.8: 11838428-11838428 on Build GRCh38
rs780886966 Chr.8: 11838444-11838446 on Build GRCh38
rs749754215 Chr.8: 11838450-11838450 on Build GRCh38
rs200261638 Chr.8: 11838417-11838417 on Build GRCh38
rs1054511924 Chr.8: 11838505-11838505 on Build GRCh38
rs766342498 Chr.8: 11838441-11838441 on Build GRCh38
rs976060505 Chr.8: 11838255-11838255 on Build GRCh38
rs1054916364 Chr.8: 11838315-11838315 on Build GRCh38
rs189378553 Chr.8: 11838461-11838461 on Build GRCh38
rs758826568 Chr.8: 11838562-11838562 on Build GRCh38
rs755822020 Chr.8: 11838566-11838567 on Build GRCh38
rs766845955 Chr.8: 11838522-11838522 on Build GRCh38
rs781109427 Chr.8: 11838518-11838518 on Build GRCh38
rs768621368 Chr.8: 11838574-11838574 on Build GRCh38
rs747188802 Chr.8: 11838506-11838506 on Build GRCh38
rs776013215 Chr.8: 11838609-11838609 on Build GRCh38
rs191277119 Chr.8: 11838221-11838221 on Build GRCh38
rs780209110 Chr.8: 11838573-11838573 on Build GRCh38
rs1018204684 Chr.8: 11838463-11838463 on Build GRCh38
rs55774923 Chr.8: 11838222-11838222 on Build GRCh38
rs745651943 Chr.8: 11838503-11838503 on Build GRCh38
rs185046623 Chr.8: 11838282-11838282 on Build GRCh38
rs909764404 Chr.8: 11838242-11838242 on Build GRCh38
rs925267284 Chr.8: 11838507-11838507 on Build GRCh38
rs772866170 Chr.8: 11838604-11838604 on Build GRCh38
rs1001512569 Chr.8: 11838294-11838294 on Build GRCh38
rs376982798 Chr.8: 11838622-11838622 on Build GRCh38
rs771629673 Chr.8: 11838589-11838589 on Build GRCh38
rs755349901 Chr.8: 11838190-11838190 on Build GRCh38
rs777386277 Chr.8: 11838404-11838404 on Build GRCh38
rs773178092 Chr.8: 11838385-11838385 on Build GRCh38
rs759357216 Chr.8: 11838473-11838473 on Build GRCh38
rs759430126 Chr.8: 11838442-11838442 on Build GRCh38
rs142452672 Chr.8: 11838301-11838301 on Build GRCh38
rs773358775 Chr.8: 11838456-11838456 on Build GRCh38
rs751401059 Chr.8: 11838374-11838375 on Build GRCh38
rs779677068 Chr.8: 11838573-11838574 on Build GRCh38
rs547996073 Chr.8: 11838259-11838259 on Build GRCh38
rs750347717 Chr.8: 11838353-11838353 on Build GRCh38
rs757104710 Chr.8: 11838380-11838380 on Build GRCh38
rs753067078 Chr.8: 11838497-11838497 on Build GRCh38
rs767152311 Chr.8: 11838541-11838541 on Build GRCh38
rs371396760 Chr.8: 11838587-11838587 on Build GRCh38
rs368991167 Chr.8: 11838390-11838390 on Build GRCh38
rs952294162 Chr.8: 11838227-11838227 on Build GRCh38
rs373392280 Chr.8: 11838374-11838374 on Build GRCh38
rs769947378 Chr.8: 11838341-11838341 on Build GRCh38
rs139749047 Chr.8: 11838480-11838480 on Build GRCh38
rs761816508 Chr.8: 11838349-11838349 on Build GRCh38
rs754341871 Chr.8: 11838523-11838523 on Build GRCh38
rs1013302694 Chr.8: 11838320-11838320 on Build GRCh38
rs763789568 Chr.8: 11838444-11838444 on Build GRCh38
rs772237955 Chr.8: 11838210-11838210 on Build GRCh38
rs976474009 Chr.8: 11838328-11838328 on Build GRCh38
rs750654067 Chr.8: 11838621-11838621 on Build GRCh38
rs757441747 Chr.8: 11838569-11838569 on Build GRCh38
rs773297012 Chr.8: 11838216-11838216 on Build GRCh38
rs751693384 Chr.8: 11838348-11838348 on Build GRCh38
rs780670590 Chr.8: 11838448-11838448 on Build GRCh38
rs779354465 Chr.8: 11838491-11838491 on Build GRCh38
rs567215246 Chr.8: 11838299-11838299 on Build GRCh38
rs762794713 Chr.8: 11838476-11838476 on Build GRCh38
rs79708434 Chr.8: 11838422-11838422 on Build GRCh38
rs550400564 Chr.8: 11838225-11838225 on Build GRCh38
rs371833870 Chr.8: 11838350-11838350 on Build GRCh38
rs754034037 Chr.8: 11838361-11838362 on Build GRCh38
rs752380491 Chr.8: 11838399-11838399 on Build GRCh38
rs903653288 Chr.8: 11838185-11838185 on Build GRCh38
rs964834142 Chr.8: 11838468-11838468 on Build GRCh38
rs375494480 Chr.8: 11838359-11838359 on Build GRCh38
rs768928848 Chr.8: 11838379-11838379 on Build GRCh38
rs778076297 Chr.8: 11838564-11838564 on Build GRCh38
rs377634114 Chr.8: 11838542-11838542 on Build GRCh38
rs905863121 Chr.8: 11838285-11838285 on Build GRCh38
rs1013249004 Chr.8: 11838211-11838211 on Build GRCh38
rs761351505 Chr.8: 11838536-11838536 on Build GRCh38
rs763177462 Chr.8: 11838378-11838378 on Build GRCh38
rs1048556100 Chr.8: 11838598-11838598 on Build GRCh38
rs937395981 Chr.8: 11838419-11838419 on Build GRCh38
rs767645435 Chr.8: 11838620-11838620 on Build GRCh38
rs760588184 Chr.8: 11838387-11838387 on Build GRCh38
rs770899743 Chr.8: 11838459-11838459 on Build GRCh38
rs943424798 Chr.8: 11838252-11838252 on Build GRCh38
rs749678951 Chr.8: 11838412-11838412 on Build GRCh38
rs373559985 Chr.8: 11838278-11838278 on Build GRCh38
rs201618620 Chr.8: 11838526-11838526 on Build GRCh38

View Full Product Details