Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 13566710-13567216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf48
Entrez Gene ID
157773
Gene Name
chromosome 8 open reading frame 48
Gene Aliases
-
UniGene
Hs.104941
Gene Chromosome Location
Chr.8: 13566843-13568288 on Build GRCh38
Transcripts
NM_001007090.2 XM_017012951.1

Gene Symbol
DLC1
Entrez Gene ID
10395
Gene Name
DLC1 Rho GTPase activating protein
Gene Aliases
ARHGAP7, HP, STARD12, p122-RhoGAP
UniGene
Hs.134296
Gene Chromosome Location
Chr.8: 13083361-13604606 on Build GRCh38
Transcripts
NM_001007090.2 XM_017012951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1096356 c.89A>G p.D30G Chr.8: 13567080-13567080 on Build GRCh38
6223407 c.115A>G p.S39G Chr.8: 13567106-13567106 on Build GRCh38
1096355 c.80A>C p.N27T Chr.8: 13567071-13567071 on Build GRCh38
1455018 c.122C>A p.S41Y Chr.8: 13567113-13567113 on Build GRCh38
4532499 c.188G>A p.S63N Chr.8: 13567179-13567179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026145754 Chr.8: 13566791-13566791 on Build GRCh38
rs533273748 Chr.8: 13566922-13566922 on Build GRCh38
rs969880281 Chr.8: 13566778-13566778 on Build GRCh38
rs78230326 Chr.8: 13566774-13566774 on Build GRCh38
rs191812233 Chr.8: 13567128-13567128 on Build GRCh38
rs116418644 Chr.8: 13566977-13566977 on Build GRCh38
rs911488627 Chr.8: 13566946-13566946 on Build GRCh38
rs1054380187 Chr.8: 13567184-13567184 on Build GRCh38
rs769191641 Chr.8: 13567154-13567154 on Build GRCh38
rs1039543267 Chr.8: 13566776-13566777 on Build GRCh38
rs13281400 Chr.8: 13566878-13566878 on Build GRCh38
rs896470367 Chr.8: 13566770-13566770 on Build GRCh38
rs386722474 Chr.8: 13566876-13566878 on Build GRCh38
rs13273355 Chr.8: 13567074-13567074 on Build GRCh38
rs573686073 Chr.8: 13567092-13567092 on Build GRCh38
rs1022468635 Chr.8: 13566838-13566838 on Build GRCh38
rs10095899 Chr.8: 13566876-13566876 on Build GRCh38
rs994591374 Chr.8: 13566764-13566764 on Build GRCh38
rs775955635 Chr.8: 13567132-13567132 on Build GRCh38
rs957120854 Chr.8: 13566806-13566806 on Build GRCh38
rs767032573 Chr.8: 13567033-13567033 on Build GRCh38
rs1014209929 Chr.8: 13566771-13566771 on Build GRCh38
rs555375875 Chr.8: 13566753-13566753 on Build GRCh38
rs141944380 Chr.8: 13566746-13566746 on Build GRCh38
rs563038052 Chr.8: 13566908-13566908 on Build GRCh38
rs1056322344 Chr.8: 13566899-13566899 on Build GRCh38
rs551907924 Chr.8: 13566925-13566925 on Build GRCh38
rs999267114 Chr.8: 13566954-13566954 on Build GRCh38
rs757469783 Chr.8: 13566850-13566850 on Build GRCh38
rs532978997 Chr.8: 13567018-13567018 on Build GRCh38
rs1030735153 Chr.8: 13566976-13566976 on Build GRCh38
rs942881987 Chr.8: 13566735-13566735 on Build GRCh38
rs751163792 Chr.8: 13567038-13567038 on Build GRCh38
rs756125886 Chr.8: 13567137-13567137 on Build GRCh38
rs780137898 Chr.8: 13567143-13567144 on Build GRCh38
rs895030384 Chr.8: 13566901-13566901 on Build GRCh38
rs572275683 Chr.8: 13566832-13566832 on Build GRCh38
rs911437545 Chr.8: 13566830-13566830 on Build GRCh38
rs747398781 Chr.8: 13567141-13567141 on Build GRCh38
rs957417070 Chr.8: 13566912-13566912 on Build GRCh38
rs114054581 Chr.8: 13566880-13566880 on Build GRCh38
rs201250059 Chr.8: 13567056-13567056 on Build GRCh38
rs905173837 Chr.8: 13567040-13567040 on Build GRCh38
rs149488225 Chr.8: 13567093-13567093 on Build GRCh38
rs764570112 Chr.8: 13566847-13566847 on Build GRCh38
rs1012325882 Chr.8: 13566915-13566915 on Build GRCh38
rs898593485 Chr.8: 13566834-13566834 on Build GRCh38
rs562740146 Chr.8: 13567107-13567107 on Build GRCh38
rs999571127 Chr.8: 13567177-13567177 on Build GRCh38
rs920170626 Chr.8: 13567116-13567116 on Build GRCh38
rs370147338 Chr.8: 13566814-13566814 on Build GRCh38
rs537888309 Chr.8: 13567020-13567020 on Build GRCh38
rs955045718 Chr.8: 13566936-13566936 on Build GRCh38
rs995082732 Chr.8: 13566781-13566781 on Build GRCh38
rs918024192 Chr.8: 13567135-13567135 on Build GRCh38
rs934926011 Chr.8: 13566856-13566856 on Build GRCh38
rs62493966 Chr.8: 13566769-13566769 on Build GRCh38
rs188494481 Chr.8: 13567091-13567091 on Build GRCh38
rs533439710 Chr.8: 13567120-13567120 on Build GRCh38
rs930128355 Chr.8: 13566979-13566979 on Build GRCh38
rs949471852 Chr.8: 13567143-13567143 on Build GRCh38
rs1047218482 Chr.8: 13566984-13566984 on Build GRCh38
rs772729057 Chr.8: 13567007-13567007 on Build GRCh38
rs924858973 Chr.8: 13566742-13566742 on Build GRCh38
rs531048512 Chr.8: 13566799-13566799 on Build GRCh38
rs1048049210 Chr.8: 13566881-13566881 on Build GRCh38
rs964988020 Chr.8: 13567010-13567010 on Build GRCh38
rs1038866390 Chr.8: 13566743-13566743 on Build GRCh38
rs1010019850 Chr.8: 13566923-13566923 on Build GRCh38
rs886391422 Chr.8: 13566800-13566800 on Build GRCh38
rs1003306976 Chr.8: 13566894-13566894 on Build GRCh38
rs974385771 Chr.8: 13567114-13567114 on Build GRCh38
rs776972388 Chr.8: 13567179-13567179 on Build GRCh38
rs898548063 Chr.8: 13566760-13566760 on Build GRCh38
rs936664250 Chr.8: 13567178-13567178 on Build GRCh38
rs886253902 Chr.8: 13566893-13566893 on Build GRCh38
rs899215443 Chr.8: 13566779-13566779 on Build GRCh38
rs192314796 Chr.8: 13566941-13566941 on Build GRCh38
rs895104102 Chr.8: 13567105-13567105 on Build GRCh38
rs1013552353 Chr.8: 13566857-13566857 on Build GRCh38
rs560098538 Chr.8: 13566826-13566826 on Build GRCh38
rs1045157915 Chr.8: 13567151-13567151 on Build GRCh38
rs896524173 Chr.8: 13566855-13566855 on Build GRCh38
rs577760423 Chr.8: 13566813-13566813 on Build GRCh38
rs183770365 Chr.8: 13567025-13567025 on Build GRCh38
rs920136002 Chr.8: 13566978-13566978 on Build GRCh38
rs933648776 Chr.8: 13566823-13566823 on Build GRCh38
rs749094023 Chr.8: 13567122-13567122 on Build GRCh38
rs374822998 Chr.8: 13566844-13566844 on Build GRCh38
rs905749996 Chr.8: 13566877-13566877 on Build GRCh38
rs556322835 Chr.8: 13566801-13566801 on Build GRCh38
rs755998709 Chr.8: 13567090-13567090 on Build GRCh38
rs903538310 Chr.8: 13567151-13567155 on Build GRCh38
rs566976024 Chr.8: 13566932-13566932 on Build GRCh38
rs964306475 Chr.8: 13566974-13566974 on Build GRCh38
rs979924531 Chr.8: 13566785-13566785 on Build GRCh38
rs542490022 Chr.8: 13567075-13567075 on Build GRCh38
rs1038510323 Chr.8: 13566833-13566833 on Build GRCh38
rs553896592 Chr.8: 13567059-13567059 on Build GRCh38
rs967782176 Chr.8: 13566840-13566840 on Build GRCh38
rs577627190 Chr.8: 13567049-13567049 on Build GRCh38
rs1017919591 Chr.8: 13567099-13567099 on Build GRCh38
rs757281084 Chr.8: 13567098-13567098 on Build GRCh38
rs912926135 Chr.8: 13566815-13566815 on Build GRCh38

View Full Product Details