Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 49074112-49074635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf22
Entrez Gene ID
492307
Gene Name
chromosome 8 open reading frame 22
Gene Aliases
-
UniGene
Hs.49890
Gene Chromosome Location
Chr.8: 49054336-49076083 on Build GRCh38
Transcripts
NM_001007176.4 NM_001256596.1 NM_001256597.1 NM_001256598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4515699 c.124_125CC>TT p.P42L Chr.8: 49074127-49074128 on Build GRCh38
6258102 c.222G>T p.L74L Chr.8: 49074322-49074322 on Build GRCh38
4925066 c.229A>G p.T77A Chr.8: 49074329-49074329 on Build GRCh38
5409653 c.166T>C p.F56L Chr.8: 49074266-49074266 on Build GRCh38
6616686 c.117C>T p.D39D Chr.8: 49074120-49074120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746461742 Chr.8: 49074599-49074599 on Build GRCh38
rs892771692 Chr.8: 49074323-49074323 on Build GRCh38
rs563492382 Chr.8: 49074443-49074443 on Build GRCh38
rs931727250 Chr.8: 49074528-49074528 on Build GRCh38
rs767519218 Chr.8: 49074148-49074148 on Build GRCh38
rs528096043 Chr.8: 49074512-49074512 on Build GRCh38
rs555263510 Chr.8: 49074263-49074263 on Build GRCh38
rs1040607724 Chr.8: 49074517-49074517 on Build GRCh38
rs780685079 Chr.8: 49074511-49074511 on Build GRCh38
rs571283618 Chr.8: 49074602-49074602 on Build GRCh38
rs765608876 Chr.8: 49074383-49074383 on Build GRCh38
rs866942819 Chr.8: 49074601-49074601 on Build GRCh38
rs573279701 Chr.8: 49074290-49074290 on Build GRCh38
rs771565058 Chr.8: 49074358-49074358 on Build GRCh38
rs577250675 Chr.8: 49074410-49074410 on Build GRCh38
rs752054476 Chr.8: 49074484-49074484 on Build GRCh38
rs900293837 Chr.8: 49074525-49074525 on Build GRCh38
rs376197239 Chr.8: 49074153-49074153 on Build GRCh38
rs530751664 Chr.8: 49074446-49074446 on Build GRCh38
rs772708144 Chr.8: 49074176-49074176 on Build GRCh38
rs769401276 Chr.8: 49074334-49074334 on Build GRCh38
rs762898192 Chr.8: 49074145-49074145 on Build GRCh38
rs766363672 Chr.8: 49074225-49074225 on Build GRCh38
rs375049537 Chr.8: 49074461-49074461 on Build GRCh38
rs776385400 Chr.8: 49074199-49074199 on Build GRCh38
rs1035142416 Chr.8: 49074399-49074399 on Build GRCh38
rs868344933 Chr.8: 49074564-49074564 on Build GRCh38
rs565253124 Chr.8: 49074366-49074366 on Build GRCh38
rs543334863 Chr.8: 49074183-49074183 on Build GRCh38
rs759416628 Chr.8: 49074363-49074363 on Build GRCh38
rs775526686 Chr.8: 49074239-49074239 on Build GRCh38
rs772978147 Chr.8: 49074349-49074349 on Build GRCh38
rs771707690 Chr.8: 49074548-49074548 on Build GRCh38
rs140271227 Chr.8: 49074468-49074468 on Build GRCh38
rs370825990 Chr.8: 49074319-49074319 on Build GRCh38
rs754045329 Chr.8: 49074270-49074270 on Build GRCh38
rs748971827 Chr.8: 49074569-49074569 on Build GRCh38
rs761202841 Chr.8: 49074319-49074319 on Build GRCh38
rs755453216 Chr.8: 49074502-49074502 on Build GRCh38
rs144567681 Chr.8: 49074299-49074299 on Build GRCh38
rs192781367 Chr.8: 49074537-49074537 on Build GRCh38
rs756315470 Chr.8: 49074154-49074154 on Build GRCh38
rs1047605664 Chr.8: 49074151-49074151 on Build GRCh38
rs967952750 Chr.8: 49074472-49074472 on Build GRCh38
rs111811921 Chr.8: 49074423-49074423 on Build GRCh38
rs757437257 Chr.8: 49074282-49074282 on Build GRCh38
rs557509687 Chr.8: 49074164-49074164 on Build GRCh38
rs201836550 Chr.8: 49074428-49074428 on Build GRCh38
rs771065642 Chr.8: 49074163-49074163 on Build GRCh38
rs749132287 Chr.8: 49074351-49074351 on Build GRCh38
rs911906152 Chr.8: 49074235-49074235 on Build GRCh38
rs762875380 Chr.8: 49074220-49074220 on Build GRCh38
rs376597217 Chr.8: 49074558-49074559 on Build GRCh38
rs150329964 Chr.8: 49074604-49074604 on Build GRCh38
rs764700481 Chr.8: 49074562-49074580 on Build GRCh38
rs944715381 Chr.8: 49074544-49074544 on Build GRCh38
rs745539037 Chr.8: 49074493-49074493 on Build GRCh38
rs746485291 Chr.8: 49074305-49074305 on Build GRCh38
rs773002703 Chr.8: 49074215-49074215 on Build GRCh38
rs778106096 Chr.8: 49074162-49074162 on Build GRCh38
rs776581633 Chr.8: 49074367-49074367 on Build GRCh38
rs377607585 Chr.8: 49074265-49074265 on Build GRCh38
rs932079822 Chr.8: 49074194-49074194 on Build GRCh38
rs769580928 Chr.8: 49074212-49074212 on Build GRCh38
rs907766432 Chr.8: 49074508-49074508 on Build GRCh38
rs538898821 Chr.8: 49074136-49074136 on Build GRCh38
rs746571193 Chr.8: 49074186-49074186 on Build GRCh38
rs770825684 Chr.8: 49074360-49074360 on Build GRCh38
rs1049712102 Chr.8: 49074422-49074422 on Build GRCh38
rs774235125 Chr.8: 49074361-49074361 on Build GRCh38
rs778579098 Chr.8: 49074310-49074310 on Build GRCh38
rs921100937 Chr.8: 49074486-49074486 on Build GRCh38
rs780753198 Chr.8: 49074304-49074304 on Build GRCh38
rs750881589 Chr.8: 49074294-49074294 on Build GRCh38
rs762086573 Chr.8: 49074377-49074377 on Build GRCh38
rs986656888 Chr.8: 49074506-49074506 on Build GRCh38
rs1002369430 Chr.8: 49074435-49074435 on Build GRCh38
rs760849612 Chr.8: 49074254-49074254 on Build GRCh38
rs747788030 Chr.8: 49074206-49074206 on Build GRCh38
rs548874243 Chr.8: 49074464-49074464 on Build GRCh38
rs548245406 Chr.8: 49074559-49074559 on Build GRCh38
rs539126370 Chr.8: 49074557-49074558 on Build GRCh38
rs779444984 Chr.8: 49074568-49074568 on Build GRCh38
rs778695600 Chr.8: 49074581-49074581 on Build GRCh38
rs770666371 Chr.8: 49074577-49074577 on Build GRCh38
rs930893291 Chr.8: 49074600-49074600 on Build GRCh38
rs747777229 Chr.8: 49074330-49074330 on Build GRCh38
rs757606081 Chr.8: 49074165-49074165 on Build GRCh38
rs188592189 Chr.8: 49074175-49074175 on Build GRCh38
rs1009421397 Chr.8: 49074467-49074467 on Build GRCh38
rs1011174548 Chr.8: 49074327-49074327 on Build GRCh38
rs368669781 Chr.8: 49074559-49074560 on Build GRCh38

View Full Product Details