Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 49074425-49074932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf22
Entrez Gene ID
492307
Gene Name
chromosome 8 open reading frame 22
Gene Aliases
-
UniGene
Hs.49890
Gene Chromosome Location
Chr.8: 49054336-49076083 on Build GRCh38
Transcripts
NM_001007176.4 NM_001256596.1 NM_001256597.1 NM_001256598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787026 c.234-4G>A p.? Chr.8: 49074506-49074506 on Build GRCh38
5799533 c.234-3C>T p.? Chr.8: 49074507-49074507 on Build GRCh38
2718154 c.237G>A p.S79S Chr.8: 49074513-49074513 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746461742 Chr.8: 49074599-49074599 on Build GRCh38
rs966358714 Chr.8: 49074780-49074780 on Build GRCh38
rs1019009297 Chr.8: 49074819-49074819 on Build GRCh38
rs931727250 Chr.8: 49074528-49074528 on Build GRCh38
rs528096043 Chr.8: 49074512-49074512 on Build GRCh38
rs747995598 Chr.8: 49074679-49074679 on Build GRCh38
rs1040607724 Chr.8: 49074517-49074517 on Build GRCh38
rs768772351 Chr.8: 49074637-49074637 on Build GRCh38
rs571283618 Chr.8: 49074602-49074602 on Build GRCh38
rs780685079 Chr.8: 49074511-49074511 on Build GRCh38
rs866942819 Chr.8: 49074601-49074601 on Build GRCh38
rs569840667 Chr.8: 49074873-49074873 on Build GRCh38
rs752054476 Chr.8: 49074484-49074484 on Build GRCh38
rs900293837 Chr.8: 49074525-49074525 on Build GRCh38
rs931779554 Chr.8: 49074863-49074863 on Build GRCh38
rs375049537 Chr.8: 49074461-49074461 on Build GRCh38
rs1056166340 Chr.8: 49074647-49074647 on Build GRCh38
rs868344933 Chr.8: 49074564-49074564 on Build GRCh38
rs184862385 Chr.8: 49074820-49074820 on Build GRCh38
rs921773903 Chr.8: 49074855-49074855 on Build GRCh38
rs555326580 Chr.8: 49074779-49074779 on Build GRCh38
rs986217510 Chr.8: 49074800-49074800 on Build GRCh38
rs771707690 Chr.8: 49074548-49074548 on Build GRCh38
rs140271227 Chr.8: 49074468-49074468 on Build GRCh38
rs748971827 Chr.8: 49074569-49074569 on Build GRCh38
rs954035020 Chr.8: 49074799-49074799 on Build GRCh38
rs1020597321 Chr.8: 49074720-49074720 on Build GRCh38
rs985485805 Chr.8: 49074887-49074887 on Build GRCh38
rs755453216 Chr.8: 49074502-49074502 on Build GRCh38
rs192781367 Chr.8: 49074537-49074537 on Build GRCh38
rs1042670430 Chr.8: 49074663-49074663 on Build GRCh38
rs967952750 Chr.8: 49074472-49074472 on Build GRCh38
rs371508815 Chr.8: 49074901-49074901 on Build GRCh38
rs879607957 Chr.8: 49074694-49074694 on Build GRCh38
rs1035603171 Chr.8: 49074674-49074674 on Build GRCh38
rs972018114 Chr.8: 49074847-49074847 on Build GRCh38
rs1028152039 Chr.8: 49074795-49074795 on Build GRCh38
rs2292399 Chr.8: 49074671-49074671 on Build GRCh38
rs930901955 Chr.8: 49074884-49074884 on Build GRCh38
rs905393128 Chr.8: 49074633-49074633 on Build GRCh38
rs376597217 Chr.8: 49074558-49074559 on Build GRCh38
rs150329964 Chr.8: 49074604-49074604 on Build GRCh38
rs764700481 Chr.8: 49074562-49074580 on Build GRCh38
rs944715381 Chr.8: 49074544-49074544 on Build GRCh38
rs745539037 Chr.8: 49074493-49074493 on Build GRCh38
rs907766432 Chr.8: 49074508-49074508 on Build GRCh38
rs35402871 Chr.8: 49074886-49074887 on Build GRCh38
rs921100937 Chr.8: 49074486-49074486 on Build GRCh38
rs986656888 Chr.8: 49074506-49074506 on Build GRCh38
rs969066 Chr.8: 49074775-49074775 on Build GRCh38
rs983844009 Chr.8: 49074764-49074764 on Build GRCh38
rs952071824 Chr.8: 49074757-49074757 on Build GRCh38
rs1001201014 Chr.8: 49074728-49074728 on Build GRCh38
rs548874243 Chr.8: 49074464-49074464 on Build GRCh38
rs919267090 Chr.8: 49074882-49074882 on Build GRCh38
rs548245406 Chr.8: 49074559-49074559 on Build GRCh38
rs550934468 Chr.8: 49074638-49074638 on Build GRCh38
rs539126370 Chr.8: 49074557-49074558 on Build GRCh38
rs779444984 Chr.8: 49074568-49074568 on Build GRCh38
rs778695600 Chr.8: 49074581-49074581 on Build GRCh38
rs770666371 Chr.8: 49074577-49074577 on Build GRCh38
rs930893291 Chr.8: 49074600-49074600 on Build GRCh38
rs1027672375 Chr.8: 49074752-49074752 on Build GRCh38
rs1009421397 Chr.8: 49074467-49074467 on Build GRCh38
rs536798685 Chr.8: 49074721-49074721 on Build GRCh38
rs368669781 Chr.8: 49074559-49074560 on Build GRCh38

View Full Product Details