Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 52533939-52534466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM150A
Entrez Gene ID
389658
Gene Name
family with sequence similarity 150 member A
Gene Aliases
UNQ9433
UniGene
Hs.527111
Gene Chromosome Location
Chr.8: 52534037-52565461 on Build GRCh38
Transcripts
NM_207413.3

SNP Detail

SNP ID Chromosome Location
rs370023713 Chr.8: 52534134-52534136 on Build GRCh38
rs562197006 Chr.8: 52534301-52534301 on Build GRCh38
rs902811770 Chr.8: 52534007-52534007 on Build GRCh38
rs540546947 Chr.8: 52534051-52534051 on Build GRCh38
rs908962842 Chr.8: 52534042-52534042 on Build GRCh38
rs1053477193 Chr.8: 52534106-52534106 on Build GRCh38
rs529804728 Chr.8: 52534432-52534432 on Build GRCh38
rs952952383 Chr.8: 52534197-52534197 on Build GRCh38
rs1043114569 Chr.8: 52533992-52533992 on Build GRCh38
rs558312022 Chr.8: 52534090-52534090 on Build GRCh38
rs184204809 Chr.8: 52534019-52534019 on Build GRCh38
rs951378293 Chr.8: 52534195-52534195 on Build GRCh38
rs895556535 Chr.8: 52534394-52534394 on Build GRCh38
rs1026331655 Chr.8: 52534194-52534194 on Build GRCh38
rs905746304 Chr.8: 52534401-52534401 on Build GRCh38
rs912625480 Chr.8: 52534275-52534275 on Build GRCh38
rs1023918148 Chr.8: 52534073-52534073 on Build GRCh38
rs998570415 Chr.8: 52534008-52534008 on Build GRCh38
rs148648836 Chr.8: 52534130-52534132 on Build GRCh38
rs563568309 Chr.8: 52534151-52534151 on Build GRCh38
rs1021376276 Chr.8: 52534138-52534138 on Build GRCh38
rs1042946144 Chr.8: 52534343-52534343 on Build GRCh38
rs773916633 Chr.8: 52533986-52533986 on Build GRCh38
rs35937020 Chr.8: 52534074-52534075 on Build GRCh38
rs902861762 Chr.8: 52534345-52534345 on Build GRCh38
rs543233147 Chr.8: 52534047-52534047 on Build GRCh38
rs573024519 Chr.8: 52533997-52533997 on Build GRCh38
rs932996034 Chr.8: 52534316-52534316 on Build GRCh38
rs755419348 Chr.8: 52534010-52534010 on Build GRCh38
rs891710706 Chr.8: 52534390-52534390 on Build GRCh38
rs1027402247 Chr.8: 52534181-52534181 on Build GRCh38
rs921232234 Chr.8: 52534311-52534311 on Build GRCh38
rs1056832829 Chr.8: 52534386-52534386 on Build GRCh38
rs972729452 Chr.8: 52534193-52534193 on Build GRCh38
rs899737715 Chr.8: 52533981-52533981 on Build GRCh38
rs1052764285 Chr.8: 52534386-52534387 on Build GRCh38
rs901545220 Chr.8: 52534066-52534068 on Build GRCh38
rs1011383730 Chr.8: 52534130-52534130 on Build GRCh38
rs1001609803 Chr.8: 52534158-52534158 on Build GRCh38
rs115005342 Chr.8: 52534175-52534175 on Build GRCh38
rs934304988 Chr.8: 52534361-52534361 on Build GRCh38
rs982875781 Chr.8: 52534229-52534229 on Build GRCh38
rs188761070 Chr.8: 52534285-52534285 on Build GRCh38
rs947296161 Chr.8: 52534325-52534325 on Build GRCh38
rs1013741157 Chr.8: 52534421-52534421 on Build GRCh38
rs975238194 Chr.8: 52534304-52534304 on Build GRCh38
rs1042948834 Chr.8: 52534396-52534396 on Build GRCh38
rs577805820 Chr.8: 52533967-52533967 on Build GRCh38
rs984296170 Chr.8: 52534205-52534205 on Build GRCh38

View Full Product Details