Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58496472-58496997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3779293 c.803C>T p.S268F Chr.8: 58496709-58496709 on Build GRCh38
4676042 c.743A>G p.Q248R Chr.8: 58496769-58496769 on Build GRCh38
2719724 c.858G>A p.S286S Chr.8: 58496654-58496654 on Build GRCh38
6639499 c.634T>C p.F212L Chr.8: 58496878-58496878 on Build GRCh38
4607222 c.908+2T>A p.? Chr.8: 58496602-58496602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866523787 Chr.8: 58496953-58496956 on Build GRCh38
rs569910870 Chr.8: 58496595-58496595 on Build GRCh38
rs1041432159 Chr.8: 58496846-58496846 on Build GRCh38
rs370924818 Chr.8: 58496780-58496780 on Build GRCh38
rs192455108 Chr.8: 58496797-58496797 on Build GRCh38
rs778528720 Chr.8: 58496809-58496809 on Build GRCh38
rs751787597 Chr.8: 58496580-58496580 on Build GRCh38
rs565488904 Chr.8: 58496706-58496706 on Build GRCh38
rs922273358 Chr.8: 58496513-58496513 on Build GRCh38
rs776702193 Chr.8: 58496940-58496940 on Build GRCh38
rs747138282 Chr.8: 58496968-58496968 on Build GRCh38
rs925723960 Chr.8: 58496872-58496872 on Build GRCh38
rs201581713 Chr.8: 58496572-58496573 on Build GRCh38
rs778330127 Chr.8: 58496720-58496720 on Build GRCh38
rs756001088 Chr.8: 58496938-58496938 on Build GRCh38
rs151320685 Chr.8: 58496721-58496721 on Build GRCh38
rs942874526 Chr.8: 58496858-58496858 on Build GRCh38
rs765911554 Chr.8: 58496920-58496920 on Build GRCh38
rs780668040 Chr.8: 58496820-58496820 on Build GRCh38
rs386352295 Chr.8: 58496605-58496605 on Build GRCh38
rs757763336 Chr.8: 58496714-58496714 on Build GRCh38
rs8192874 Chr.8: 58496814-58496814 on Build GRCh38
rs560268189 Chr.8: 58496588-58496588 on Build GRCh38
rs746711199 Chr.8: 58496818-58496818 on Build GRCh38
rs767871705 Chr.8: 58496665-58496665 on Build GRCh38
rs748074785 Chr.8: 58496753-58496753 on Build GRCh38
rs747942969 Chr.8: 58496859-58496859 on Build GRCh38
rs1040725233 Chr.8: 58496511-58496511 on Build GRCh38
rs756548694 Chr.8: 58496686-58496686 on Build GRCh38
rs771901033 Chr.8: 58496929-58496929 on Build GRCh38
rs779616560 Chr.8: 58496553-58496553 on Build GRCh38
rs72647413 Chr.8: 58496934-58496934 on Build GRCh38
rs778199686 Chr.8: 58496694-58496694 on Build GRCh38
rs150039957 Chr.8: 58496763-58496763 on Build GRCh38
rs376522590 Chr.8: 58496769-58496769 on Build GRCh38
rs769413815 Chr.8: 58496828-58496828 on Build GRCh38
rs201236201 Chr.8: 58496744-58496744 on Build GRCh38
rs149183844 Chr.8: 58496533-58496538 on Build GRCh38
rs754627598 Chr.8: 58496819-58496819 on Build GRCh38
rs367814767 Chr.8: 58496792-58496792 on Build GRCh38
rs767634039 Chr.8: 58496765-58496765 on Build GRCh38
rs768382295 Chr.8: 58496634-58496634 on Build GRCh38
rs770963981 Chr.8: 58496953-58496953 on Build GRCh38
rs757525794 Chr.8: 58496788-58496788 on Build GRCh38
rs745559202 Chr.8: 58496610-58496610 on Build GRCh38
rs139396617 Chr.8: 58496733-58496733 on Build GRCh38
rs911358597 Chr.8: 58496834-58496834 on Build GRCh38
rs771684358 Chr.8: 58496623-58496623 on Build GRCh38
rs776499055 Chr.8: 58496641-58496641 on Build GRCh38
rs1041461475 Chr.8: 58496569-58496569 on Build GRCh38
rs771524490 Chr.8: 58496730-58496730 on Build GRCh38
rs942372487 Chr.8: 58496501-58496501 on Build GRCh38
rs141299456 Chr.8: 58496808-58496808 on Build GRCh38
rs895423346 Chr.8: 58496654-58496654 on Build GRCh38
rs531164077 Chr.8: 58496499-58496499 on Build GRCh38
rs761796743 Chr.8: 58496645-58496645 on Build GRCh38
rs776464588 Chr.8: 58496741-58496741 on Build GRCh38
rs778958664 Chr.8: 58496958-58496958 on Build GRCh38
rs760487952 Chr.8: 58496628-58496628 on Build GRCh38
rs902658512 Chr.8: 58496572-58496572 on Build GRCh38
rs528732489 Chr.8: 58496661-58496661 on Build GRCh38
rs187984940 Chr.8: 58496723-58496723 on Build GRCh38
rs142476981 Chr.8: 58496604-58496604 on Build GRCh38
rs567669521 Chr.8: 58496737-58496737 on Build GRCh38
rs201046553 Chr.8: 58496928-58496928 on Build GRCh38
rs1024942782 Chr.8: 58496697-58496697 on Build GRCh38
rs200100159 Chr.8: 58496742-58496742 on Build GRCh38
rs568918027 Chr.8: 58496893-58496893 on Build GRCh38
rs373514835 Chr.8: 58496598-58496598 on Build GRCh38
rs945764471 Chr.8: 58496563-58496563 on Build GRCh38
rs370087640 Chr.8: 58496876-58496876 on Build GRCh38
rs754464992 Chr.8: 58496870-58496871 on Build GRCh38
rs758879425 Chr.8: 58496912-58496912 on Build GRCh38
rs558718775 Chr.8: 58496891-58496891 on Build GRCh38
rs146594981 Chr.8: 58496790-58496790 on Build GRCh38
rs755226965 Chr.8: 58496581-58496581 on Build GRCh38
rs747994158 Chr.8: 58496821-58496821 on Build GRCh38
rs945734831 Chr.8: 58496842-58496842 on Build GRCh38
rs749290143 Chr.8: 58496944-58496944 on Build GRCh38
rs373964068 Chr.8: 58496784-58496784 on Build GRCh38
rs535428616 Chr.8: 58496624-58496624 on Build GRCh38
rs766417104 Chr.8: 58496655-58496655 on Build GRCh38
rs1026171025 Chr.8: 58496734-58496734 on Build GRCh38
rs753362618 Chr.8: 58496704-58496705 on Build GRCh38
rs749230759 Chr.8: 58496875-58496875 on Build GRCh38
rs762252165 Chr.8: 58496897-58496897 on Build GRCh38
rs761022978 Chr.8: 58496878-58496878 on Build GRCh38
rs530012264 Chr.8: 58496755-58496755 on Build GRCh38
rs765455964 Chr.8: 58496910-58496910 on Build GRCh38
rs930310389 Chr.8: 58496909-58496909 on Build GRCh38
rs1003369754 Chr.8: 58496621-58496621 on Build GRCh38
rs922242272 Chr.8: 58496839-58496839 on Build GRCh38
rs374585203 Chr.8: 58496931-58496931 on Build GRCh38
rs545609682 Chr.8: 58496672-58496672 on Build GRCh38
rs773285427 Chr.8: 58496874-58496874 on Build GRCh38
rs750996469 Chr.8: 58496911-58496911 on Build GRCh38
rs753140939 Chr.8: 58496587-58496587 on Build GRCh38
rs760969537 Chr.8: 58496778-58496778 on Build GRCh38

View Full Product Details