Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58498101-58498608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
751104 c.124C>A p.L42M Chr.8: 58498426-58498426 on Build GRCh38
1100722 c.145G>A p.G49S Chr.8: 58498405-58498405 on Build GRCh38
7363581 c.127G>T p.G43C Chr.8: 58498423-58498423 on Build GRCh38
6639498 c.87G>A p.T29T Chr.8: 58498463-58498463 on Build GRCh38
1100721 c.229C>A p.H77N Chr.8: 58498321-58498321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766241318 Chr.8: 58498237-58498237 on Build GRCh38
rs772489091 Chr.8: 58498208-58498208 on Build GRCh38
rs201412261 Chr.8: 58498244-58498244 on Build GRCh38
rs138113674 Chr.8: 58498442-58498442 on Build GRCh38
rs765283240 Chr.8: 58498513-58498513 on Build GRCh38
rs777813512 Chr.8: 58498252-58498252 on Build GRCh38
rs141308879 Chr.8: 58498409-58498409 on Build GRCh38
rs753399056 Chr.8: 58498264-58498264 on Build GRCh38
rs7825751 Chr.8: 58498196-58498196 on Build GRCh38
rs757315114 Chr.8: 58498346-58498346 on Build GRCh38
rs921237347 Chr.8: 58498370-58498370 on Build GRCh38
rs189424028 Chr.8: 58498393-58498393 on Build GRCh38
rs201327280 Chr.8: 58498475-58498475 on Build GRCh38
rs113258453 Chr.8: 58498361-58498361 on Build GRCh38
rs377254635 Chr.8: 58498326-58498326 on Build GRCh38
rs756127511 Chr.8: 58498185-58498185 on Build GRCh38
rs772428979 Chr.8: 58498263-58498263 on Build GRCh38
rs759284212 Chr.8: 58498430-58498430 on Build GRCh38
rs761546326 Chr.8: 58498230-58498230 on Build GRCh38
rs983005287 Chr.8: 58498310-58498310 on Build GRCh38
rs750214616 Chr.8: 58498233-58498233 on Build GRCh38
rs779960617 Chr.8: 58498134-58498134 on Build GRCh38
rs761807364 Chr.8: 58498456-58498456 on Build GRCh38
rs199703290 Chr.8: 58498289-58498289 on Build GRCh38
rs576051298 Chr.8: 58498222-58498222 on Build GRCh38
rs747673005 Chr.8: 58498390-58498390 on Build GRCh38
rs779020982 Chr.8: 58498352-58498352 on Build GRCh38
rs748660005 Chr.8: 58498480-58498480 on Build GRCh38
rs535212546 Chr.8: 58498545-58498545 on Build GRCh38
rs771792299 Chr.8: 58498499-58498499 on Build GRCh38
rs143198855 Chr.8: 58498347-58498347 on Build GRCh38
rs746243515 Chr.8: 58498262-58498262 on Build GRCh38
rs750486706 Chr.8: 58498518-58498518 on Build GRCh38
rs750668958 Chr.8: 58498463-58498463 on Build GRCh38
rs375792484 Chr.8: 58498485-58498485 on Build GRCh38
rs555015632 Chr.8: 58498574-58498574 on Build GRCh38
rs759343375 Chr.8: 58498494-58498494 on Build GRCh38
rs569592628 Chr.8: 58498502-58498502 on Build GRCh38
rs763632176 Chr.8: 58498226-58498226 on Build GRCh38
rs770645008 Chr.8: 58498487-58498487 on Build GRCh38
rs775869319 Chr.8: 58498276-58498276 on Build GRCh38
rs750595419 Chr.8: 58498357-58498357 on Build GRCh38
rs762196592 Chr.8: 58498533-58498533 on Build GRCh38
rs753952585 Chr.8: 58498332-58498332 on Build GRCh38
rs2162459 Chr.8: 58498483-58498483 on Build GRCh38
rs886044571 Chr.8: 58498479-58498479 on Build GRCh38
rs755335090 Chr.8: 58498473-58498473 on Build GRCh38
rs754562223 Chr.8: 58498462-58498462 on Build GRCh38
rs200836103 Chr.8: 58498251-58498251 on Build GRCh38
rs969443490 Chr.8: 58498381-58498381 on Build GRCh38
rs751553125 Chr.8: 58498320-58498320 on Build GRCh38
rs762661210 Chr.8: 58498315-58498315 on Build GRCh38
rs780606427 Chr.8: 58498464-58498464 on Build GRCh38
rs192803612 Chr.8: 58498205-58498205 on Build GRCh38
rs757436505 Chr.8: 58498254-58498254 on Build GRCh38
rs931298152 Chr.8: 58498517-58498517 on Build GRCh38
rs760278695 Chr.8: 58498224-58498224 on Build GRCh38
rs906849086 Chr.8: 58498491-58498491 on Build GRCh38
rs150351766 Chr.8: 58498427-58498427 on Build GRCh38
rs752006313 Chr.8: 58498468-58498468 on Build GRCh38
rs545869846 Chr.8: 58498296-58498296 on Build GRCh38
rs771340637 Chr.8: 58498199-58498199 on Build GRCh38
rs200839929 Chr.8: 58498232-58498232 on Build GRCh38
rs563046069 Chr.8: 58498304-58498304 on Build GRCh38
rs539045136 Chr.8: 58498187-58498187 on Build GRCh38
rs767467704 Chr.8: 58498432-58498432 on Build GRCh38
rs899803163 Chr.8: 58498239-58498239 on Build GRCh38
rs60059260 Chr.8: 58498325-58498325 on Build GRCh38
rs762648850 Chr.8: 58498405-58498405 on Build GRCh38
rs780453940 Chr.8: 58498363-58498363 on Build GRCh38
rs777312596 Chr.8: 58498484-58498484 on Build GRCh38
rs1002567960 Chr.8: 58498508-58498508 on Build GRCh38
rs535484729 Chr.8: 58498229-58498229 on Build GRCh38
rs184906718 Chr.8: 58498253-58498253 on Build GRCh38
rs180916341 Chr.8: 58498404-58498404 on Build GRCh38
rs747570322 Chr.8: 58498372-58498372 on Build GRCh38
rs62621283 Chr.8: 58498294-58498294 on Build GRCh38
rs148278254 Chr.8: 58498388-58498388 on Build GRCh38
rs549512565 Chr.8: 58498492-58498492 on Build GRCh38
rs766189113 Chr.8: 58498318-58498318 on Build GRCh38
rs769256926 Chr.8: 58498377-58498377 on Build GRCh38
rs775477625 Chr.8: 58498500-58498500 on Build GRCh38
rs1026092643 Chr.8: 58498280-58498280 on Build GRCh38
rs747513533 Chr.8: 58498277-58498277 on Build GRCh38
rs746430425 Chr.8: 58498202-58498202 on Build GRCh38
rs267601959 Chr.8: 58498275-58498275 on Build GRCh38

View Full Product Details