Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58499807-58500332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874848 c.22T>G p.W8G Chr.8: 58500077-58500077 on Build GRCh38
5358823 c.38C>T p.A13V Chr.8: 58500061-58500061 on Build GRCh38
1249619 c.79A>C p.R27R Chr.8: 58500020-58500020 on Build GRCh38
4138721 c.50G>T p.C17F Chr.8: 58500049-58500049 on Build GRCh38
3900809 c.65T>C p.L22P Chr.8: 58500034-58500034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963226859 Chr.8: 58500182-58500182 on Build GRCh38
rs987894650 Chr.8: 58500303-58500303 on Build GRCh38
rs1015202964 Chr.8: 58500148-58500148 on Build GRCh38
rs61683973 Chr.8: 58499869-58499869 on Build GRCh38
rs768030841 Chr.8: 58500131-58500131 on Build GRCh38
rs111811966 Chr.8: 58500097-58500097 on Build GRCh38
rs533285898 Chr.8: 58499911-58499911 on Build GRCh38
rs371251420 Chr.8: 58500121-58500121 on Build GRCh38
rs980538720 Chr.8: 58499831-58499831 on Build GRCh38
rs372058087 Chr.8: 58499921-58499921 on Build GRCh38
rs944896206 Chr.8: 58499915-58499915 on Build GRCh38
rs976692810 Chr.8: 58500279-58500279 on Build GRCh38
rs528066345 Chr.8: 58500074-58500074 on Build GRCh38
rs773606282 Chr.8: 58500247-58500247 on Build GRCh38
rs1028838884 Chr.8: 58500196-58500196 on Build GRCh38
rs778099251 Chr.8: 58500047-58500047 on Build GRCh38
rs953057984 Chr.8: 58500217-58500217 on Build GRCh38
rs373181079 Chr.8: 58499978-58499978 on Build GRCh38
rs1004963084 Chr.8: 58500127-58500127 on Build GRCh38
rs755459892 Chr.8: 58499981-58499981 on Build GRCh38
rs571033590 Chr.8: 58500308-58500308 on Build GRCh38
rs911558515 Chr.8: 58500271-58500271 on Build GRCh38
rs376891373 Chr.8: 58500072-58500072 on Build GRCh38
rs966226876 Chr.8: 58500275-58500276 on Build GRCh38
rs769167524 Chr.8: 58500014-58500018 on Build GRCh38
rs973313394 Chr.8: 58500189-58500189 on Build GRCh38
rs564210114 Chr.8: 58499848-58499848 on Build GRCh38
rs771917069 Chr.8: 58500071-58500071 on Build GRCh38
rs989031155 Chr.8: 58499884-58499884 on Build GRCh38
rs973355162 Chr.8: 58499858-58499858 on Build GRCh38
rs762903208 Chr.8: 58499972-58499972 on Build GRCh38
rs375388089 Chr.8: 58500267-58500267 on Build GRCh38
rs911676383 Chr.8: 58499927-58499927 on Build GRCh38
rs921576715 Chr.8: 58499862-58499862 on Build GRCh38
rs763096344 Chr.8: 58500088-58500088 on Build GRCh38
rs1020518824 Chr.8: 58500273-58500273 on Build GRCh38
rs774771534 Chr.8: 58500112-58500112 on Build GRCh38
rs778544430 Chr.8: 58500062-58500062 on Build GRCh38
rs550994436 Chr.8: 58500183-58500183 on Build GRCh38
rs200846540 Chr.8: 58500019-58500019 on Build GRCh38
rs1014526226 Chr.8: 58500056-58500056 on Build GRCh38
rs147162838 Chr.8: 58500061-58500061 on Build GRCh38
rs745519772 Chr.8: 58500067-58500067 on Build GRCh38
rs748850568 Chr.8: 58500052-58500052 on Build GRCh38
rs375604247 Chr.8: 58499990-58499990 on Build GRCh38
rs371390410 Chr.8: 58499952-58499952 on Build GRCh38
rs756610472 Chr.8: 58500045-58500045 on Build GRCh38
rs1049902632 Chr.8: 58499965-58499965 on Build GRCh38
rs896965456 Chr.8: 58500012-58500012 on Build GRCh38
rs935109473 Chr.8: 58500290-58500290 on Build GRCh38
rs8192869 Chr.8: 58500267-58500267 on Build GRCh38
rs772451119 Chr.8: 58500087-58500087 on Build GRCh38
rs932328821 Chr.8: 58499958-58499958 on Build GRCh38
rs895846752 Chr.8: 58500125-58500125 on Build GRCh38
rs753133818 Chr.8: 58500150-58500150 on Build GRCh38
rs1051446990 Chr.8: 58499924-58499924 on Build GRCh38
rs766561777 Chr.8: 58500110-58500110 on Build GRCh38
rs556059266 Chr.8: 58500264-58500264 on Build GRCh38
rs963589952 Chr.8: 58499839-58499839 on Build GRCh38
rs987058301 Chr.8: 58500230-58500230 on Build GRCh38

View Full Product Details