Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60188778-60189277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA8
Entrez Gene ID
767
Gene Name
carbonic anhydrase 8
Gene Aliases
CA-RP, CA-VIII, CALS, CAMRQ3, CARP
UniGene
Hs.654388
Gene Chromosome Location
Chr.8: 60185420-60281423 on Build GRCh38
Transcripts
NM_001321839.1 NM_004056.5 NR_135821.1 XM_017013818.1

SNP Detail

SNP ID Chromosome Location
rs3140 Chr.8: 60189151-60189151 on Build GRCh38
rs978078720 Chr.8: 60188853-60188853 on Build GRCh38
rs906352259 Chr.8: 60188972-60188972 on Build GRCh38
rs187722285 Chr.8: 60189230-60189230 on Build GRCh38
rs965272092 Chr.8: 60189117-60189117 on Build GRCh38
rs560622576 Chr.8: 60188886-60188888 on Build GRCh38
rs907130695 Chr.8: 60189023-60189023 on Build GRCh38
rs745829696 Chr.8: 60189060-60189060 on Build GRCh38
rs910307788 Chr.8: 60189178-60189178 on Build GRCh38
rs923531451 Chr.8: 60188834-60188834 on Build GRCh38
rs1002981997 Chr.8: 60188977-60188982 on Build GRCh38
rs1030889613 Chr.8: 60189163-60189163 on Build GRCh38
rs572817985 Chr.8: 60189237-60189237 on Build GRCh38
rs555094000 Chr.8: 60189209-60189209 on Build GRCh38
rs574427596 Chr.8: 60188922-60188922 on Build GRCh38
rs925294345 Chr.8: 60189197-60189197 on Build GRCh38
rs1034431443 Chr.8: 60188980-60188982 on Build GRCh38
rs954859327 Chr.8: 60189133-60189133 on Build GRCh38
rs941742613 Chr.8: 60189186-60189186 on Build GRCh38
rs1047430635 Chr.8: 60189061-60189061 on Build GRCh38
rs887406060 Chr.8: 60189075-60189075 on Build GRCh38
rs140836397 Chr.8: 60188924-60188924 on Build GRCh38
rs910243596 Chr.8: 60188817-60188817 on Build GRCh38
rs1011592845 Chr.8: 60188998-60188998 on Build GRCh38
rs142386673 Chr.8: 60189247-60189247 on Build GRCh38
rs770984438 Chr.8: 60188995-60188995 on Build GRCh38
rs73683370 Chr.8: 60188824-60188824 on Build GRCh38
rs1055182282 Chr.8: 60188908-60188908 on Build GRCh38
rs77070861 Chr.8: 60189192-60189192 on Build GRCh38
rs991352919 Chr.8: 60189235-60189235 on Build GRCh38
rs1046088481 Chr.8: 60188996-60188996 on Build GRCh38
rs915764668 Chr.8: 60188944-60188944 on Build GRCh38
rs567190198 Chr.8: 60188840-60188840 on Build GRCh38
rs993403738 Chr.8: 60188865-60188865 on Build GRCh38
rs978274630 Chr.8: 60189195-60189195 on Build GRCh38
rs1017350256 Chr.8: 60189113-60189113 on Build GRCh38
rs1046671902 Chr.8: 60188882-60188882 on Build GRCh38
rs555692295 Chr.8: 60188917-60188917 on Build GRCh38
rs1006008197 Chr.8: 60189076-60189076 on Build GRCh38
rs558465687 Chr.8: 60188836-60188836 on Build GRCh38
rs923465439 Chr.8: 60189121-60189121 on Build GRCh38
rs906398417 Chr.8: 60189196-60189196 on Build GRCh38
rs781048307 Chr.8: 60189054-60189054 on Build GRCh38
rs534365904 Chr.8: 60188859-60188859 on Build GRCh38
rs936761370 Chr.8: 60188899-60188899 on Build GRCh38
rs958025532 Chr.8: 60189222-60189222 on Build GRCh38
rs182200309 Chr.8: 60188990-60188990 on Build GRCh38

View Full Product Details