Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 60279587-60280099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA8
Entrez Gene ID
767
Gene Name
carbonic anhydrase 8
Gene Aliases
CA-RP, CA-VIII, CALS, CAMRQ3, CARP
UniGene
Hs.654388
Gene Chromosome Location
Chr.8: 60185420-60281423 on Build GRCh38
Transcripts
NM_001321837.1 NM_001321838.1 NM_001321839.1 NM_004056.5 NR_135821.1 XM_011517587.2 XM_011517588.2 XM_017013818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5794041 c.212C>T p.S71F Chr.8: 60279769-60279769 on Build GRCh38
3900844 c.192G>A p.S64S Chr.8: 60279789-60279789 on Build GRCh38
5629189 c.167C>G p.S56* Chr.8: 60279814-60279814 on Build GRCh38
3900843 c.251A>G p.N84S Chr.8: 60279730-60279730 on Build GRCh38
7242904 c.276C>A p.I92I Chr.8: 60279705-60279705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144915033 Chr.8: 60279699-60279699 on Build GRCh38
rs778724631 Chr.8: 60279842-60279842 on Build GRCh38
rs767300932 Chr.8: 60279666-60279666 on Build GRCh38
rs374430594 Chr.8: 60279776-60279776 on Build GRCh38
rs755803333 Chr.8: 60279800-60279800 on Build GRCh38
rs781229474 Chr.8: 60279869-60279869 on Build GRCh38
rs777655754 Chr.8: 60279679-60279679 on Build GRCh38
rs780227062 Chr.8: 60279726-60279726 on Build GRCh38
rs757956541 Chr.8: 60280038-60280060 on Build GRCh38
rs752417587 Chr.8: 60279794-60279794 on Build GRCh38
rs547419049 Chr.8: 60280043-60280043 on Build GRCh38
rs142471870 Chr.8: 60279775-60279775 on Build GRCh38
rs763353966 Chr.8: 60279641-60279641 on Build GRCh38
rs770200174 Chr.8: 60279764-60279764 on Build GRCh38
rs748309428 Chr.8: 60279755-60279755 on Build GRCh38
rs769999610 Chr.8: 60279912-60279912 on Build GRCh38
rs1028494750 Chr.8: 60279827-60279827 on Build GRCh38
rs532104510 Chr.8: 60280016-60280016 on Build GRCh38
rs760002973 Chr.8: 60279968-60279968 on Build GRCh38
rs767070057 Chr.8: 60279788-60279788 on Build GRCh38
rs912501757 Chr.8: 60279638-60279638 on Build GRCh38
rs898205495 Chr.8: 60280066-60280066 on Build GRCh38
rs1037248115 Chr.8: 60279732-60279732 on Build GRCh38
rs191276716 Chr.8: 60279953-60279953 on Build GRCh38
rs896125931 Chr.8: 60279689-60279689 on Build GRCh38
rs768165240 Chr.8: 60279965-60279965 on Build GRCh38
rs1032432994 Chr.8: 60280031-60280031 on Build GRCh38
rs917662295 Chr.8: 60280001-60280001 on Build GRCh38
rs55948763 Chr.8: 60279983-60279983 on Build GRCh38
rs771310005 Chr.8: 60279874-60279874 on Build GRCh38
rs377244093 Chr.8: 60279674-60279674 on Build GRCh38
rs28429093 Chr.8: 60279975-60279975 on Build GRCh38
rs747077440 Chr.8: 60279728-60279728 on Build GRCh38
rs937855196 Chr.8: 60279998-60279998 on Build GRCh38
rs754709169 Chr.8: 60279660-60279660 on Build GRCh38
rs776839353 Chr.8: 60279751-60279751 on Build GRCh38
rs184481078 Chr.8: 60280045-60280045 on Build GRCh38
rs988773087 Chr.8: 60279974-60279974 on Build GRCh38
rs751002003 Chr.8: 60279647-60279647 on Build GRCh38
rs528594297 Chr.8: 60279972-60279972 on Build GRCh38
rs757024364 Chr.8: 60279807-60279807 on Build GRCh38
rs889976787 Chr.8: 60279843-60279843 on Build GRCh38
rs768738585 Chr.8: 60279730-60279730 on Build GRCh38
rs762520438 Chr.8: 60279931-60279931 on Build GRCh38
rs926464505 Chr.8: 60279993-60279993 on Build GRCh38
rs748411544 Chr.8: 60279883-60279883 on Build GRCh38
rs541432962 Chr.8: 60280000-60280000 on Build GRCh38
rs745780775 Chr.8: 60279844-60279844 on Build GRCh38
rs149029428 Chr.8: 60279710-60279710 on Build GRCh38
rs749308896 Chr.8: 60279680-60279680 on Build GRCh38
rs553201107 Chr.8: 60279670-60279670 on Build GRCh38
rs369675880 Chr.8: 60279642-60279642 on Build GRCh38
rs114775598 Chr.8: 60279618-60279618 on Build GRCh38
rs1000919784 Chr.8: 60280010-60280010 on Build GRCh38
rs144653268 Chr.8: 60279966-60279966 on Build GRCh38
rs956505262 Chr.8: 60280037-60280040 on Build GRCh38
rs867958016 Chr.8: 60279725-60279725 on Build GRCh38
rs774872914 Chr.8: 60279777-60279777 on Build GRCh38
rs931537503 Chr.8: 60280067-60280067 on Build GRCh38
rs773560890 Chr.8: 60279769-60279769 on Build GRCh38
rs764017237 Chr.8: 60279802-60279802 on Build GRCh38
rs759861082 Chr.8: 60279856-60279856 on Build GRCh38
rs967928895 Chr.8: 60279980-60279980 on Build GRCh38

View Full Product Details