Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 68439104-68439627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf34
Entrez Gene ID
116328
Gene Name
chromosome 8 open reading frame 34
Gene Aliases
VEST-1, VEST1
UniGene
Hs.491941
Gene Chromosome Location
Chr.8: 68330722-68819023 on Build GRCh38
Transcripts
NM_001195639.1 NM_052958.2 XM_011517445.2 XM_011517446.2 XM_011517448.2 XM_011517449.2 XM_011517450.2 XM_017013019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4547668 c.170G>A p.R57K Chr.8: 68439599-68439599 on Build GRCh38
486625 c.197G>A p.W66* Chr.8: 68439626-68439626 on Build GRCh38
6181096 c.127G>A p.D43N Chr.8: 68439556-68439556 on Build GRCh38
5156495 c.151A>T p.N51Y Chr.8: 68439580-68439580 on Build GRCh38
2874215 c.107A>G p.Q36R Chr.8: 68439536-68439536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751846808 Chr.8: 68439583-68439583 on Build GRCh38
rs899107321 Chr.8: 68439505-68439505 on Build GRCh38
rs759635250 Chr.8: 68439584-68439584 on Build GRCh38
rs971385816 Chr.8: 68439477-68439477 on Build GRCh38
rs73260794 Chr.8: 68439411-68439411 on Build GRCh38
rs749904423 Chr.8: 68439170-68439170 on Build GRCh38
rs779415019 Chr.8: 68439298-68439298 on Build GRCh38
rs34403530 Chr.8: 68439576-68439577 on Build GRCh38
rs935181361 Chr.8: 68439527-68439527 on Build GRCh38
rs892573891 Chr.8: 68439175-68439175 on Build GRCh38
rs771191923 Chr.8: 68439529-68439529 on Build GRCh38
rs1005693143 Chr.8: 68439396-68439396 on Build GRCh38
rs1024353291 Chr.8: 68439425-68439425 on Build GRCh38
rs767654979 Chr.8: 68439585-68439585 on Build GRCh38
rs935144757 Chr.8: 68439267-68439267 on Build GRCh38
rs766690262 Chr.8: 68439449-68439449 on Build GRCh38
rs1052941079 Chr.8: 68439161-68439161 on Build GRCh38
rs564459048 Chr.8: 68439412-68439412 on Build GRCh38
rs556662095 Chr.8: 68439333-68439333 on Build GRCh38
rs963534377 Chr.8: 68439278-68439278 on Build GRCh38
rs867888447 Chr.8: 68439293-68439293 on Build GRCh38
rs77327351 Chr.8: 68439397-68439397 on Build GRCh38
rs764917193 Chr.8: 68439582-68439582 on Build GRCh38
rs756689865 Chr.8: 68439595-68439595 on Build GRCh38
rs761655185 Chr.8: 68439577-68439577 on Build GRCh38
rs971963388 Chr.8: 68439280-68439280 on Build GRCh38
rs542167755 Chr.8: 68439395-68439395 on Build GRCh38
rs774816194 Chr.8: 68439537-68439537 on Build GRCh38
rs756638564 Chr.8: 68439512-68439512 on Build GRCh38
rs981398659 Chr.8: 68439354-68439354 on Build GRCh38
rs757990389 Chr.8: 68439258-68439258 on Build GRCh38
rs781099468 Chr.8: 68439480-68439480 on Build GRCh38
rs573374937 Chr.8: 68439390-68439390 on Build GRCh38
rs942657916 Chr.8: 68439561-68439561 on Build GRCh38
rs752764833 Chr.8: 68439586-68439586 on Build GRCh38
rs749594943 Chr.8: 68439528-68439528 on Build GRCh38
rs925592165 Chr.8: 68439365-68439365 on Build GRCh38
rs950215399 Chr.8: 68439252-68439252 on Build GRCh38
rs1054126439 Chr.8: 68439277-68439277 on Build GRCh38
rs576896094 Chr.8: 68439350-68439350 on Build GRCh38
rs547068572 Chr.8: 68439479-68439479 on Build GRCh38
rs1007848519 Chr.8: 68439253-68439253 on Build GRCh38
rs936863506 Chr.8: 68439138-68439138 on Build GRCh38
rs915557234 Chr.8: 68439328-68439328 on Build GRCh38
rs992593458 Chr.8: 68439308-68439308 on Build GRCh38
rs533294481 Chr.8: 68439426-68439426 on Build GRCh38
rs765540930 Chr.8: 68439474-68439474 on Build GRCh38
rs867140234 Chr.8: 68439360-68439360 on Build GRCh38
rs900844207 Chr.8: 68439338-68439344 on Build GRCh38
rs530950266 Chr.8: 68439405-68439405 on Build GRCh38
rs1051797703 Chr.8: 68439428-68439428 on Build GRCh38
rs923773181 Chr.8: 68439257-68439257 on Build GRCh38
rs1017167763 Chr.8: 68439147-68439147 on Build GRCh38
rs959807908 Chr.8: 68439301-68439301 on Build GRCh38
rs997458816 Chr.8: 68439207-68439207 on Build GRCh38
rs760620872 Chr.8: 68439553-68439553 on Build GRCh38
rs893702534 Chr.8: 68439288-68439288 on Build GRCh38
rs778680320 Chr.8: 68439531-68439531 on Build GRCh38
rs1026053392 Chr.8: 68439300-68439300 on Build GRCh38
rs745482172 Chr.8: 68439533-68439533 on Build GRCh38
rs958711871 Chr.8: 68439168-68439168 on Build GRCh38
rs72664946 Chr.8: 68439241-68439241 on Build GRCh38
rs200035591 Chr.8: 68439485-68439485 on Build GRCh38
rs553336171 Chr.8: 68439380-68439380 on Build GRCh38
rs751718139 Chr.8: 68439451-68439451 on Build GRCh38
rs141345932 Chr.8: 68439567-68439567 on Build GRCh38
rs991512542 Chr.8: 68439186-68439186 on Build GRCh38
rs184740450 Chr.8: 68439351-68439351 on Build GRCh38
rs147842454 Chr.8: 68439554-68439554 on Build GRCh38
rs778486798 Chr.8: 68439597-68439597 on Build GRCh38
rs1030260230 Chr.8: 68439232-68439232 on Build GRCh38
rs754379412 Chr.8: 68439602-68439602 on Build GRCh38
rs778292497 Chr.8: 68439520-68439520 on Build GRCh38

View Full Product Details