Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 68533827-68534349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf34
Entrez Gene ID
116328
Gene Name
chromosome 8 open reading frame 34
Gene Aliases
VEST-1, VEST1
UniGene
Hs.491941
Gene Chromosome Location
Chr.8: 68330722-68819023 on Build GRCh38
Transcripts
NM_001195639.1 NM_052958.2 XM_011517445.2 XM_011517446.2 XM_011517448.2 XM_011517449.2 XM_011517450.2 XM_017013019.1

SNP Detail

SNP ID Chromosome Location
rs866605211 Chr.8: 68533985-68533985 on Build GRCh38
rs990989747 Chr.8: 68534179-68534179 on Build GRCh38
rs949816294 Chr.8: 68533962-68533962 on Build GRCh38
rs950602638 Chr.8: 68534125-68534125 on Build GRCh38
rs575085444 Chr.8: 68534308-68534308 on Build GRCh38
rs536297675 Chr.8: 68534245-68534245 on Build GRCh38
rs1030278891 Chr.8: 68534114-68534114 on Build GRCh38
rs999510144 Chr.8: 68534041-68534041 on Build GRCh38
rs893931229 Chr.8: 68534065-68534065 on Build GRCh38
rs11988777 Chr.8: 68534227-68534227 on Build GRCh38
rs924230200 Chr.8: 68534257-68534257 on Build GRCh38
rs751027491 Chr.8: 68534191-68534191 on Build GRCh38
rs917042025 Chr.8: 68533943-68533943 on Build GRCh38
rs1024115894 Chr.8: 68534144-68534144 on Build GRCh38
rs908871479 Chr.8: 68534165-68534165 on Build GRCh38
rs975892451 Chr.8: 68534105-68534105 on Build GRCh38
rs926957789 Chr.8: 68534192-68534192 on Build GRCh38
rs900462240 Chr.8: 68534066-68534066 on Build GRCh38
rs895309986 Chr.8: 68534018-68534018 on Build GRCh38
rs1008956777 Chr.8: 68534035-68534035 on Build GRCh38
rs765846716 Chr.8: 68534174-68534174 on Build GRCh38
rs1018325949 Chr.8: 68534096-68534096 on Build GRCh38
rs566655117 Chr.8: 68534054-68534054 on Build GRCh38
rs773151099 Chr.8: 68534039-68534039 on Build GRCh38
rs762381919 Chr.8: 68534141-68534141 on Build GRCh38
rs901242026 Chr.8: 68534328-68534328 on Build GRCh38
rs140506642 Chr.8: 68534031-68534031 on Build GRCh38
rs868738074 Chr.8: 68534027-68534027 on Build GRCh38
rs938263729 Chr.8: 68534000-68534000 on Build GRCh38
rs945621520 Chr.8: 68534269-68534269 on Build GRCh38
rs773332844 Chr.8: 68534323-68534323 on Build GRCh38
rs1027607769 Chr.8: 68534095-68534095 on Build GRCh38
rs945585600 Chr.8: 68534019-68534019 on Build GRCh38
rs531961599 Chr.8: 68533978-68533978 on Build GRCh38
rs112130514 Chr.8: 68534142-68534142 on Build GRCh38
rs549609212 Chr.8: 68533933-68533933 on Build GRCh38
rs973322239 Chr.8: 68533857-68533857 on Build GRCh38
rs146525135 Chr.8: 68534015-68534015 on Build GRCh38
rs1042549473 Chr.8: 68534282-68534282 on Build GRCh38
rs1054002997 Chr.8: 68534304-68534304 on Build GRCh38
rs1027493174 Chr.8: 68533875-68533875 on Build GRCh38
rs980188523 Chr.8: 68533972-68533972 on Build GRCh38
rs1054005788 Chr.8: 68534009-68534009 on Build GRCh38
rs11987961 Chr.8: 68533884-68533884 on Build GRCh38
rs555188112 Chr.8: 68534247-68534247 on Build GRCh38
rs571589646 Chr.8: 68534021-68534021 on Build GRCh38
rs964917469 Chr.8: 68534099-68534099 on Build GRCh38
rs1032298446 Chr.8: 68534060-68534060 on Build GRCh38
rs976007005 Chr.8: 68533919-68533919 on Build GRCh38
rs995172130 Chr.8: 68534074-68534074 on Build GRCh38
rs59050152 Chr.8: 68533935-68533935 on Build GRCh38
rs113341263 Chr.8: 68533995-68533995 on Build GRCh38
rs989769151 Chr.8: 68534237-68534237 on Build GRCh38
rs555729124 Chr.8: 68534288-68534288 on Build GRCh38
rs1046535347 Chr.8: 68534240-68534240 on Build GRCh38
rs181669364 Chr.8: 68534322-68534322 on Build GRCh38
rs1038972304 Chr.8: 68533934-68533934 on Build GRCh38
rs971572421 Chr.8: 68534173-68534173 on Build GRCh38
rs949577067 Chr.8: 68534229-68534229 on Build GRCh38
rs893807627 Chr.8: 68534317-68534317 on Build GRCh38
rs915514999 Chr.8: 68534262-68534262 on Build GRCh38

View Full Product Details