Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17229931-17230434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT7
Entrez Gene ID
29883
Gene Name
CCR4-NOT transcription complex subunit 7
Gene Aliases
CAF-1, CAF1, Caf1a, hCAF-1
UniGene
Hs.587465
Gene Chromosome Location
Chr.8: 17226030-17246909 on Build GRCh38
Transcripts
NM_001322090.1 NM_001322091.1 NM_001322092.1 NM_001322093.1 NM_001322094.1 NM_001322095.1 NM_001322096.1 NM_001322097.1 NM_013354.6 XM_005273481.2

SNP Detail

SNP ID Chromosome Location
rs979074589 Chr.8: 17230118-17230118 on Build GRCh38
rs1002365616 Chr.8: 17230049-17230049 on Build GRCh38
rs985579396 Chr.8: 17230132-17230133 on Build GRCh38
rs537006827 Chr.8: 17230282-17230282 on Build GRCh38
rs1021233204 Chr.8: 17230299-17230299 on Build GRCh38
rs897064479 Chr.8: 17230191-17230191 on Build GRCh38
rs887894740 Chr.8: 17230247-17230247 on Build GRCh38
rs779662265 Chr.8: 17230287-17230288 on Build GRCh38
rs1033842450 Chr.8: 17230060-17230060 on Build GRCh38
rs11554623 Chr.8: 17230408-17230408 on Build GRCh38
rs966396436 Chr.8: 17230106-17230106 on Build GRCh38
rs745729835 Chr.8: 17230282-17230283 on Build GRCh38
rs191737917 Chr.8: 17229991-17229991 on Build GRCh38
rs775442791 Chr.8: 17230304-17230304 on Build GRCh38
rs1049047632 Chr.8: 17230222-17230222 on Build GRCh38
rs545559906 Chr.8: 17230353-17230353 on Build GRCh38
rs966949722 Chr.8: 17230300-17230300 on Build GRCh38
rs912269365 Chr.8: 17230374-17230375 on Build GRCh38
rs1032020139 Chr.8: 17230325-17230325 on Build GRCh38
rs1034158590 Chr.8: 17230281-17230281 on Build GRCh38
rs749268949 Chr.8: 17229981-17229981 on Build GRCh38
rs1002249718 Chr.8: 17230274-17230274 on Build GRCh38
rs78750338 Chr.8: 17230128-17230128 on Build GRCh38
rs961124242 Chr.8: 17230080-17230080 on Build GRCh38
rs929330927 Chr.8: 17230218-17230218 on Build GRCh38
rs953655430 Chr.8: 17230340-17230340 on Build GRCh38
rs564621311 Chr.8: 17230144-17230144 on Build GRCh38
rs979127020 Chr.8: 17230312-17230312 on Build GRCh38
rs11554625 Chr.8: 17230151-17230151 on Build GRCh38
rs777683819 Chr.8: 17230186-17230186 on Build GRCh38
rs568217085 Chr.8: 17230234-17230234 on Build GRCh38
rs557363977 Chr.8: 17230293-17230293 on Build GRCh38
rs770853110 Chr.8: 17230126-17230126 on Build GRCh38
rs985245123 Chr.8: 17230341-17230344 on Build GRCh38
rs992443798 Chr.8: 17230084-17230084 on Build GRCh38
rs1036963014 Chr.8: 17230181-17230181 on Build GRCh38
rs758126733 Chr.8: 17230087-17230087 on Build GRCh38
rs373869725 Chr.8: 17230376-17230376 on Build GRCh38
rs912399389 Chr.8: 17230149-17230149 on Build GRCh38
rs140042337 Chr.8: 17230130-17230130 on Build GRCh38
rs185769502 Chr.8: 17229970-17229970 on Build GRCh38
rs11554626 Chr.8: 17230160-17230160 on Build GRCh38
rs887842634 Chr.8: 17230021-17230021 on Build GRCh38
rs1014159013 Chr.8: 17230292-17230292 on Build GRCh38
rs572020992 Chr.8: 17230283-17230283 on Build GRCh38
rs746864989 Chr.8: 17230232-17230232 on Build GRCh38

View Full Product Details