Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17868761-17869272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGL1
Entrez Gene ID
2267
Gene Name
fibrinogen like 1
Gene Aliases
HFREP1, HP-041, LFIRE-1, LFIRE1
UniGene
Hs.491143
Gene Chromosome Location
Chr.8: 17864380-17909983 on Build GRCh38
Transcripts
NM_004467.3 NM_147203.2 NM_201552.1 NM_201553.1 XM_017013232.1 XM_017013233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3982446 c.591+9A>T p.? Chr.8: 17868907-17868907 on Build GRCh38
7176771 c.551G>A p.R184H Chr.8: 17868956-17868956 on Build GRCh38
5409145 c.580G>A p.G194R Chr.8: 17868927-17868927 on Build GRCh38
1455866 c.570T>G p.N190K Chr.8: 17868937-17868937 on Build GRCh38
4684638 c.539A>G p.E180G Chr.8: 17868968-17868968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559066372 Chr.8: 17869061-17869061 on Build GRCh38
rs773643114 Chr.8: 17868960-17868960 on Build GRCh38
rs1033320028 Chr.8: 17868827-17868827 on Build GRCh38
rs541923505 Chr.8: 17869191-17869192 on Build GRCh38
rs1002458088 Chr.8: 17868811-17868811 on Build GRCh38
rs896725110 Chr.8: 17869217-17869217 on Build GRCh38
rs765692505 Chr.8: 17868957-17868957 on Build GRCh38
rs752150477 Chr.8: 17868978-17868978 on Build GRCh38
rs372421521 Chr.8: 17868952-17868952 on Build GRCh38
rs770083259 Chr.8: 17869046-17869046 on Build GRCh38
rs375555356 Chr.8: 17868922-17868922 on Build GRCh38
rs747017998 Chr.8: 17868949-17868949 on Build GRCh38
rs775368303 Chr.8: 17868866-17868866 on Build GRCh38
rs986288821 Chr.8: 17868923-17868923 on Build GRCh38
rs758239987 Chr.8: 17869011-17869011 on Build GRCh38
rs530738953 Chr.8: 17869177-17869177 on Build GRCh38
rs758615311 Chr.8: 17868935-17868935 on Build GRCh38
rs779623400 Chr.8: 17869068-17869068 on Build GRCh38
rs758894433 Chr.8: 17868888-17868888 on Build GRCh38
rs3213604 Chr.8: 17868907-17868907 on Build GRCh38
rs138480437 Chr.8: 17869139-17869139 on Build GRCh38
rs898762705 Chr.8: 17868800-17868800 on Build GRCh38
rs768150864 Chr.8: 17869169-17869169 on Build GRCh38
rs1055339694 Chr.8: 17869210-17869210 on Build GRCh38
rs776915892 Chr.8: 17869027-17869027 on Build GRCh38
rs1013668268 Chr.8: 17869064-17869064 on Build GRCh38
rs370228078 Chr.8: 17869055-17869055 on Build GRCh38
rs574875120 Chr.8: 17868829-17868829 on Build GRCh38
rs141817672 Chr.8: 17869206-17869206 on Build GRCh38
rs75939304 Chr.8: 17868887-17868887 on Build GRCh38
rs771850046 Chr.8: 17868943-17868943 on Build GRCh38
rs745604389 Chr.8: 17869008-17869008 on Build GRCh38
rs200822392 Chr.8: 17868976-17868976 on Build GRCh38
rs370213414 Chr.8: 17868876-17868876 on Build GRCh38
rs753488417 Chr.8: 17868985-17868985 on Build GRCh38
rs1032112652 Chr.8: 17869115-17869115 on Build GRCh38
rs549938225 Chr.8: 17868832-17868832 on Build GRCh38
rs535281825 Chr.8: 17868946-17868946 on Build GRCh38
rs201941547 Chr.8: 17868961-17868962 on Build GRCh38
rs773590136 Chr.8: 17869053-17869053 on Build GRCh38
rs757034174 Chr.8: 17868915-17868915 on Build GRCh38
rs186386014 Chr.8: 17869171-17869171 on Build GRCh38
rs766757109 Chr.8: 17868963-17868963 on Build GRCh38
rs139691812 Chr.8: 17868984-17868984 on Build GRCh38
rs908541932 Chr.8: 17869180-17869180 on Build GRCh38
rs909232455 Chr.8: 17869238-17869238 on Build GRCh38
rs1013284307 Chr.8: 17868854-17868854 on Build GRCh38
rs964501416 Chr.8: 17869130-17869130 on Build GRCh38
rs938145258 Chr.8: 17869209-17869209 on Build GRCh38
rs949671670 Chr.8: 17869221-17869221 on Build GRCh38
rs985222502 Chr.8: 17869233-17869233 on Build GRCh38
rs747013658 Chr.8: 17869021-17869021 on Build GRCh38
rs751678342 Chr.8: 17869020-17869020 on Build GRCh38
rs374549113 Chr.8: 17868879-17868879 on Build GRCh38
rs902227602 Chr.8: 17869084-17869084 on Build GRCh38
rs1000573691 Chr.8: 17869101-17869101 on Build GRCh38
rs145867321 Chr.8: 17869065-17869065 on Build GRCh38
rs147845891 Chr.8: 17868956-17868956 on Build GRCh38
rs537721215 Chr.8: 17868995-17868995 on Build GRCh38
rs1021106989 Chr.8: 17868869-17868869 on Build GRCh38
rs550819613 Chr.8: 17869178-17869178 on Build GRCh38
rs778552509 Chr.8: 17868996-17868996 on Build GRCh38
rs181272361 Chr.8: 17869017-17869017 on Build GRCh38
rs570007513 Chr.8: 17868877-17868877 on Build GRCh38
rs768602203 Chr.8: 17869023-17869023 on Build GRCh38
rs748241020 Chr.8: 17869039-17869039 on Build GRCh38
rs765741976 Chr.8: 17868883-17868883 on Build GRCh38
rs368295325 Chr.8: 17869054-17869054 on Build GRCh38
rs887609179 Chr.8: 17868843-17868843 on Build GRCh38
rs759122923 Chr.8: 17869056-17869056 on Build GRCh38
rs896760342 Chr.8: 17869058-17869058 on Build GRCh38
rs974165867 Chr.8: 17869202-17869202 on Build GRCh38
rs772192167 Chr.8: 17868865-17868865 on Build GRCh38
rs1000974251 Chr.8: 17869085-17869085 on Build GRCh38
rs763387971 Chr.8: 17868961-17868961 on Build GRCh38
rs745671326 Chr.8: 17868940-17868940 on Build GRCh38
rs372212133 Chr.8: 17869103-17869103 on Build GRCh38
rs368200345 Chr.8: 17868924-17868924 on Build GRCh38
rs940061890 Chr.8: 17869200-17869200 on Build GRCh38
rs200048711 Chr.8: 17868944-17868944 on Build GRCh38
rs896261405 Chr.8: 17868861-17868861 on Build GRCh38
rs371681226 Chr.8: 17868894-17868894 on Build GRCh38
rs752088937 Chr.8: 17868898-17868898 on Build GRCh38
rs755683462 Chr.8: 17868903-17868903 on Build GRCh38
rs769806615 Chr.8: 17869231-17869231 on Build GRCh38
rs368031186 Chr.8: 17868897-17868897 on Build GRCh38
rs1002072087 Chr.8: 17868846-17868846 on Build GRCh38
rs867705325 Chr.8: 17869232-17869232 on Build GRCh38
rs937883652 Chr.8: 17869248-17869248 on Build GRCh38

View Full Product Details