Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17885355-17885829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGL1
Entrez Gene ID
2267
Gene Name
fibrinogen like 1
Gene Aliases
HFREP1, HP-041, LFIRE-1, LFIRE1
UniGene
Hs.491143
Gene Chromosome Location
Chr.8: 17864380-17909983 on Build GRCh38
Transcripts
NM_004467.3 NM_147203.2 NM_201552.1 NM_201553.1 XM_017013232.1 XM_017013233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5409147 c.48G>A p.M16I Chr.8: 17885507-17885507 on Build GRCh38
3698996 c.13T>A p.F5I Chr.8: 17885542-17885542 on Build GRCh38
6663867 c.18T>G p.S6R Chr.8: 17885537-17885537 on Build GRCh38
3646948 c.54G>A p.R18R Chr.8: 17885501-17885501 on Build GRCh38
4569621 c.18T>C p.S6S Chr.8: 17885537-17885537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764401009 Chr.8: 17885605-17885605 on Build GRCh38
rs775490810 Chr.8: 17885475-17885475 on Build GRCh38
rs757936689 Chr.8: 17885601-17885601 on Build GRCh38
rs77473833 Chr.8: 17885629-17885629 on Build GRCh38
rs770330224 Chr.8: 17885455-17885457 on Build GRCh38
rs369898 Chr.8: 17885747-17885747 on Build GRCh38
rs34880881 Chr.8: 17885621-17885621 on Build GRCh38
rs952000680 Chr.8: 17885407-17885407 on Build GRCh38
rs774186621 Chr.8: 17885468-17885468 on Build GRCh38
rs1017023495 Chr.8: 17885433-17885433 on Build GRCh38
rs770659402 Chr.8: 17885464-17885464 on Build GRCh38
rs1013095183 Chr.8: 17885762-17885762 on Build GRCh38
rs187245262 Chr.8: 17885483-17885483 on Build GRCh38
rs998475761 Chr.8: 17885381-17885381 on Build GRCh38
rs61745164 Chr.8: 17885542-17885542 on Build GRCh38
rs182097449 Chr.8: 17885693-17885693 on Build GRCh38
rs995355946 Chr.8: 17885604-17885604 on Build GRCh38
rs367786231 Chr.8: 17885750-17885750 on Build GRCh38
rs576658380 Chr.8: 17885791-17885791 on Build GRCh38
rs149721242 Chr.8: 17885756-17885756 on Build GRCh38
rs896887251 Chr.8: 17885678-17885678 on Build GRCh38
rs779782679 Chr.8: 17885609-17885609 on Build GRCh38
rs373985863 Chr.8: 17885543-17885543 on Build GRCh38
rs575563596 Chr.8: 17885780-17885780 on Build GRCh38
rs924712767 Chr.8: 17885640-17885640 on Build GRCh38
rs762171697 Chr.8: 17885465-17885465 on Build GRCh38
rs577822711 Chr.8: 17885706-17885706 on Build GRCh38
rs1036757333 Chr.8: 17885676-17885676 on Build GRCh38
rs777082688 Chr.8: 17885448-17885448 on Build GRCh38
rs745580725 Chr.8: 17885541-17885541 on Build GRCh38
rs484373 Chr.8: 17885511-17885511 on Build GRCh38
rs755402959 Chr.8: 17885578-17885578 on Build GRCh38
rs887349026 Chr.8: 17885746-17885746 on Build GRCh38
rs2073562 Chr.8: 17885420-17885420 on Build GRCh38
rs776540301 Chr.8: 17885620-17885620 on Build GRCh38
rs376327490 Chr.8: 17885546-17885546 on Build GRCh38
rs1032110448 Chr.8: 17885800-17885800 on Build GRCh38
rs896960496 Chr.8: 17885588-17885588 on Build GRCh38
rs201345330 Chr.8: 17885486-17885486 on Build GRCh38
rs61745168 Chr.8: 17885521-17885521 on Build GRCh38
rs2073563 Chr.8: 17885444-17885444 on Build GRCh38
rs369087559 Chr.8: 17885441-17885441 on Build GRCh38
rs201530014 Chr.8: 17885499-17885499 on Build GRCh38
rs892882972 Chr.8: 17885599-17885599 on Build GRCh38
rs932893108 Chr.8: 17885659-17885659 on Build GRCh38
rs755383800 Chr.8: 17885505-17885505 on Build GRCh38
rs1012540986 Chr.8: 17885694-17885694 on Build GRCh38
rs990260075 Chr.8: 17885650-17885650 on Build GRCh38
rs995282235 Chr.8: 17885682-17885682 on Build GRCh38
rs752012351 Chr.8: 17885593-17885593 on Build GRCh38
rs553043332 Chr.8: 17885577-17885577 on Build GRCh38
rs182210693 Chr.8: 17885385-17885385 on Build GRCh38
rs763345269 Chr.8: 17885582-17885582 on Build GRCh38
rs761908696 Chr.8: 17885480-17885480 on Build GRCh38
rs540017594 Chr.8: 17885723-17885723 on Build GRCh38
rs1024901378 Chr.8: 17885733-17885733 on Build GRCh38
rs372906220 Chr.8: 17885624-17885624 on Build GRCh38
rs900075325 Chr.8: 17885381-17885382 on Build GRCh38
rs768548660 Chr.8: 17885613-17885613 on Build GRCh38
rs755364464 Chr.8: 17885639-17885639 on Build GRCh38
rs1049854242 Chr.8: 17885540-17885540 on Build GRCh38
rs112333965 Chr.8: 17885490-17885490 on Build GRCh38
rs112281518 Chr.8: 17885550-17885550 on Build GRCh38
rs759700090 Chr.8: 17885587-17885587 on Build GRCh38
rs761863714 Chr.8: 17885549-17885549 on Build GRCh38
rs191917407 Chr.8: 17885590-17885590 on Build GRCh38
rs17850145 Chr.8: 17885513-17885513 on Build GRCh38
rs766639216 Chr.8: 17885558-17885558 on Build GRCh38
rs1048252371 Chr.8: 17885687-17885687 on Build GRCh38
rs370686839 Chr.8: 17885553-17885553 on Build GRCh38
rs185824929 Chr.8: 17885801-17885801 on Build GRCh38
rs951553797 Chr.8: 17885419-17885419 on Build GRCh38
rs990232600 Chr.8: 17885787-17885787 on Build GRCh38
rs755175584 Chr.8: 17885481-17885483 on Build GRCh38
rs542439985 Chr.8: 17885770-17885770 on Build GRCh38
rs981900676 Chr.8: 17885631-17885631 on Build GRCh38
rs549303237 Chr.8: 17885469-17885469 on Build GRCh38
rs991878886 Chr.8: 17885412-17885412 on Build GRCh38
rs750696507 Chr.8: 17885484-17885484 on Build GRCh38
rs866985820 Chr.8: 17885674-17885674 on Build GRCh38
rs956111192 Chr.8: 17885768-17885768 on Build GRCh38
rs770614549 Chr.8: 17885518-17885518 on Build GRCh38
rs912178738 Chr.8: 17885669-17885669 on Build GRCh38
rs758752599 Chr.8: 17885491-17885491 on Build GRCh38
rs971883246 Chr.8: 17885434-17885434 on Build GRCh38
rs570617482 Chr.8: 17885537-17885537 on Build GRCh38
rs1026914519 Chr.8: 17885396-17885396 on Build GRCh38
rs950109741 Chr.8: 17885504-17885504 on Build GRCh38
rs376579189 Chr.8: 17885574-17885574 on Build GRCh38
rs914775754 Chr.8: 17885658-17885658 on Build GRCh38
rs373358723 Chr.8: 17885668-17885668 on Build GRCh38
rs780353033 Chr.8: 17885493-17885493 on Build GRCh38
rs1047890827 Chr.8: 17885696-17885696 on Build GRCh38
rs1017138420 Chr.8: 17885628-17885628 on Build GRCh38
rs748821535 Chr.8: 17885463-17885463 on Build GRCh38
rs376028403 Chr.8: 17885478-17885478 on Build GRCh38
rs763096688 Chr.8: 17885557-17885557 on Build GRCh38
rs756820868 Chr.8: 17885592-17885592 on Build GRCh38
rs566772667 Chr.8: 17885581-17885581 on Build GRCh38
rs778694966 Chr.8: 17885519-17885519 on Build GRCh38
rs956184507 Chr.8: 17885647-17885647 on Build GRCh38
rs773421975 Chr.8: 17885556-17885556 on Build GRCh38

View Full Product Details