Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18062174-18062719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH1
Entrez Gene ID
427
Gene Name
N-acylsphingosine amidohydrolase 1
Gene Aliases
AC, ACDase, ASAH, PHP, PHP32, SMAPME
UniGene
Hs.527412
Gene Chromosome Location
Chr.8: 18056299-18084998 on Build GRCh38
Transcripts
NM_001127505.2 NM_004315.5 NM_177924.4 XM_005273504.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5887999 c.695C>T p.P232L Chr.8: 18062280-18062280 on Build GRCh38
1154970 c.526C>G p.L176V Chr.8: 18062383-18062383 on Build GRCh38
6837205 c.638C>G p.T213S Chr.8: 18062337-18062337 on Build GRCh38
1097634 c.592C>G p.L198V Chr.8: 18062383-18062383 on Build GRCh38
4150690 c.540G>C p.E180D Chr.8: 18062387-18062387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750226117 Chr.8: 18062318-18062318 on Build GRCh38
rs754996239 Chr.8: 18062626-18062626 on Build GRCh38
rs567573744 Chr.8: 18062594-18062594 on Build GRCh38
rs761086598 Chr.8: 18062523-18062523 on Build GRCh38
rs889664197 Chr.8: 18062670-18062670 on Build GRCh38
rs143902833 Chr.8: 18062645-18062645 on Build GRCh38
rs781515666 Chr.8: 18062285-18062285 on Build GRCh38
rs985034577 Chr.8: 18062552-18062552 on Build GRCh38
rs780718023 Chr.8: 18062466-18062466 on Build GRCh38
rs1041112756 Chr.8: 18062499-18062499 on Build GRCh38
rs143342058 Chr.8: 18062462-18062462 on Build GRCh38
rs774882799 Chr.8: 18062363-18062363 on Build GRCh38
rs756455049 Chr.8: 18062422-18062422 on Build GRCh38
rs749643743 Chr.8: 18062351-18062351 on Build GRCh38
rs780061103 Chr.8: 18062253-18062253 on Build GRCh38
rs763715494 Chr.8: 18062242-18062242 on Build GRCh38
rs938165185 Chr.8: 18062562-18062562 on Build GRCh38
rs901342439 Chr.8: 18062505-18062505 on Build GRCh38
rs566970608 Chr.8: 18062395-18062395 on Build GRCh38
rs12541567 Chr.8: 18062568-18062568 on Build GRCh38
rs767762678 Chr.8: 18062419-18062419 on Build GRCh38
rs538175037 Chr.8: 18062522-18062522 on Build GRCh38
rs956781540 Chr.8: 18062571-18062571 on Build GRCh38
rs753659692 Chr.8: 18062243-18062243 on Build GRCh38
rs755120590 Chr.8: 18062406-18062406 on Build GRCh38
rs138332332 Chr.8: 18062533-18062533 on Build GRCh38
rs140561726 Chr.8: 18062368-18062368 on Build GRCh38
rs919528988 Chr.8: 18062280-18062280 on Build GRCh38
rs767741196 Chr.8: 18062508-18062508 on Build GRCh38
rs150268016 Chr.8: 18062307-18062307 on Build GRCh38
rs756389508 Chr.8: 18062343-18062343 on Build GRCh38
rs199909023 Chr.8: 18062345-18062345 on Build GRCh38
rs151083567 Chr.8: 18062289-18062289 on Build GRCh38
rs769601591 Chr.8: 18062231-18062235 on Build GRCh38
rs911240547 Chr.8: 18062606-18062606 on Build GRCh38
rs1000723723 Chr.8: 18062485-18062485 on Build GRCh38
rs147027727 Chr.8: 18062532-18062532 on Build GRCh38
rs535336475 Chr.8: 18062222-18062222 on Build GRCh38
rs766314172 Chr.8: 18062328-18062328 on Build GRCh38
rs766257867 Chr.8: 18062391-18062391 on Build GRCh38
rs1041536550 Chr.8: 18062629-18062629 on Build GRCh38
rs757579385 Chr.8: 18062444-18062444 on Build GRCh38
rs151320126 Chr.8: 18062335-18062335 on Build GRCh38
rs141068211 Chr.8: 18062298-18062298 on Build GRCh38
rs180947624 Chr.8: 18062641-18062641 on Build GRCh38
rs983781009 Chr.8: 18062321-18062321 on Build GRCh38
rs761518207 Chr.8: 18062377-18062377 on Build GRCh38
rs762756953 Chr.8: 18062389-18062389 on Build GRCh38
rs760287134 Chr.8: 18062234-18062234 on Build GRCh38
rs370655492 Chr.8: 18062250-18062250 on Build GRCh38
rs751515915 Chr.8: 18062620-18062620 on Build GRCh38
rs758339998 Chr.8: 18062251-18062251 on Build GRCh38
rs138920776 Chr.8: 18062427-18062427 on Build GRCh38
rs1050276941 Chr.8: 18062604-18062604 on Build GRCh38
rs539898166 Chr.8: 18062632-18062632 on Build GRCh38
rs115756271 Chr.8: 18062599-18062599 on Build GRCh38
rs186378479 Chr.8: 18062227-18062227 on Build GRCh38
rs779745050 Chr.8: 18062600-18062600 on Build GRCh38
rs774912301 Chr.8: 18062305-18062305 on Build GRCh38
rs374289481 Chr.8: 18062259-18062259 on Build GRCh38
rs540913734 Chr.8: 18062376-18062376 on Build GRCh38
rs1019842179 Chr.8: 18062678-18062678 on Build GRCh38
rs532912450 Chr.8: 18062226-18062226 on Build GRCh38
rs771365660 Chr.8: 18062358-18062358 on Build GRCh38
rs909422096 Chr.8: 18062556-18062556 on Build GRCh38
rs751609815 Chr.8: 18062330-18062330 on Build GRCh38
rs947770502 Chr.8: 18062366-18062366 on Build GRCh38
rs761612547 Chr.8: 18062310-18062310 on Build GRCh38
rs747632799 Chr.8: 18062475-18062475 on Build GRCh38
rs775288080 Chr.8: 18062262-18062264 on Build GRCh38
rs772925787 Chr.8: 18062380-18062380 on Build GRCh38
rs746434957 Chr.8: 18062454-18062454 on Build GRCh38
rs116160319 Chr.8: 18062534-18062534 on Build GRCh38
rs576151596 Chr.8: 18062674-18062674 on Build GRCh38
rs1013596750 Chr.8: 18062387-18062387 on Build GRCh38
rs778830228 Chr.8: 18062246-18062246 on Build GRCh38
rs929574620 Chr.8: 18062547-18062547 on Build GRCh38
rs145381040 Chr.8: 18062215-18062215 on Build GRCh38
rs368212646 Chr.8: 18062371-18062371 on Build GRCh38
rs189703202 Chr.8: 18062480-18062480 on Build GRCh38
rs551625847 Chr.8: 18062291-18062291 on Build GRCh38
rs1032168895 Chr.8: 18062490-18062490 on Build GRCh38
rs758304705 Chr.8: 18062325-18062325 on Build GRCh38
rs759113229 Chr.8: 18062648-18062648 on Build GRCh38
rs749646498 Chr.8: 18062433-18062433 on Build GRCh38
rs765078195 Chr.8: 18062315-18062315 on Build GRCh38
rs757009749 Chr.8: 18062245-18062245 on Build GRCh38
rs889897500 Chr.8: 18062488-18062488 on Build GRCh38
rs1052336290 Chr.8: 18062213-18062213 on Build GRCh38
rs137853597 Chr.8: 18062383-18062383 on Build GRCh38
rs748322562 Chr.8: 18062336-18062336 on Build GRCh38
rs530829304 Chr.8: 18062538-18062538 on Build GRCh38
rs1007399083 Chr.8: 18062496-18062496 on Build GRCh38
rs763439501 Chr.8: 18062296-18062296 on Build GRCh38

View Full Product Details