Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 19403869-19404380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT1
Entrez Gene ID
55790
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CSGalNAcT-1, ChGn, beta4GalNAcT
UniGene
Hs.613729
Gene Chromosome Location
Chr.8: 19404161-19757940 on Build GRCh38
Transcripts
NM_001130518.1 NM_018371.4 NR_024040.1 XM_006716358.2 XM_006716359.2 XM_006716360.2 XM_006716361.2 XM_006716362.2 XM_006716363.1 XM_006716364.3 XM_011544578.1 XM_011544579.1 XM_011544580.1 XM_011544582.1 XM_011544583.1 XM_011544584.1 XM_011544585.2 XM_017013623.1 XM_017013624.1 XM_017013625.1 XM_017013626.1 XM_017013627.1 XM_017013628.1 XM_017013629.1 XM_017013630.1 XM_017013631.1 XM_017013632.1 XM_017013633.1 XM_017013634.1 XM_017013635.1 XM_017013636.1 XM_017013637.1 XM_017013638.1 XM_017013639.1

SNP Detail

SNP ID Chromosome Location
rs1004552901 Chr.8: 19404315-19404315 on Build GRCh38
rs925156617 Chr.8: 19404040-19404040 on Build GRCh38
rs377086566 Chr.8: 19403927-19403927 on Build GRCh38
rs540162753 Chr.8: 19404348-19404351 on Build GRCh38
rs548476516 Chr.8: 19404050-19404050 on Build GRCh38
rs562936755 Chr.8: 19404333-19404333 on Build GRCh38
rs553039039 Chr.8: 19404304-19404304 on Build GRCh38
rs914666478 Chr.8: 19404157-19404157 on Build GRCh38
rs774111927 Chr.8: 19404193-19404193 on Build GRCh38
rs1024763312 Chr.8: 19404251-19404251 on Build GRCh38
rs927504822 Chr.8: 19403950-19403950 on Build GRCh38
rs770095531 Chr.8: 19403929-19403929 on Build GRCh38
rs141955684 Chr.8: 19404179-19404179 on Build GRCh38
rs914761500 Chr.8: 19404000-19404000 on Build GRCh38
rs934768235 Chr.8: 19403985-19403985 on Build GRCh38
rs983260481 Chr.8: 19403901-19403901 on Build GRCh38
rs1005090170 Chr.8: 19403924-19403924 on Build GRCh38
rs1051837484 Chr.8: 19404103-19404103 on Build GRCh38
rs532321542 Chr.8: 19404215-19404215 on Build GRCh38
rs1023942134 Chr.8: 19404123-19404123 on Build GRCh38
rs960260848 Chr.8: 19404203-19404203 on Build GRCh38
rs945060995 Chr.8: 19404025-19404025 on Build GRCh38
rs947663063 Chr.8: 19404014-19404014 on Build GRCh38
rs752824226 Chr.8: 19404308-19404308 on Build GRCh38
rs997562612 Chr.8: 19404028-19404028 on Build GRCh38
rs1034920405 Chr.8: 19404161-19404161 on Build GRCh38
rs963671176 Chr.8: 19403942-19403942 on Build GRCh38
rs1039657133 Chr.8: 19404024-19404024 on Build GRCh38
rs147998103 Chr.8: 19404126-19404126 on Build GRCh38
rs981388389 Chr.8: 19404139-19404139 on Build GRCh38
rs1004668736 Chr.8: 19404085-19404085 on Build GRCh38
rs767766982 Chr.8: 19404306-19404306 on Build GRCh38
rs796235789 Chr.8: 19404358-19404358 on Build GRCh38
rs1037821766 Chr.8: 19404326-19404326 on Build GRCh38
rs373884795 Chr.8: 19404199-19404199 on Build GRCh38
rs577012605 Chr.8: 19404206-19404206 on Build GRCh38
rs997542639 Chr.8: 19404092-19404092 on Build GRCh38
rs756737024 Chr.8: 19404116-19404116 on Build GRCh38
rs192109535 Chr.8: 19404042-19404042 on Build GRCh38
rs948693456 Chr.8: 19403947-19403950 on Build GRCh38
rs1001809289 Chr.8: 19404150-19404150 on Build GRCh38
rs540510170 Chr.8: 19404056-19404056 on Build GRCh38
rs936601559 Chr.8: 19404052-19404052 on Build GRCh38
rs565223418 Chr.8: 19404142-19404142 on Build GRCh38
rs62493852 Chr.8: 19403946-19403946 on Build GRCh38
rs963943590 Chr.8: 19404148-19404148 on Build GRCh38
rs759843866 Chr.8: 19403977-19403977 on Build GRCh38
rs7001231 Chr.8: 19404219-19404219 on Build GRCh38
rs1004806618 Chr.8: 19404138-19404138 on Build GRCh38
rs950461682 Chr.8: 19403890-19403891 on Build GRCh38
rs552687284 Chr.8: 19403965-19403965 on Build GRCh38
rs767888280 Chr.8: 19403999-19403999 on Build GRCh38
rs963504926 Chr.8: 19404096-19404096 on Build GRCh38
rs187255505 Chr.8: 19404026-19404026 on Build GRCh38
rs568377815 Chr.8: 19404094-19404094 on Build GRCh38
rs1023489580 Chr.8: 19404348-19404349 on Build GRCh38
rs563943111 Chr.8: 19403940-19403940 on Build GRCh38
rs763429382 Chr.8: 19404273-19404273 on Build GRCh38
rs1016101535 Chr.8: 19404093-19404093 on Build GRCh38
rs550124784 Chr.8: 19403932-19403932 on Build GRCh38
rs924989741 Chr.8: 19404343-19404343 on Build GRCh38
rs764062391 Chr.8: 19404066-19404066 on Build GRCh38

View Full Product Details