Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 20204401-20204907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1B2
Entrez Gene ID
526
Gene Name
ATPase H+ transporting V1 subunit B2
Gene Aliases
ATP6B1B2, ATP6B2, DOOD, HO57, VATB, VPP3, Vma2, ZLS2
UniGene
Hs.295917
Gene Chromosome Location
Chr.8: 20197193-20226852 on Build GRCh38
Transcripts
NM_001693.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298647 c.192+2T>C p.? Chr.8: 20204541-20204541 on Build GRCh38
7186963 c.182A>G p.D61G Chr.8: 20204529-20204529 on Build GRCh38
6112834 c.178T>G p.L60V Chr.8: 20204525-20204525 on Build GRCh38
354642 c.171A>G p.L57L Chr.8: 20204518-20204518 on Build GRCh38
3834384 c.137-1G>T p.? Chr.8: 20204483-20204483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907023557 Chr.8: 20204582-20204582 on Build GRCh38
rs117177999 Chr.8: 20204835-20204835 on Build GRCh38
rs564214885 Chr.8: 20204681-20204697 on Build GRCh38
rs889170497 Chr.8: 20204630-20204630 on Build GRCh38
rs766487263 Chr.8: 20204525-20204525 on Build GRCh38
rs570667716 Chr.8: 20204854-20204854 on Build GRCh38
rs768400416 Chr.8: 20204499-20204499 on Build GRCh38
rs776556396 Chr.8: 20204506-20204506 on Build GRCh38
rs560333267 Chr.8: 20204810-20204810 on Build GRCh38
rs373240364 Chr.8: 20204598-20204598 on Build GRCh38
rs556241850 Chr.8: 20204472-20204472 on Build GRCh38
rs763274868 Chr.8: 20204516-20204516 on Build GRCh38
rs1045508085 Chr.8: 20204570-20204570 on Build GRCh38
rs774663114 Chr.8: 20204534-20204534 on Build GRCh38
rs562318749 Chr.8: 20204737-20204737 on Build GRCh38
rs776034125 Chr.8: 20204679-20204680 on Build GRCh38
rs897391670 Chr.8: 20204815-20204815 on Build GRCh38
rs755587622 Chr.8: 20204459-20204459 on Build GRCh38
rs1049706824 Chr.8: 20204784-20204784 on Build GRCh38
rs764845794 Chr.8: 20204553-20204553 on Build GRCh38
rs749883642 Chr.8: 20204559-20204559 on Build GRCh38
rs919539237 Chr.8: 20204430-20204430 on Build GRCh38
rs112063532 Chr.8: 20204546-20204546 on Build GRCh38
rs558158291 Chr.8: 20204650-20204650 on Build GRCh38
rs914434756 Chr.8: 20204750-20204750 on Build GRCh38
rs934552383 Chr.8: 20204583-20204583 on Build GRCh38
rs1026951690 Chr.8: 20204653-20204653 on Build GRCh38
rs200124277 Chr.8: 20204508-20204508 on Build GRCh38
rs757961145 Chr.8: 20204485-20204485 on Build GRCh38
rs888624096 Chr.8: 20204851-20204851 on Build GRCh38
rs934476085 Chr.8: 20204746-20204746 on Build GRCh38
rs757880067 Chr.8: 20204576-20204576 on Build GRCh38
rs188636896 Chr.8: 20204659-20204659 on Build GRCh38
rs576289068 Chr.8: 20204657-20204657 on Build GRCh38
rs192525820 Chr.8: 20204679-20204679 on Build GRCh38
rs753267498 Chr.8: 20204551-20204551 on Build GRCh38
rs754783173 Chr.8: 20204588-20204588 on Build GRCh38
rs948477636 Chr.8: 20204562-20204562 on Build GRCh38
rs994327752 Chr.8: 20204581-20204581 on Build GRCh38
rs760006623 Chr.8: 20204540-20204540 on Build GRCh38
rs145647828 Chr.8: 20204748-20204748 on Build GRCh38
rs776932730 Chr.8: 20204687-20204687 on Build GRCh38
rs746600467 Chr.8: 20204587-20204587 on Build GRCh38
rs779952110 Chr.8: 20204493-20204493 on Build GRCh38
rs756828947 Chr.8: 20204470-20204470 on Build GRCh38
rs751941103 Chr.8: 20204451-20204451 on Build GRCh38
rs765396291 Chr.8: 20204807-20204807 on Build GRCh38
rs746829559 Chr.8: 20204495-20204495 on Build GRCh38
rs1048505966 Chr.8: 20204850-20204850 on Build GRCh38
rs753212630 Chr.8: 20204469-20204469 on Build GRCh38
rs768155931 Chr.8: 20204464-20204464 on Build GRCh38
rs1036461942 Chr.8: 20204802-20204802 on Build GRCh38
rs981155699 Chr.8: 20204704-20204704 on Build GRCh38
rs928428166 Chr.8: 20204727-20204727 on Build GRCh38
rs185371803 Chr.8: 20204613-20204613 on Build GRCh38
rs901937432 Chr.8: 20204873-20204873 on Build GRCh38
rs966590644 Chr.8: 20204647-20204647 on Build GRCh38
rs779712186 Chr.8: 20204586-20204586 on Build GRCh38
rs773158882 Chr.8: 20204512-20204512 on Build GRCh38
rs527477044 Chr.8: 20204827-20204827 on Build GRCh38
rs183455699 Chr.8: 20204794-20204794 on Build GRCh38
rs769091323 Chr.8: 20204824-20204824 on Build GRCh38
rs140696770 Chr.8: 20204605-20204605 on Build GRCh38
rs947294604 Chr.8: 20204767-20204767 on Build GRCh38
rs763459289 Chr.8: 20204705-20204705 on Build GRCh38
rs564344209 Chr.8: 20204655-20204655 on Build GRCh38
rs1007639955 Chr.8: 20204639-20204639 on Build GRCh38
rs373517039 Chr.8: 20204858-20204858 on Build GRCh38
rs1050084523 Chr.8: 20204465-20204465 on Build GRCh38
rs931679776 Chr.8: 20204747-20204747 on Build GRCh38
rs745604509 Chr.8: 20204473-20204473 on Build GRCh38
rs1019067623 Chr.8: 20204645-20204645 on Build GRCh38
rs538138975 Chr.8: 20204468-20204468 on Build GRCh38
rs984383421 Chr.8: 20204673-20204673 on Build GRCh38

View Full Product Details