Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 20211541-20212045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1B2
Entrez Gene ID
526
Gene Name
ATPase H+ transporting V1 subunit B2
Gene Aliases
ATP6B1B2, ATP6B2, DOOD, HO57, VATB, VPP3, Vma2, ZLS2
UniGene
Hs.295917
Gene Chromosome Location
Chr.8: 20197193-20226852 on Build GRCh38
Transcripts
NM_001693.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354699 c.665A>G p.Y222C Chr.8: 20211713-20211713 on Build GRCh38
7357031 c.702G>A p.M234I Chr.8: 20211750-20211750 on Build GRCh38
7400237 c.702G>C p.M234I Chr.8: 20211750-20211750 on Build GRCh38
3647086 c.648C>T p.S216S Chr.8: 20211696-20211696 on Build GRCh38
7414734 c.682G>T p.A228S Chr.8: 20211730-20211730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016721214 Chr.8: 20211946-20211946 on Build GRCh38
rs769843486 Chr.8: 20211783-20211783 on Build GRCh38
rs553807546 Chr.8: 20211831-20211831 on Build GRCh38
rs746992809 Chr.8: 20211769-20211769 on Build GRCh38
rs369235753 Chr.8: 20211738-20211738 on Build GRCh38
rs80121793 Chr.8: 20211763-20211764 on Build GRCh38
rs748210018 Chr.8: 20211779-20211779 on Build GRCh38
rs771485223 Chr.8: 20211802-20211802 on Build GRCh38
rs180737041 Chr.8: 20211851-20211851 on Build GRCh38
rs1034512702 Chr.8: 20211993-20211993 on Build GRCh38
rs941779397 Chr.8: 20211889-20211889 on Build GRCh38
rs142952762 Chr.8: 20211762-20211763 on Build GRCh38
rs1016380250 Chr.8: 20211620-20211620 on Build GRCh38
rs376254752 Chr.8: 20211711-20211711 on Build GRCh38
rs998057563 Chr.8: 20211877-20211877 on Build GRCh38
rs372607416 Chr.8: 20211837-20211837 on Build GRCh38
rs546878455 Chr.8: 20211617-20211617 on Build GRCh38
rs139800942 Chr.8: 20211705-20211705 on Build GRCh38
rs1046993415 Chr.8: 20211880-20211880 on Build GRCh38
rs200664744 Chr.8: 20211671-20211671 on Build GRCh38
rs1004900362 Chr.8: 20211929-20211929 on Build GRCh38
rs766788781 Chr.8: 20211714-20211714 on Build GRCh38
rs752043229 Chr.8: 20211607-20211607 on Build GRCh38
rs773316883 Chr.8: 20211786-20211786 on Build GRCh38
rs538948064 Chr.8: 20211918-20211918 on Build GRCh38
rs114753726 Chr.8: 20211732-20211732 on Build GRCh38
rs778815381 Chr.8: 20211642-20211642 on Build GRCh38
rs763412091 Chr.8: 20211713-20211713 on Build GRCh38
rs186120877 Chr.8: 20211992-20211992 on Build GRCh38
rs775420686 Chr.8: 20211670-20211670 on Build GRCh38
rs143242270 Chr.8: 20211847-20211847 on Build GRCh38
rs758248437 Chr.8: 20211766-20211766 on Build GRCh38
rs751961802 Chr.8: 20211631-20211633 on Build GRCh38
rs143922607 Chr.8: 20211726-20211726 on Build GRCh38
rs963499884 Chr.8: 20211953-20211953 on Build GRCh38
rs1047449920 Chr.8: 20211578-20211578 on Build GRCh38
rs750559171 Chr.8: 20211790-20211791 on Build GRCh38
rs1018029832 Chr.8: 20211846-20211846 on Build GRCh38
rs368469837 Chr.8: 20211764-20211764 on Build GRCh38
rs543687429 Chr.8: 20211908-20211908 on Build GRCh38
rs774806095 Chr.8: 20211865-20211865 on Build GRCh38
rs894688976 Chr.8: 20211706-20211706 on Build GRCh38
rs776546672 Chr.8: 20211694-20211694 on Build GRCh38
rs375143864 Chr.8: 20211803-20211803 on Build GRCh38
rs975987071 Chr.8: 20211855-20211855 on Build GRCh38
rs762147835 Chr.8: 20211704-20211704 on Build GRCh38
rs755695034 Chr.8: 20211613-20211613 on Build GRCh38
rs894818994 Chr.8: 20211952-20211952 on Build GRCh38
rs572383531 Chr.8: 20211601-20211601 on Build GRCh38
rs779820435 Chr.8: 20211767-20211767 on Build GRCh38
rs1002057043 Chr.8: 20211985-20211985 on Build GRCh38
rs749036513 Chr.8: 20211625-20211625 on Build GRCh38
rs768791251 Chr.8: 20211683-20211683 on Build GRCh38
rs541896931 Chr.8: 20211760-20211760 on Build GRCh38
rs768562483 Chr.8: 20211774-20211774 on Build GRCh38
rs771790292 Chr.8: 20211663-20211663 on Build GRCh38
rs372596079 Chr.8: 20211777-20211777 on Build GRCh38
rs763251381 Chr.8: 20211789-20211789 on Build GRCh38
rs572132909 Chr.8: 20211840-20211840 on Build GRCh38
rs137894692 Chr.8: 20211845-20211845 on Build GRCh38
rs960217860 Chr.8: 20212007-20212007 on Build GRCh38
rs756827467 Chr.8: 20211638-20211638 on Build GRCh38
rs1054765844 Chr.8: 20211944-20211944 on Build GRCh38
rs576185708 Chr.8: 20211857-20211857 on Build GRCh38
rs945760052 Chr.8: 20211827-20211827 on Build GRCh38
rs753264154 Chr.8: 20211735-20211735 on Build GRCh38
rs901048281 Chr.8: 20211872-20211872 on Build GRCh38
rs763698959 Chr.8: 20211733-20211733 on Build GRCh38

View Full Product Details