Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 21912137-21912647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK2
Entrez Gene ID
9046
Gene Name
docking protein 2
Gene Aliases
p56DOK, p56dok-2
UniGene
Hs.71215
Gene Chromosome Location
Chr.8: 21908873-21913717 on Build GRCh38
Transcripts
NM_001317800.1 NM_003974.3 NM_201349.2 XM_005273680.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4411584 c.173A>C p.K58T Chr.8: 21912401-21912401 on Build GRCh38
4429855 c.310G>A p.D104N Chr.8: 21912264-21912264 on Build GRCh38
228481 c.301G>A p.E101K Chr.8: 21912273-21912273 on Build GRCh38
5757184 c.199C>T p.R67W Chr.8: 21912375-21912375 on Build GRCh38
4531833 c.182G>A p.R61H Chr.8: 21912392-21912392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770573554 Chr.8: 21912484-21912484 on Build GRCh38
rs972673135 Chr.8: 21912357-21912357 on Build GRCh38
rs760058864 Chr.8: 21912260-21912260 on Build GRCh38
rs769706992 Chr.8: 21912455-21912455 on Build GRCh38
rs2957337 Chr.8: 21912519-21912519 on Build GRCh38
rs750297661 Chr.8: 21912210-21912210 on Build GRCh38
rs370181142 Chr.8: 21912453-21912453 on Build GRCh38
rs529243950 Chr.8: 21912486-21912486 on Build GRCh38
rs368280286 Chr.8: 21912386-21912386 on Build GRCh38
rs866790904 Chr.8: 21912448-21912448 on Build GRCh38
rs577698027 Chr.8: 21912447-21912447 on Build GRCh38
rs756042144 Chr.8: 21912359-21912359 on Build GRCh38
rs539886209 Chr.8: 21912541-21912541 on Build GRCh38
rs749348418 Chr.8: 21912361-21912361 on Build GRCh38
rs779935483 Chr.8: 21912218-21912218 on Build GRCh38
rs1057407704 Chr.8: 21912456-21912456 on Build GRCh38
rs771734200 Chr.8: 21912491-21912491 on Build GRCh38
rs775844922 Chr.8: 21912317-21912317 on Build GRCh38
rs764063442 Chr.8: 21912427-21912427 on Build GRCh38
rs777847247 Chr.8: 21912294-21912294 on Build GRCh38
rs772334877 Chr.8: 21912380-21912380 on Build GRCh38
rs771293728 Chr.8: 21912237-21912237 on Build GRCh38
rs368204724 Chr.8: 21912580-21912580 on Build GRCh38
rs990624463 Chr.8: 21912324-21912324 on Build GRCh38
rs767250698 Chr.8: 21912421-21912421 on Build GRCh38
rs746491737 Chr.8: 21912165-21912165 on Build GRCh38
rs73541515 Chr.8: 21912236-21912236 on Build GRCh38
rs753987268 Chr.8: 21912433-21912433 on Build GRCh38
rs747500789 Chr.8: 21912392-21912392 on Build GRCh38
rs762517469 Chr.8: 21912399-21912399 on Build GRCh38
rs773296143 Chr.8: 21912234-21912234 on Build GRCh38
rs200971271 Chr.8: 21912187-21912187 on Build GRCh38
rs746943260 Chr.8: 21912220-21912220 on Build GRCh38
rs777271397 Chr.8: 21912472-21912472 on Build GRCh38
rs911913808 Chr.8: 21912563-21912563 on Build GRCh38
rs552782288 Chr.8: 21912347-21912347 on Build GRCh38
rs770853017 Chr.8: 21912600-21912600 on Build GRCh38
rs769080665 Chr.8: 21912470-21912470 on Build GRCh38
rs766198984 Chr.8: 21912405-21912405 on Build GRCh38
rs369991976 Chr.8: 21912485-21912485 on Build GRCh38
rs986408112 Chr.8: 21912569-21912569 on Build GRCh38
rs752492055 Chr.8: 21912423-21912423 on Build GRCh38
rs771093305 Chr.8: 21912302-21912302 on Build GRCh38
rs199740321 Chr.8: 21912268-21912268 on Build GRCh38
rs947406040 Chr.8: 21912366-21912366 on Build GRCh38
rs775189966 Chr.8: 21912495-21912495 on Build GRCh38
rs772735107 Chr.8: 21912334-21912334 on Build GRCh38
rs372882463 Chr.8: 21912422-21912422 on Build GRCh38
rs570637744 Chr.8: 21912607-21912607 on Build GRCh38
rs937874835 Chr.8: 21912182-21912182 on Build GRCh38
rs531906443 Chr.8: 21912465-21912465 on Build GRCh38
rs370594320 Chr.8: 21912360-21912360 on Build GRCh38
rs538174636 Chr.8: 21912619-21912619 on Build GRCh38
rs969172517 Chr.8: 21912598-21912598 on Build GRCh38
rs868648335 Chr.8: 21912511-21912511 on Build GRCh38
rs933929306 Chr.8: 21912538-21912538 on Build GRCh38
rs747457180 Chr.8: 21912488-21912519 on Build GRCh38
rs918458875 Chr.8: 21912524-21912524 on Build GRCh38
rs867912936 Chr.8: 21912490-21912490 on Build GRCh38
rs754809072 Chr.8: 21912344-21912344 on Build GRCh38
rs770123577 Chr.8: 21912229-21912229 on Build GRCh38
rs568152896 Chr.8: 21912473-21912473 on Build GRCh38
rs890625197 Chr.8: 21912253-21912253 on Build GRCh38
rs574468600 Chr.8: 21912222-21912222 on Build GRCh38
rs903193113 Chr.8: 21912434-21912434 on Build GRCh38
rs375844220 Chr.8: 21912321-21912321 on Build GRCh38
rs760515264 Chr.8: 21912498-21912498 on Build GRCh38
rs147786765 Chr.8: 21912311-21912311 on Build GRCh38
rs972670959 Chr.8: 21912503-21912503 on Build GRCh38
rs375816776 Chr.8: 21912457-21912457 on Build GRCh38
rs749504624 Chr.8: 21912297-21912297 on Build GRCh38
rs772642448 Chr.8: 21912396-21912396 on Build GRCh38
rs201025320 Chr.8: 21912500-21912500 on Build GRCh38
rs748321214 Chr.8: 21912283-21912283 on Build GRCh38
rs769780504 Chr.8: 21912285-21912285 on Build GRCh38
rs778166605 Chr.8: 21912338-21912339 on Build GRCh38
rs779104485 Chr.8: 21912374-21912374 on Build GRCh38
rs146943090 Chr.8: 21912393-21912393 on Build GRCh38
rs546091332 Chr.8: 21912303-21912303 on Build GRCh38
rs371879955 Chr.8: 21912274-21912274 on Build GRCh38
rs758056880 Chr.8: 21912269-21912269 on Build GRCh38
rs11313762 Chr.8: 21912209-21912209 on Build GRCh38
rs756056583 Chr.8: 21912426-21912426 on Build GRCh38
rs1035827426 Chr.8: 21912264-21912264 on Build GRCh38
rs374735206 Chr.8: 21912265-21912265 on Build GRCh38
rs765331663 Chr.8: 21912540-21912540 on Build GRCh38
rs1023378123 Chr.8: 21912476-21912476 on Build GRCh38
rs141129446 Chr.8: 21912337-21912337 on Build GRCh38
rs182205822 Chr.8: 21912186-21912186 on Build GRCh38
rs767991803 Chr.8: 21912262-21912262 on Build GRCh38
rs749216314 Chr.8: 21912547-21912547 on Build GRCh38
rs780958143 Chr.8: 21912279-21912279 on Build GRCh38
rs757512119 Chr.8: 21912367-21912367 on Build GRCh38
rs753784888 Chr.8: 21912515-21912515 on Build GRCh38
rs774659649 Chr.8: 21912248-21912248 on Build GRCh38
rs375699639 Chr.8: 21912466-21912466 on Build GRCh38
rs751346454 Chr.8: 21912273-21912273 on Build GRCh38
rs759200913 Chr.8: 21912419-21912419 on Build GRCh38
rs780023106 Chr.8: 21912272-21912272 on Build GRCh38
rs865865361 Chr.8: 21912492-21912492 on Build GRCh38
rs910388186 Chr.8: 21912586-21912586 on Build GRCh38
rs556222778 Chr.8: 21912193-21912193 on Build GRCh38
rs866821306 Chr.8: 21912369-21912369 on Build GRCh38
rs367990658 Chr.8: 21912267-21912267 on Build GRCh38
rs756983935 Chr.8: 21912195-21912195 on Build GRCh38
rs917541912 Chr.8: 21912622-21912622 on Build GRCh38
rs758114610 Chr.8: 21912216-21912216 on Build GRCh38
rs924584872 Chr.8: 21912601-21912601 on Build GRCh38
rs759864633 Chr.8: 21912310-21912310 on Build GRCh38
rs746287821 Chr.8: 21912378-21912378 on Build GRCh38
rs897844136 Chr.8: 21912231-21912231 on Build GRCh38
rs780288303 Chr.8: 21912560-21912560 on Build GRCh38
rs1050486455 Chr.8: 21912194-21912194 on Build GRCh38
rs757884231 Chr.8: 21912339-21912339 on Build GRCh38
rs558493086 Chr.8: 21912594-21912594 on Build GRCh38
rs752585347 Chr.8: 21912358-21912358 on Build GRCh38
rs748150790 Chr.8: 21912228-21912228 on Build GRCh38
rs185989167 Chr.8: 21912522-21912522 on Build GRCh38
rs778397645 Chr.8: 21912203-21912203 on Build GRCh38
rs766105823 Chr.8: 21912340-21912340 on Build GRCh38
rs751438222 Chr.8: 21912342-21912342 on Build GRCh38
rs777896625 Chr.8: 21912494-21912494 on Build GRCh38
rs1039157032 Chr.8: 21912402-21912402 on Build GRCh38

View Full Product Details