Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22046285-22046791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF17
Entrez Gene ID
8822
Gene Name
fibroblast growth factor 17
Gene Aliases
FGF-13, FGF-17, HH20
UniGene
Hs.248192
Gene Chromosome Location
Chr.8: 22039708-22048809 on Build GRCh38
Transcripts
NM_001304478.1 NM_003867.3 XM_005273675.1 XM_011544683.1 XM_011544684.2 XM_011544685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207064 c.287G>A p.R96Q Chr.8: 22046563-22046563 on Build GRCh38
6663519 c.270G>A p.T90T Chr.8: 22046546-22046546 on Build GRCh38
2785589 c.288G>T p.R96R Chr.8: 22046564-22046564 on Build GRCh38
1699785 c.292C>T p.R98C Chr.8: 22046568-22046568 on Build GRCh38
1284961 c.294C>T p.R98R Chr.8: 22046570-22046570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946253872 Chr.8: 22046454-22046454 on Build GRCh38
rs776733398 Chr.8: 22046311-22046311 on Build GRCh38
rs190867302 Chr.8: 22046490-22046490 on Build GRCh38
rs574112401 Chr.8: 22046453-22046453 on Build GRCh38
rs370093040 Chr.8: 22046676-22046676 on Build GRCh38
rs560392250 Chr.8: 22046469-22046469 on Build GRCh38
rs902962443 Chr.8: 22046764-22046764 on Build GRCh38
rs749233488 Chr.8: 22046627-22046627 on Build GRCh38
rs1041423261 Chr.8: 22046395-22046395 on Build GRCh38
rs773246227 Chr.8: 22046495-22046495 on Build GRCh38
rs368916503 Chr.8: 22046498-22046498 on Build GRCh38
rs780943518 Chr.8: 22046594-22046594 on Build GRCh38
rs1008767952 Chr.8: 22046566-22046566 on Build GRCh38
rs946907121 Chr.8: 22046715-22046715 on Build GRCh38
rs911414025 Chr.8: 22046372-22046372 on Build GRCh38
rs932907856 Chr.8: 22046417-22046417 on Build GRCh38
rs745402028 Chr.8: 22046342-22046342 on Build GRCh38
rs754727639 Chr.8: 22046700-22046700 on Build GRCh38
rs1016623494 Chr.8: 22046519-22046519 on Build GRCh38
rs372548164 Chr.8: 22046694-22046694 on Build GRCh38
rs145470556 Chr.8: 22046674-22046674 on Build GRCh38
rs202160304 Chr.8: 22046630-22046630 on Build GRCh38
rs757790277 Chr.8: 22046680-22046680 on Build GRCh38
rs976832825 Chr.8: 22046675-22046675 on Build GRCh38
rs1042561666 Chr.8: 22046721-22046721 on Build GRCh38
rs1041424543 Chr.8: 22046481-22046481 on Build GRCh38
rs771016753 Chr.8: 22046335-22046335 on Build GRCh38
rs764613581 Chr.8: 22046524-22046524 on Build GRCh38
rs762983576 Chr.8: 22046497-22046497 on Build GRCh38
rs144144739 Chr.8: 22046534-22046534 on Build GRCh38
rs1042864834 Chr.8: 22046488-22046488 on Build GRCh38
rs576825379 Chr.8: 22046321-22046321 on Build GRCh38
rs959704829 Chr.8: 22046545-22046545 on Build GRCh38
rs373308419 Chr.8: 22046315-22046315 on Build GRCh38
rs897758780 Chr.8: 22046521-22046521 on Build GRCh38
rs774505557 Chr.8: 22046358-22046358 on Build GRCh38
rs990347940 Chr.8: 22046579-22046579 on Build GRCh38
rs932631384 Chr.8: 22046682-22046682 on Build GRCh38
rs398123024 Chr.8: 22046599-22046599 on Build GRCh38
rs541459497 Chr.8: 22046463-22046463 on Build GRCh38
rs775839407 Chr.8: 22046540-22046540 on Build GRCh38
rs779616279 Chr.8: 22046546-22046546 on Build GRCh38
rs984177327 Chr.8: 22046363-22046363 on Build GRCh38
rs942880576 Chr.8: 22046428-22046428 on Build GRCh38
rs774492730 Chr.8: 22046649-22046649 on Build GRCh38
rs555859502 Chr.8: 22046452-22046452 on Build GRCh38
rs778190743 Chr.8: 22046333-22046333 on Build GRCh38
rs1078363 Chr.8: 22046701-22046701 on Build GRCh38
rs371279355 Chr.8: 22046663-22046663 on Build GRCh38
rs528017799 Chr.8: 22046476-22046476 on Build GRCh38
rs1007254524 Chr.8: 22046457-22046457 on Build GRCh38
rs182343990 Chr.8: 22046754-22046754 on Build GRCh38
rs2957330 Chr.8: 22046314-22046314 on Build GRCh38
rs1008217276 Chr.8: 22046766-22046766 on Build GRCh38
rs777426592 Chr.8: 22046333-22046348 on Build GRCh38
rs746441702 Chr.8: 22046569-22046569 on Build GRCh38
rs756613942 Chr.8: 22046324-22046324 on Build GRCh38
rs544300366 Chr.8: 22046389-22046389 on Build GRCh38
rs1051629149 Chr.8: 22046757-22046758 on Build GRCh38
rs908625057 Chr.8: 22046383-22046383 on Build GRCh38
rs758193197 Chr.8: 22046318-22046319 on Build GRCh38
rs774492997 Chr.8: 22046502-22046502 on Build GRCh38
rs897623096 Chr.8: 22046710-22046710 on Build GRCh38
rs3176295 Chr.8: 22046615-22046615 on Build GRCh38
rs200608386 Chr.8: 22046612-22046612 on Build GRCh38
rs118120728 Chr.8: 22046625-22046625 on Build GRCh38
rs142215324 Chr.8: 22046628-22046628 on Build GRCh38
rs922906064 Chr.8: 22046397-22046397 on Build GRCh38
rs993383661 Chr.8: 22046712-22046712 on Build GRCh38
rs769875515 Chr.8: 22046493-22046493 on Build GRCh38
rs2960137 Chr.8: 22046307-22046307 on Build GRCh38
rs199703542 Chr.8: 22046518-22046518 on Build GRCh38
rs201297515 Chr.8: 22046563-22046563 on Build GRCh38
rs370290685 Chr.8: 22046306-22046306 on Build GRCh38
rs752572149 Chr.8: 22046307-22046307 on Build GRCh38
rs952507755 Chr.8: 22046347-22046347 on Build GRCh38
rs759799719 Chr.8: 22046512-22046512 on Build GRCh38
rs941745134 Chr.8: 22046691-22046691 on Build GRCh38
rs767658166 Chr.8: 22046513-22046513 on Build GRCh38
rs374424349 Chr.8: 22046668-22046668 on Build GRCh38
rs369682032 Chr.8: 22046570-22046570 on Build GRCh38
rs776579827 Chr.8: 22046312-22046312 on Build GRCh38
rs552040652 Chr.8: 22046753-22046753 on Build GRCh38
rs764788712 Chr.8: 22046770-22046770 on Build GRCh38
rs867130159 Chr.8: 22046621-22046621 on Build GRCh38
rs376460755 Chr.8: 22046522-22046522 on Build GRCh38
rs759473624 Chr.8: 22046659-22046659 on Build GRCh38
rs2960138 Chr.8: 22046328-22046328 on Build GRCh38
rs767745650 Chr.8: 22046661-22046661 on Build GRCh38
rs767670173 Chr.8: 22046491-22046491 on Build GRCh38
rs768406192 Chr.8: 22046482-22046482 on Build GRCh38
rs754337894 Chr.8: 22046687-22046687 on Build GRCh38
rs746663363 Chr.8: 22046479-22046479 on Build GRCh38
rs1049982571 Chr.8: 22046450-22046450 on Build GRCh38
rs149341718 Chr.8: 22046528-22046528 on Build GRCh38

View Full Product Details