Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22069335-22069827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMTN
Entrez Gene ID
2039
Gene Name
dematin actin binding protein
Gene Aliases
DMT, EPB49
UniGene
Hs.106124
Gene Chromosome Location
Chr.8: 22048955-22082527 on Build GRCh38
Transcripts
NM_001114135.4 NM_001114136.2 NM_001114137.3 NM_001114138.2 NM_001114139.3 NM_001302816.2 NM_001302817.2 NM_001323378.1 NM_001323380.1 NM_001323381.1 NM_001323382.1 NM_001323383.1 NM_001323384.1 NM_001323385.1 NM_001323387.1 NM_001323388.1 NM_001323389.1 NM_001323390.1 NM_001323391.1 NM_001323392.1 NM_001323393.1 NM_001323394.1 NM_001323395.1 NM_001323396.1 NM_001323397.1 NM_001323398.1 NM_001323399.1 NM_001323400.1 NM_001323401.1 NM_001978.4 XM_005273432.1 XM_005273433.1 XM_005273442.1 XM_011544435.1 XM_017013189.1 XM_017013190.1 XM_017013191.1 XM_017013192.1 XM_017013193.1 XM_017013194.1 XM_017013195.1 XM_017013196.1 XM_017013197.1 XM_017013198.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355668 c.295-1G>T p.? Chr.8: 22069418-22069418 on Build GRCh38
5514526 c.365G>A p.R122Q Chr.8: 22069489-22069489 on Build GRCh38
1456023 c.359delC p.R122fs*109 Chr.8: 22069483-22069483 on Build GRCh38
4791398 c.303G>A p.A101A Chr.8: 22069427-22069427 on Build GRCh38
6536481 c.331C>G p.Q111E Chr.8: 22069455-22069455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760897459 Chr.8: 22069416-22069416 on Build GRCh38
rs769796263 Chr.8: 22069392-22069392 on Build GRCh38
rs760749884 Chr.8: 22069370-22069371 on Build GRCh38
rs768890543 Chr.8: 22069488-22069488 on Build GRCh38
rs12680100 Chr.8: 22069626-22069626 on Build GRCh38
rs949443558 Chr.8: 22069368-22069368 on Build GRCh38
rs376609490 Chr.8: 22069564-22069564 on Build GRCh38
rs546172597 Chr.8: 22069586-22069586 on Build GRCh38
rs763788185 Chr.8: 22069563-22069563 on Build GRCh38
rs772215398 Chr.8: 22069414-22069414 on Build GRCh38
rs532164106 Chr.8: 22069439-22069439 on Build GRCh38
rs571127179 Chr.8: 22069679-22069679 on Build GRCh38
rs747897075 Chr.8: 22069386-22069386 on Build GRCh38
rs775647615 Chr.8: 22069486-22069486 on Build GRCh38
rs145460473 Chr.8: 22069435-22069435 on Build GRCh38
rs763796953 Chr.8: 22069385-22069385 on Build GRCh38
rs539385159 Chr.8: 22069426-22069426 on Build GRCh38
rs757199597 Chr.8: 22069566-22069566 on Build GRCh38
rs149018201 Chr.8: 22069466-22069466 on Build GRCh38
rs745889849 Chr.8: 22069471-22069471 on Build GRCh38
rs529582238 Chr.8: 22069557-22069557 on Build GRCh38
rs200406300 Chr.8: 22069427-22069427 on Build GRCh38
rs929059774 Chr.8: 22069390-22069390 on Build GRCh38
rs879718507 Chr.8: 22069373-22069373 on Build GRCh38
rs901457962 Chr.8: 22069787-22069787 on Build GRCh38
rs565232601 Chr.8: 22069357-22069357 on Build GRCh38
rs905932377 Chr.8: 22069735-22069735 on Build GRCh38
rs765786457 Chr.8: 22069529-22069529 on Build GRCh38
rs371952202 Chr.8: 22069485-22069485 on Build GRCh38
rs762583204 Chr.8: 22069430-22069430 on Build GRCh38
rs781143626 Chr.8: 22069375-22069375 on Build GRCh38
rs970376818 Chr.8: 22069623-22069623 on Build GRCh38
rs1001449511 Chr.8: 22069499-22069499 on Build GRCh38
rs972899987 Chr.8: 22069361-22069361 on Build GRCh38
rs375614247 Chr.8: 22069535-22069535 on Build GRCh38
rs183408735 Chr.8: 22069722-22069722 on Build GRCh38
rs556729981 Chr.8: 22069688-22069688 on Build GRCh38
rs780947354 Chr.8: 22069449-22069449 on Build GRCh38
rs766413952 Chr.8: 22069362-22069362 on Build GRCh38
rs538013769 Chr.8: 22069683-22069683 on Build GRCh38
rs146615047 Chr.8: 22069501-22069501 on Build GRCh38
rs141336563 Chr.8: 22069508-22069508 on Build GRCh38
rs79543612 Chr.8: 22069573-22069573 on Build GRCh38
rs368383977 Chr.8: 22069462-22069462 on Build GRCh38
rs376084791 Chr.8: 22069403-22069403 on Build GRCh38
rs759079340 Chr.8: 22069531-22069531 on Build GRCh38
rs1048474756 Chr.8: 22069680-22069680 on Build GRCh38
rs891617692 Chr.8: 22069691-22069691 on Build GRCh38
rs190703612 Chr.8: 22069699-22069699 on Build GRCh38
rs919013660 Chr.8: 22069369-22069369 on Build GRCh38
rs898767975 Chr.8: 22069792-22069792 on Build GRCh38
rs755990971 Chr.8: 22069539-22069539 on Build GRCh38
rs1053400515 Chr.8: 22069664-22069664 on Build GRCh38
rs997518232 Chr.8: 22069800-22069800 on Build GRCh38
rs774610512 Chr.8: 22069404-22069404 on Build GRCh38
rs987213301 Chr.8: 22069600-22069600 on Build GRCh38
rs368652897 Chr.8: 22069536-22069536 on Build GRCh38
rs1051453898 Chr.8: 22069395-22069395 on Build GRCh38
rs1045792608 Chr.8: 22069705-22069705 on Build GRCh38
rs772734374 Chr.8: 22069770-22069770 on Build GRCh38
rs574871738 Chr.8: 22069757-22069757 on Build GRCh38
rs778969258 Chr.8: 22069467-22069467 on Build GRCh38
rs550763719 Chr.8: 22069464-22069464 on Build GRCh38
rs778771330 Chr.8: 22069569-22069569 on Build GRCh38
rs968341534 Chr.8: 22069528-22069528 on Build GRCh38
rs761380868 Chr.8: 22069690-22069690 on Build GRCh38
rs79079952 Chr.8: 22069651-22069651 on Build GRCh38
rs774024887 Chr.8: 22069530-22069530 on Build GRCh38
rs1038977778 Chr.8: 22069438-22069438 on Build GRCh38
rs752574575 Chr.8: 22069452-22069452 on Build GRCh38
rs145887401 Chr.8: 22069490-22069490 on Build GRCh38
rs775845525 Chr.8: 22069415-22069415 on Build GRCh38
rs146914678 Chr.8: 22069587-22069587 on Build GRCh38
rs777812551 Chr.8: 22069463-22069463 on Build GRCh38
rs12681799 Chr.8: 22069700-22069700 on Build GRCh38
rs753875036 Chr.8: 22069632-22069632 on Build GRCh38
rs935913911 Chr.8: 22069663-22069663 on Build GRCh38
rs566116238 Chr.8: 22069611-22069611 on Build GRCh38
rs1016876921 Chr.8: 22069790-22069790 on Build GRCh38
rs370461300 Chr.8: 22069406-22069406 on Build GRCh38
rs970039627 Chr.8: 22069496-22069496 on Build GRCh38
rs112125971 Chr.8: 22069743-22069743 on Build GRCh38
rs911609992 Chr.8: 22069609-22069609 on Build GRCh38
rs1014338666 Chr.8: 22069489-22069489 on Build GRCh38
rs1001502109 Chr.8: 22069761-22069761 on Build GRCh38

View Full Product Details