Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22689772-22690266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR3
Entrez Gene ID
1960
Gene Name
early growth response 3
Gene Aliases
EGR-3, PILOT
UniGene
Hs.534313
Gene Chromosome Location
Chr.8: 22687661-22693302 on Build GRCh38
Transcripts
NM_001199880.1 NM_001199881.1 NM_004430.2 XM_005273425.3 XM_005273426.3 XM_011544429.2

SNP Detail

SNP ID Chromosome Location
rs142737697 Chr.8: 22689972-22689972 on Build GRCh38
rs1040542533 Chr.8: 22690166-22690166 on Build GRCh38
rs1028890498 Chr.8: 22689956-22689956 on Build GRCh38
rs74304574 Chr.8: 22690029-22690029 on Build GRCh38
rs1047604145 Chr.8: 22690089-22690089 on Build GRCh38
rs1020629982 Chr.8: 22689859-22689859 on Build GRCh38
rs959676781 Chr.8: 22689931-22689931 on Build GRCh38
rs569867611 Chr.8: 22690064-22690064 on Build GRCh38
rs1044717059 Chr.8: 22689921-22689922 on Build GRCh38
rs764396821 Chr.8: 22689876-22689876 on Build GRCh38
rs983542853 Chr.8: 22689996-22689996 on Build GRCh38
rs549322093 Chr.8: 22690091-22690091 on Build GRCh38
rs1034724924 Chr.8: 22690245-22690245 on Build GRCh38
rs921503681 Chr.8: 22689821-22689821 on Build GRCh38
rs930595787 Chr.8: 22690086-22690086 on Build GRCh38
rs561319224 Chr.8: 22689831-22689831 on Build GRCh38
rs965949515 Chr.8: 22689946-22689946 on Build GRCh38
rs1018659933 Chr.8: 22689945-22689946 on Build GRCh38
rs774960526 Chr.8: 22690052-22690052 on Build GRCh38
rs932624909 Chr.8: 22690059-22690059 on Build GRCh38
rs533096601 Chr.8: 22690017-22690017 on Build GRCh38
rs886111574 Chr.8: 22690118-22690118 on Build GRCh38
rs867469635 Chr.8: 22690056-22690056 on Build GRCh38
rs543733332 Chr.8: 22689800-22689800 on Build GRCh38
rs934607764 Chr.8: 22690009-22690009 on Build GRCh38
rs1010669170 Chr.8: 22690072-22690072 on Build GRCh38
rs867282466 Chr.8: 22689933-22689933 on Build GRCh38
rs893650183 Chr.8: 22690071-22690071 on Build GRCh38
rs930758825 Chr.8: 22689983-22689983 on Build GRCh38
rs541187152 Chr.8: 22690232-22690232 on Build GRCh38
rs934764279 Chr.8: 22689909-22689909 on Build GRCh38
rs540684319 Chr.8: 22689921-22689921 on Build GRCh38
rs994695935 Chr.8: 22689930-22689933 on Build GRCh38
rs975579769 Chr.8: 22689953-22689953 on Build GRCh38
rs373801955 Chr.8: 22690210-22690210 on Build GRCh38
rs1008542728 Chr.8: 22690157-22690157 on Build GRCh38
rs530806776 Chr.8: 22690083-22690083 on Build GRCh38
rs924612731 Chr.8: 22690008-22690008 on Build GRCh38
rs948556566 Chr.8: 22689920-22689923 on Build GRCh38
rs868394131 Chr.8: 22689844-22689844 on Build GRCh38
rs1003688823 Chr.8: 22690137-22690137 on Build GRCh38
rs183177754 Chr.8: 22689919-22689919 on Build GRCh38
rs1049594658 Chr.8: 22690060-22690060 on Build GRCh38
rs764359347 Chr.8: 22690030-22690030 on Build GRCh38
rs754236899 Chr.8: 22689883-22689883 on Build GRCh38
rs551277884 Chr.8: 22690027-22690027 on Build GRCh38
rs902281212 Chr.8: 22690120-22690120 on Build GRCh38
rs976075923 Chr.8: 22689809-22689809 on Build GRCh38
rs947384323 Chr.8: 22689881-22689881 on Build GRCh38
rs966794459 Chr.8: 22689894-22689894 on Build GRCh38
rs978999104 Chr.8: 22689991-22689991 on Build GRCh38
rs1057182923 Chr.8: 22689914-22689914 on Build GRCh38
rs376046298 Chr.8: 22690122-22690122 on Build GRCh38

View Full Product Details