Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25296222-25296767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK5
Entrez Gene ID
80005
Gene Name
dedicator of cytokinesis 5
Gene Aliases
-
UniGene
Hs.195403
Gene Chromosome Location
Chr.8: 25184723-25415716 on Build GRCh38
Transcripts
NM_024940.7 XM_011544661.1 XM_011544662.1 XM_011544663.2 XM_017013865.1 XM_017013866.1 XM_017013868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3929517 c.499G>C p.D167H Chr.8: 25296541-25296541 on Build GRCh38
291015 c.501T>C p.D167D Chr.8: 25296543-25296543 on Build GRCh38
6495645 c.495G>A p.V165V Chr.8: 25296537-25296537 on Build GRCh38
3899100 c.571T>C p.S191P Chr.8: 25296613-25296613 on Build GRCh38
6013098 c.516C>T p.I172I Chr.8: 25296558-25296558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753825164 Chr.8: 25296446-25296446 on Build GRCh38
rs997942774 Chr.8: 25296405-25296405 on Build GRCh38
rs61732777 Chr.8: 25296561-25296561 on Build GRCh38
rs758080324 Chr.8: 25296709-25296709 on Build GRCh38
rs1034253686 Chr.8: 25296437-25296437 on Build GRCh38
rs753460222 Chr.8: 25296484-25296484 on Build GRCh38
rs778032313 Chr.8: 25296598-25296598 on Build GRCh38
rs765837278 Chr.8: 25296571-25296571 on Build GRCh38
rs780973314 Chr.8: 25296532-25296532 on Build GRCh38
rs374015497 Chr.8: 25296541-25296541 on Build GRCh38
rs201659492 Chr.8: 25296508-25296508 on Build GRCh38
rs149326440 Chr.8: 25296308-25296308 on Build GRCh38
rs779947182 Chr.8: 25296520-25296520 on Build GRCh38
rs887682048 Chr.8: 25296457-25296457 on Build GRCh38
rs914920101 Chr.8: 25296458-25296458 on Build GRCh38
rs755799941 Chr.8: 25296512-25296512 on Build GRCh38
rs1018986169 Chr.8: 25296383-25296383 on Build GRCh38
rs774894495 Chr.8: 25296464-25296464 on Build GRCh38
rs770043731 Chr.8: 25296538-25296538 on Build GRCh38
rs766134056 Chr.8: 25296445-25296445 on Build GRCh38
rs758374912 Chr.8: 25296514-25296514 on Build GRCh38
rs759953877 Chr.8: 25296471-25296471 on Build GRCh38
rs546947818 Chr.8: 25296360-25296360 on Build GRCh38
rs369984539 Chr.8: 25296539-25296539 on Build GRCh38
rs965189048 Chr.8: 25296397-25296397 on Build GRCh38
rs756863186 Chr.8: 25296498-25296498 on Build GRCh38
rs1005403839 Chr.8: 25296361-25296361 on Build GRCh38
rs747353064 Chr.8: 25296583-25296583 on Build GRCh38
rs760078239 Chr.8: 25296552-25296552 on Build GRCh38
rs772324434 Chr.8: 25296559-25296559 on Build GRCh38
rs901908776 Chr.8: 25296662-25296662 on Build GRCh38
rs978283637 Chr.8: 25296526-25296526 on Build GRCh38
rs955583289 Chr.8: 25296554-25296554 on Build GRCh38
rs961450354 Chr.8: 25296510-25296510 on Build GRCh38
rs775017076 Chr.8: 25296542-25296542 on Build GRCh38
rs997993593 Chr.8: 25296706-25296706 on Build GRCh38
rs748332549 Chr.8: 25296536-25296536 on Build GRCh38
rs746387195 Chr.8: 25296612-25296612 on Build GRCh38
rs193230791 Chr.8: 25296414-25296414 on Build GRCh38
rs767942865 Chr.8: 25296482-25296482 on Build GRCh38
rs779064994 Chr.8: 25296607-25296607 on Build GRCh38
rs761081395 Chr.8: 25296643-25296643 on Build GRCh38
rs780906985 Chr.8: 25296586-25296586 on Build GRCh38
rs1049507775 Chr.8: 25296336-25296336 on Build GRCh38
rs772632928 Chr.8: 25296628-25296628 on Build GRCh38
rs945745918 Chr.8: 25296314-25296314 on Build GRCh38
rs966395206 Chr.8: 25296496-25296496 on Build GRCh38
rs749987596 Chr.8: 25296568-25296568 on Build GRCh38
rs73673885 Chr.8: 25296657-25296657 on Build GRCh38
rs150049709 Chr.8: 25296564-25296564 on Build GRCh38
rs113835979 Chr.8: 25296430-25296430 on Build GRCh38
rs749345836 Chr.8: 25296540-25296540 on Build GRCh38
rs376012368 Chr.8: 25296263-25296263 on Build GRCh38
rs764714809 Chr.8: 25296500-25296500 on Build GRCh38
rs749470233 Chr.8: 25296603-25296603 on Build GRCh38
rs34576741 Chr.8: 25296570-25296570 on Build GRCh38
rs377088823 Chr.8: 25296606-25296606 on Build GRCh38
rs916850632 Chr.8: 25296563-25296563 on Build GRCh38
rs754799894 Chr.8: 25296529-25296529 on Build GRCh38
rs1054622008 Chr.8: 25296253-25296253 on Build GRCh38
rs775762790 Chr.8: 25296636-25296636 on Build GRCh38
rs376663203 Chr.8: 25296527-25296527 on Build GRCh38
rs561486482 Chr.8: 25296264-25296264 on Build GRCh38
rs751502358 Chr.8: 25296580-25296580 on Build GRCh38
rs772884845 Chr.8: 25296685-25296685 on Build GRCh38
rs527949746 Chr.8: 25296455-25296455 on Build GRCh38
rs989396002 Chr.8: 25296440-25296440 on Build GRCh38
rs752373973 Chr.8: 25296489-25296489 on Build GRCh38
rs756329033 Chr.8: 25296595-25296595 on Build GRCh38

View Full Product Details