Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25423734-25424235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNRH1
Entrez Gene ID
2796
Gene Name
gonadotropin releasing hormone 1
Gene Aliases
GNRH, GRH, LHRH, LNRH
UniGene
Hs.82963
Gene Chromosome Location
Chr.8: 25419258-25425040 on Build GRCh38
Transcripts
NM_000825.3 NM_001083111.1

SNP Detail

SNP ID Chromosome Location
rs1024482079 Chr.8: 25424010-25424010 on Build GRCh38
rs972063506 Chr.8: 25424082-25424082 on Build GRCh38
rs962938386 Chr.8: 25424142-25424142 on Build GRCh38
rs368021772 Chr.8: 25424186-25424186 on Build GRCh38
rs980254568 Chr.8: 25424092-25424092 on Build GRCh38
rs1032750626 Chr.8: 25423888-25423888 on Build GRCh38
rs1028036261 Chr.8: 25423918-25423918 on Build GRCh38
rs530580841 Chr.8: 25424041-25424041 on Build GRCh38
rs185048861 Chr.8: 25423862-25423862 on Build GRCh38
rs902182855 Chr.8: 25423771-25423771 on Build GRCh38
rs532608268 Chr.8: 25423761-25423761 on Build GRCh38
rs890383544 Chr.8: 25423798-25423798 on Build GRCh38
rs575887190 Chr.8: 25423816-25423816 on Build GRCh38
rs974522296 Chr.8: 25424093-25424093 on Build GRCh38
rs950743581 Chr.8: 25423927-25423927 on Build GRCh38
rs559177514 Chr.8: 25424149-25424149 on Build GRCh38
rs1034775671 Chr.8: 25423780-25423780 on Build GRCh38
rs573738491 Chr.8: 25424063-25424063 on Build GRCh38
rs577750806 Chr.8: 25424173-25424173 on Build GRCh38
rs373861534 Chr.8: 25423993-25423993 on Build GRCh38
rs999641495 Chr.8: 25423772-25423772 on Build GRCh38
rs566896578 Chr.8: 25423843-25423843 on Build GRCh38
rs534899690 Chr.8: 25424007-25424007 on Build GRCh38
rs544978358 Chr.8: 25424193-25424193 on Build GRCh38
rs867048611 Chr.8: 25424190-25424190 on Build GRCh38
rs78322917 Chr.8: 25424196-25424196 on Build GRCh38
rs569784222 Chr.8: 25423974-25423974 on Build GRCh38
rs547583807 Chr.8: 25424125-25424125 on Build GRCh38
rs999892407 Chr.8: 25423884-25423884 on Build GRCh38
rs867890095 Chr.8: 25423760-25423760 on Build GRCh38
rs983891239 Chr.8: 25423945-25423945 on Build GRCh38
rs17790824 Chr.8: 25423813-25423813 on Build GRCh38
rs555503339 Chr.8: 25423914-25423914 on Build GRCh38
rs911772501 Chr.8: 25424197-25424197 on Build GRCh38
rs918691959 Chr.8: 25424188-25424188 on Build GRCh38
rs970274871 Chr.8: 25423862-25423863 on Build GRCh38

View Full Product Details