Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25424925-25425428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNRH1
Entrez Gene ID
2796
Gene Name
gonadotropin releasing hormone 1
Gene Aliases
GNRH, GRH, LHRH, LNRH
UniGene
Hs.82963
Gene Chromosome Location
Chr.8: 25419258-25425040 on Build GRCh38
Transcripts
NM_000825.3 NM_001083111.1

SNP Detail

SNP ID Chromosome Location
rs917550276 Chr.8: 25425088-25425088 on Build GRCh38
rs975453948 Chr.8: 25425143-25425143 on Build GRCh38
rs898756128 Chr.8: 25425040-25425040 on Build GRCh38
rs1001226325 Chr.8: 25424988-25424988 on Build GRCh38
rs906843604 Chr.8: 25424982-25424982 on Build GRCh38
rs542748304 Chr.8: 25425343-25425343 on Build GRCh38
rs769458170 Chr.8: 25425213-25425216 on Build GRCh38
rs769143143 Chr.8: 25425200-25425200 on Build GRCh38
rs189563909 Chr.8: 25424951-25424951 on Build GRCh38
rs562234587 Chr.8: 25425148-25425148 on Build GRCh38
rs533217785 Chr.8: 25425314-25425314 on Build GRCh38
rs1055465223 Chr.8: 25425271-25425271 on Build GRCh38
rs902136338 Chr.8: 25425231-25425231 on Build GRCh38
rs1005050731 Chr.8: 25425194-25425194 on Build GRCh38
rs957287635 Chr.8: 25425188-25425188 on Build GRCh38
rs931041172 Chr.8: 25425372-25425372 on Build GRCh38
rs913060069 Chr.8: 25425193-25425193 on Build GRCh38
rs2321048 Chr.8: 25425076-25425076 on Build GRCh38
rs917825577 Chr.8: 25425317-25425317 on Build GRCh38
rs535046522 Chr.8: 25425179-25425179 on Build GRCh38
rs199511048 Chr.8: 25425266-25425267 on Build GRCh38
rs1029292812 Chr.8: 25425154-25425154 on Build GRCh38
rs1044780637 Chr.8: 25425144-25425144 on Build GRCh38
rs773484263 Chr.8: 25425363-25425363 on Build GRCh38
rs1028976125 Chr.8: 25425248-25425248 on Build GRCh38
rs1056420502 Chr.8: 25425030-25425030 on Build GRCh38
rs964369133 Chr.8: 25425133-25425133 on Build GRCh38
rs548578379 Chr.8: 25425316-25425316 on Build GRCh38
rs559821666 Chr.8: 25425251-25425251 on Build GRCh38
rs992032575 Chr.8: 25425315-25425315 on Build GRCh38
rs557032411 Chr.8: 25424978-25424978 on Build GRCh38
rs770796366 Chr.8: 25425334-25425335 on Build GRCh38
rs981574057 Chr.8: 25425235-25425235 on Build GRCh38
rs1049201060 Chr.8: 25425165-25425165 on Build GRCh38
rs776497850 Chr.8: 25425107-25425109 on Build GRCh38
rs931543121 Chr.8: 25425301-25425301 on Build GRCh38
rs996122056 Chr.8: 25425236-25425236 on Build GRCh38
rs796349528 Chr.8: 25425287-25425287 on Build GRCh38
rs571535123 Chr.8: 25425367-25425367 on Build GRCh38
rs1008713833 Chr.8: 25425087-25425087 on Build GRCh38
rs923791322 Chr.8: 25424996-25424996 on Build GRCh38
rs2321049 Chr.8: 25425333-25425333 on Build GRCh38
rs995633911 Chr.8: 25425059-25425059 on Build GRCh38
rs945906358 Chr.8: 25425206-25425206 on Build GRCh38
rs541220028 Chr.8: 25425189-25425189 on Build GRCh38
rs549296865 Chr.8: 25425395-25425395 on Build GRCh38
rs1020090178 Chr.8: 25425358-25425358 on Build GRCh38
rs1025813416 Chr.8: 25425065-25425065 on Build GRCh38
rs187271288 Chr.8: 25425255-25425255 on Build GRCh38
rs149931306 Chr.8: 25425359-25425359 on Build GRCh38
rs529500147 Chr.8: 25425187-25425187 on Build GRCh38
rs968567852 Chr.8: 25424985-25424985 on Build GRCh38
rs956621195 Chr.8: 25425007-25425007 on Build GRCh38
rs773853007 Chr.8: 25424958-25424958 on Build GRCh38
rs1045519607 Chr.8: 25424966-25424966 on Build GRCh38
rs180879343 Chr.8: 25424980-25424980 on Build GRCh38
rs1019721724 Chr.8: 25425124-25425124 on Build GRCh38

View Full Product Details