Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27461191-27461702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755677 c.1543G>T p.G515W Chr.8: 27461676-27461676 on Build GRCh38
1098733 c.1555C>A p.L519I Chr.8: 27461664-27461664 on Build GRCh38
3647808 c.1580G>A p.G527E Chr.8: 27461639-27461639 on Build GRCh38
4397625 c.1567C>T p.P523S Chr.8: 27461652-27461652 on Build GRCh38
3647809 c.1553G>A p.G518D Chr.8: 27461666-27461666 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1054185789 Chr.8: 27461363-27461363 on Build GRCh38
rs940210331 Chr.8: 27461247-27461247 on Build GRCh38
rs1050136144 Chr.8: 27461338-27461338 on Build GRCh38
rs934551659 Chr.8: 27461606-27461606 on Build GRCh38
rs199993564 Chr.8: 27461650-27461650 on Build GRCh38
rs924571569 Chr.8: 27461318-27461318 on Build GRCh38
rs776005376 Chr.8: 27461619-27461619 on Build GRCh38
rs1050962776 Chr.8: 27461304-27461304 on Build GRCh38
rs902992226 Chr.8: 27461391-27461391 on Build GRCh38
rs780560860 Chr.8: 27461574-27461574 on Build GRCh38
rs1044177621 Chr.8: 27461424-27461424 on Build GRCh38
rs147882344 Chr.8: 27461521-27461521 on Build GRCh38
rs748966513 Chr.8: 27461599-27461599 on Build GRCh38
rs762018858 Chr.8: 27461645-27461645 on Build GRCh38
rs560295005 Chr.8: 27461519-27461519 on Build GRCh38
rs535402948 Chr.8: 27461483-27461483 on Build GRCh38
rs76428534 Chr.8: 27461245-27461245 on Build GRCh38
rs978024767 Chr.8: 27461583-27461583 on Build GRCh38
rs755631456 Chr.8: 27461667-27461667 on Build GRCh38
rs542172546 Chr.8: 27461243-27461243 on Build GRCh38
rs930285465 Chr.8: 27461281-27461281 on Build GRCh38
rs1038996400 Chr.8: 27461254-27461254 on Build GRCh38
rs370844410 Chr.8: 27461336-27461336 on Build GRCh38
rs943412642 Chr.8: 27461251-27461251 on Build GRCh38
rs145238683 Chr.8: 27461651-27461651 on Build GRCh38
rs964926183 Chr.8: 27461517-27461517 on Build GRCh38
rs570546296 Chr.8: 27461248-27461248 on Build GRCh38
rs779112876 Chr.8: 27461615-27461615 on Build GRCh38
rs572033587 Chr.8: 27461518-27461518 on Build GRCh38
rs752689106 Chr.8: 27461662-27461662 on Build GRCh38
rs577770461 Chr.8: 27461525-27461525 on Build GRCh38
rs1040135347 Chr.8: 27461644-27461644 on Build GRCh38
rs1026564153 Chr.8: 27461470-27461470 on Build GRCh38
rs974292330 Chr.8: 27461582-27461582 on Build GRCh38
rs777504286 Chr.8: 27461596-27461596 on Build GRCh38
rs569975109 Chr.8: 27461354-27461354 on Build GRCh38
rs908646142 Chr.8: 27461554-27461554 on Build GRCh38
rs111565880 Chr.8: 27461344-27461344 on Build GRCh38
rs759372688 Chr.8: 27461488-27461488 on Build GRCh38
rs752061425 Chr.8: 27461417-27461417 on Build GRCh38
rs950904165 Chr.8: 27461480-27461480 on Build GRCh38
rs866429918 Chr.8: 27461280-27461280 on Build GRCh38
rs950852359 Chr.8: 27461220-27461220 on Build GRCh38
rs112062126 Chr.8: 27461287-27461287 on Build GRCh38
rs753434826 Chr.8: 27461672-27461672 on Build GRCh38
rs772407561 Chr.8: 27461618-27461618 on Build GRCh38
rs770505681 Chr.8: 27461621-27461621 on Build GRCh38
rs906848200 Chr.8: 27461440-27461440 on Build GRCh38
rs77335612 Chr.8: 27461233-27461233 on Build GRCh38
rs971488359 Chr.8: 27461221-27461221 on Build GRCh38
rs760680104 Chr.8: 27461621-27461621 on Build GRCh38
rs998718351 Chr.8: 27461396-27461396 on Build GRCh38
rs768734385 Chr.8: 27461626-27461626 on Build GRCh38
rs75394008 Chr.8: 27461575-27461575 on Build GRCh38
rs920217846 Chr.8: 27461257-27461257 on Build GRCh38
rs552243531 Chr.8: 27461249-27461249 on Build GRCh38
rs888852413 Chr.8: 27461343-27461343 on Build GRCh38
rs368132207 Chr.8: 27461627-27461627 on Build GRCh38
rs1002574858 Chr.8: 27461451-27461451 on Build GRCh38
rs777412930 Chr.8: 27461668-27461668 on Build GRCh38
rs375304755 Chr.8: 27461625-27461625 on Build GRCh38
rs1040147966 Chr.8: 27461372-27461372 on Build GRCh38
rs555088381 Chr.8: 27461497-27461497 on Build GRCh38
rs186509115 Chr.8: 27461600-27461600 on Build GRCh38
rs1022931614 Chr.8: 27461481-27461481 on Build GRCh38
rs906989445 Chr.8: 27461677-27461677 on Build GRCh38
rs746289570 Chr.8: 27461616-27461616 on Build GRCh38
rs957351653 Chr.8: 27461456-27461456 on Build GRCh38
rs767320056 Chr.8: 27461654-27461654 on Build GRCh38
rs141508075 Chr.8: 27461592-27461592 on Build GRCh38
rs981468883 Chr.8: 27461534-27461534 on Build GRCh38
rs534403222 Chr.8: 27461520-27461520 on Build GRCh38
rs1013326490 Chr.8: 27461471-27461471 on Build GRCh38
rs1016411801 Chr.8: 27461506-27461506 on Build GRCh38
rs368039286 Chr.8: 27461447-27461447 on Build GRCh38

View Full Product Details