Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27490922-27491431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX2
Entrez Gene ID
2053
Gene Name
epoxide hydrolase 2
Gene Aliases
CEH, SEH
UniGene
Hs.212088
Gene Chromosome Location
Chr.8: 27491002-27597559 on Build GRCh38
Transcripts
NM_001256482.1 NM_001256483.1 NM_001256484.1 NM_001979.5 XM_017013199.1 XM_017013200.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5358837 c.63C>A p.G21G Chr.8: 27491271-27491271 on Build GRCh38
5651741 c.17C>T p.A6V Chr.8: 27491225-27491225 on Build GRCh38
6655313 c.84G>A p.E28E Chr.8: 27491292-27491292 on Build GRCh38
1623881 c.16G>A p.A6T Chr.8: 27491224-27491224 on Build GRCh38
3925096 c.35G>A p.G12E Chr.8: 27491243-27491243 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763888849 Chr.8: 27491285-27491285 on Build GRCh38
rs567677146 Chr.8: 27491102-27491102 on Build GRCh38
rs982996777 Chr.8: 27491191-27491191 on Build GRCh38
rs750640017 Chr.8: 27491222-27491222 on Build GRCh38
rs921694046 Chr.8: 27491108-27491108 on Build GRCh38
rs778657890 Chr.8: 27491194-27491194 on Build GRCh38
rs752596927 Chr.8: 27491174-27491174 on Build GRCh38
rs962189445 Chr.8: 27491163-27491163 on Build GRCh38
rs764546851 Chr.8: 27491181-27491203 on Build GRCh38
rs550501190 Chr.8: 27491080-27491080 on Build GRCh38
rs945892982 Chr.8: 27491259-27491259 on Build GRCh38
rs962506425 Chr.8: 27491324-27491324 on Build GRCh38
rs889608638 Chr.8: 27490945-27490945 on Build GRCh38
rs778334106 Chr.8: 27491241-27491241 on Build GRCh38
rs1020278201 Chr.8: 27490949-27490949 on Build GRCh38
rs927482301 Chr.8: 27491067-27491067 on Build GRCh38
rs756108612 Chr.8: 27491176-27491176 on Build GRCh38
rs72473927 Chr.8: 27491050-27491050 on Build GRCh38
rs769301606 Chr.8: 27491209-27491209 on Build GRCh38
rs72473931 Chr.8: 27491377-27491377 on Build GRCh38
rs951527124 Chr.8: 27491183-27491183 on Build GRCh38
rs979005914 Chr.8: 27491055-27491055 on Build GRCh38
rs755984255 Chr.8: 27491227-27491227 on Build GRCh38
rs774010128 Chr.8: 27490976-27490976 on Build GRCh38
rs905914545 Chr.8: 27491199-27491199 on Build GRCh38
rs752596505 Chr.8: 27491340-27491340 on Build GRCh38
rs1041204418 Chr.8: 27491120-27491120 on Build GRCh38
rs973499372 Chr.8: 27491203-27491203 on Build GRCh38
rs773228498 Chr.8: 27491268-27491268 on Build GRCh38
rs1034615886 Chr.8: 27491261-27491261 on Build GRCh38
rs921691798 Chr.8: 27491204-27491204 on Build GRCh38
rs371631220 Chr.8: 27491226-27491226 on Build GRCh38
rs775195378 Chr.8: 27491275-27491275 on Build GRCh38
rs1000481380 Chr.8: 27491141-27491141 on Build GRCh38
rs922491873 Chr.8: 27491389-27491389 on Build GRCh38
rs904499226 Chr.8: 27491131-27491131 on Build GRCh38
rs115318000 Chr.8: 27491037-27491037 on Build GRCh38
rs532505914 Chr.8: 27491368-27491368 on Build GRCh38
rs759850446 Chr.8: 27491239-27491239 on Build GRCh38
rs959072192 Chr.8: 27491157-27491157 on Build GRCh38
rs865942990 Chr.8: 27491125-27491125 on Build GRCh38
rs1014415902 Chr.8: 27491165-27491165 on Build GRCh38
rs1056232768 Chr.8: 27491342-27491342 on Build GRCh38
rs536308432 Chr.8: 27491100-27491100 on Build GRCh38
rs1041620379 Chr.8: 27491127-27491127 on Build GRCh38
rs764791006 Chr.8: 27491345-27491345 on Build GRCh38
rs774558387 Chr.8: 27491344-27491344 on Build GRCh38
rs776509267 Chr.8: 27491213-27491213 on Build GRCh38
rs561296411 Chr.8: 27491217-27491217 on Build GRCh38
rs72473929 Chr.8: 27491109-27491109 on Build GRCh38
rs575890391 Chr.8: 27491159-27491159 on Build GRCh38
rs904183879 Chr.8: 27491134-27491134 on Build GRCh38
rs777221050 Chr.8: 27491262-27491262 on Build GRCh38
rs777767435 Chr.8: 27491228-27491228 on Build GRCh38
rs781016516 Chr.8: 27491173-27491173 on Build GRCh38
rs760494070 Chr.8: 27491277-27491277 on Build GRCh38
rs553614248 Chr.8: 27491244-27491244 on Build GRCh38
rs72473930 Chr.8: 27491270-27491270 on Build GRCh38
rs1000247848 Chr.8: 27491010-27491010 on Build GRCh38
rs765504821 Chr.8: 27491158-27491158 on Build GRCh38
rs1024516860 Chr.8: 27491175-27491175 on Build GRCh38
rs1019341318 Chr.8: 27490985-27490985 on Build GRCh38
rs72473928 Chr.8: 27491098-27491098 on Build GRCh38
rs753247698 Chr.8: 27491232-27491232 on Build GRCh38
rs747384326 Chr.8: 27491248-27491248 on Build GRCh38
rs1034229629 Chr.8: 27491373-27491390 on Build GRCh38
rs375398506 Chr.8: 27491265-27491265 on Build GRCh38
rs1036323631 Chr.8: 27491082-27491082 on Build GRCh38
rs965434324 Chr.8: 27491013-27491013 on Build GRCh38
rs1041477228 Chr.8: 27491236-27491236 on Build GRCh38
rs866460354 Chr.8: 27490959-27490959 on Build GRCh38
rs1040846410 Chr.8: 27491205-27491205 on Build GRCh38
rs1020001183 Chr.8: 27491133-27491133 on Build GRCh38
rs201585078 Chr.8: 27491253-27491253 on Build GRCh38
rs567156447 Chr.8: 27491091-27491091 on Build GRCh38
rs748840273 Chr.8: 27491189-27491189 on Build GRCh38
rs1052063568 Chr.8: 27491118-27491118 on Build GRCh38
rs2234912 Chr.8: 27491271-27491271 on Build GRCh38
rs75302548 Chr.8: 27491156-27491156 on Build GRCh38
rs972146085 Chr.8: 27491328-27491328 on Build GRCh38
rs777247946 Chr.8: 27491181-27491181 on Build GRCh38
rs780508291 Chr.8: 27491245-27491245 on Build GRCh38
rs772540676 Chr.8: 27491198-27491198 on Build GRCh38
rs890366618 Chr.8: 27491121-27491121 on Build GRCh38
rs371669961 Chr.8: 27491027-27491027 on Build GRCh38
rs776066069 Chr.8: 27491200-27491200 on Build GRCh38
rs746168469 Chr.8: 27491197-27491197 on Build GRCh38
rs983112812 Chr.8: 27491076-27491076 on Build GRCh38
rs1034583809 Chr.8: 27491142-27491142 on Build GRCh38
rs753790720 Chr.8: 27491295-27491295 on Build GRCh38
rs951642521 Chr.8: 27491074-27491074 on Build GRCh38
rs376357487 Chr.8: 27491362-27491362 on Build GRCh38
rs968773209 Chr.8: 27490979-27490979 on Build GRCh38
rs757180889 Chr.8: 27491334-27491334 on Build GRCh38
rs544582307 Chr.8: 27491160-27491160 on Build GRCh38
rs150415266 Chr.8: 27490949-27490950 on Build GRCh38
rs749968315 Chr.8: 27491352-27491352 on Build GRCh38
rs367892722 Chr.8: 27491202-27491202 on Build GRCh38
rs551191344 Chr.8: 27491021-27491021 on Build GRCh38
rs770423406 Chr.8: 27491193-27491193 on Build GRCh38
rs371664562 Chr.8: 27491150-27491150 on Build GRCh38

View Full Product Details