Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27503578-27504087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX2
Entrez Gene ID
2053
Gene Name
epoxide hydrolase 2
Gene Aliases
CEH, SEH
UniGene
Hs.212088
Gene Chromosome Location
Chr.8: 27491002-27597559 on Build GRCh38
Transcripts
NM_001256482.1 NM_001256483.1 NM_001256484.1 NM_001979.5 XM_017013199.1 XM_017013200.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
749941 c.193C>A p.P65T Chr.8: 27503610-27503610 on Build GRCh38
5922676 c.346+5G>A p.? Chr.8: 27503768-27503768 on Build GRCh38
1313973 c.339G>T p.R113S Chr.8: 27503756-27503756 on Build GRCh38
5938212 c.247C>T p.P83S Chr.8: 27503664-27503664 on Build GRCh38
1313972 c.318C>T p.L106L Chr.8: 27503735-27503735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950333274 Chr.8: 27503856-27503856 on Build GRCh38
rs747196050 Chr.8: 27503704-27503704 on Build GRCh38
rs367992021 Chr.8: 27503872-27503872 on Build GRCh38
rs530604104 Chr.8: 27503875-27503875 on Build GRCh38
rs201924747 Chr.8: 27503774-27503774 on Build GRCh38
rs745941583 Chr.8: 27503618-27503618 on Build GRCh38
rs775132454 Chr.8: 27503782-27503782 on Build GRCh38
rs770949786 Chr.8: 27503759-27503759 on Build GRCh38
rs747308108 Chr.8: 27503637-27503637 on Build GRCh38
rs17057255 Chr.8: 27503724-27503724 on Build GRCh38
rs762916759 Chr.8: 27503795-27503795 on Build GRCh38
rs760755782 Chr.8: 27503726-27503726 on Build GRCh38
rs778908072 Chr.8: 27503609-27503609 on Build GRCh38
rs766377908 Chr.8: 27503752-27503753 on Build GRCh38
rs977518725 Chr.8: 27503776-27503776 on Build GRCh38
rs899634859 Chr.8: 27504026-27504026 on Build GRCh38
rs991577298 Chr.8: 27503997-27503997 on Build GRCh38
rs908765054 Chr.8: 27504027-27504027 on Build GRCh38
rs575264078 Chr.8: 27504016-27504016 on Build GRCh38
rs780226946 Chr.8: 27503702-27503702 on Build GRCh38
rs777655940 Chr.8: 27503727-27503727 on Build GRCh38
rs376705396 Chr.8: 27503661-27503661 on Build GRCh38
rs929259952 Chr.8: 27503966-27503966 on Build GRCh38
rs761816520 Chr.8: 27503844-27503844 on Build GRCh38
rs28556875 Chr.8: 27503899-27503899 on Build GRCh38
rs146918355 Chr.8: 27503683-27503683 on Build GRCh38
rs142029565 Chr.8: 27503725-27503725 on Build GRCh38
rs536862077 Chr.8: 27503631-27503631 on Build GRCh38
rs4149238 Chr.8: 27503835-27503835 on Build GRCh38
rs903195208 Chr.8: 27503968-27503968 on Build GRCh38
rs971516101 Chr.8: 27503999-27503999 on Build GRCh38
rs764435677 Chr.8: 27503675-27503675 on Build GRCh38
rs750645541 Chr.8: 27503882-27503882 on Build GRCh38
rs981672228 Chr.8: 27504022-27504022 on Build GRCh38
rs1018848807 Chr.8: 27503967-27503967 on Build GRCh38
rs750468213 Chr.8: 27503687-27503687 on Build GRCh38
rs72475809 Chr.8: 27503902-27503902 on Build GRCh38
rs760306398 Chr.8: 27503787-27503787 on Build GRCh38
rs1005421165 Chr.8: 27503788-27503788 on Build GRCh38
rs553359086 Chr.8: 27503642-27503642 on Build GRCh38
rs776277631 Chr.8: 27503693-27503693 on Build GRCh38
rs997963807 Chr.8: 27504036-27504036 on Build GRCh38
rs760415141 Chr.8: 27503665-27503665 on Build GRCh38
rs372389894 Chr.8: 27503762-27503762 on Build GRCh38
rs778159836 Chr.8: 27503602-27503602 on Build GRCh38
rs747123450 Chr.8: 27503873-27503873 on Build GRCh38
rs758386893 Chr.8: 27503701-27503701 on Build GRCh38
rs377439293 Chr.8: 27503652-27503652 on Build GRCh38
rs752269686 Chr.8: 27503837-27503837 on Build GRCh38
rs545029636 Chr.8: 27503679-27503679 on Build GRCh38
rs1051246215 Chr.8: 27504038-27504038 on Build GRCh38
rs762189404 Chr.8: 27503800-27503800 on Build GRCh38
rs759198021 Chr.8: 27503606-27503606 on Build GRCh38
rs141148096 Chr.8: 27503648-27503648 on Build GRCh38
rs141863165 Chr.8: 27503933-27503933 on Build GRCh38
rs888941714 Chr.8: 27503937-27503937 on Build GRCh38
rs542194263 Chr.8: 27503956-27503956 on Build GRCh38
rs992109537 Chr.8: 27503817-27503817 on Build GRCh38
rs1033013483 Chr.8: 27503976-27503976 on Build GRCh38
rs771451583 Chr.8: 27503781-27503781 on Build GRCh38
rs372597795 Chr.8: 27503649-27503649 on Build GRCh38
rs754312530 Chr.8: 27503604-27503604 on Build GRCh38
rs573493340 Chr.8: 27503677-27503677 on Build GRCh38
rs745864157 Chr.8: 27503779-27503779 on Build GRCh38
rs17057269 Chr.8: 27503812-27503812 on Build GRCh38
rs372745275 Chr.8: 27503932-27503932 on Build GRCh38
rs780162329 Chr.8: 27503632-27503632 on Build GRCh38
rs559345117 Chr.8: 27503813-27503813 on Build GRCh38
rs374746559 Chr.8: 27503643-27503643 on Build GRCh38
rs17057262 Chr.8: 27503794-27503794 on Build GRCh38
rs899602797 Chr.8: 27503803-27503803 on Build GRCh38
rs558666351 Chr.8: 27503998-27503998 on Build GRCh38
rs765765941 Chr.8: 27503686-27503686 on Build GRCh38
rs528390099 Chr.8: 27503824-27503824 on Build GRCh38
rs369978603 Chr.8: 27503662-27503662 on Build GRCh38
rs765640978 Chr.8: 27503808-27503808 on Build GRCh38
rs769613726 Chr.8: 27503764-27503764 on Build GRCh38
rs957722712 Chr.8: 27503980-27503980 on Build GRCh38
rs888290322 Chr.8: 27503770-27503770 on Build GRCh38
rs749096467 Chr.8: 27503750-27503750 on Build GRCh38
rs550721510 Chr.8: 27503881-27503881 on Build GRCh38
rs771002906 Chr.8: 27503645-27503645 on Build GRCh38
rs752661968 Chr.8: 27503754-27503761 on Build GRCh38
rs940364109 Chr.8: 27504055-27504055 on Build GRCh38
rs998819887 Chr.8: 27503974-27503974 on Build GRCh38
rs778894848 Chr.8: 27503639-27503639 on Build GRCh38

View Full Product Details