Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27604053-27604456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLU
Entrez Gene ID
1191
Gene Name
clusterin
Gene Aliases
AAG4, APO-J, APOJ, CLI, CLU1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2
UniGene
Hs.436657
Gene Chromosome Location
Chr.8: 27596917-27616717 on Build GRCh38
Transcripts
NM_001831.3 NR_038335.1 NR_045494.1 XM_006716284.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361613 c.1004C>T p.T335I Chr.8: 27604377-27604377 on Build GRCh38
5485011 c.1022G>A p.R341H Chr.8: 27604359-27604359 on Build GRCh38
3647824 c.994G>A p.D332N Chr.8: 27604387-27604387 on Build GRCh38
6215177 c.986A>G p.E329G Chr.8: 27604395-27604395 on Build GRCh38
5420961 c.1046G>T p.R349L Chr.8: 27604335-27604335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776894641 Chr.8: 27604435-27604435 on Build GRCh38
rs754372820 Chr.8: 27604293-27604293 on Build GRCh38
rs930142450 Chr.8: 27604167-27604167 on Build GRCh38
rs563432916 Chr.8: 27604347-27604347 on Build GRCh38
rs769745301 Chr.8: 27604259-27604259 on Build GRCh38
rs150552076 Chr.8: 27604377-27604377 on Build GRCh38
rs775045101 Chr.8: 27604359-27604359 on Build GRCh38
rs964803949 Chr.8: 27604091-27604091 on Build GRCh38
rs528753124 Chr.8: 27604387-27604387 on Build GRCh38
rs933127763 Chr.8: 27604141-27604141 on Build GRCh38
rs777141082 Chr.8: 27604366-27604366 on Build GRCh38
rs923117356 Chr.8: 27604135-27604135 on Build GRCh38
rs771775730 Chr.8: 27604404-27604404 on Build GRCh38
rs754448694 Chr.8: 27604257-27604257 on Build GRCh38
rs762517794 Chr.8: 27604265-27604265 on Build GRCh38
rs549647960 Chr.8: 27604357-27604357 on Build GRCh38
rs759358037 Chr.8: 27604256-27604258 on Build GRCh38
rs552103421 Chr.8: 27604390-27604390 on Build GRCh38
rs9331919 Chr.8: 27604122-27604122 on Build GRCh38
rs770620635 Chr.8: 27604271-27604271 on Build GRCh38
rs1803289 Chr.8: 27604318-27604318 on Build GRCh38
rs765965098 Chr.8: 27604228-27604236 on Build GRCh38
rs374780183 Chr.8: 27604407-27604407 on Build GRCh38
rs777432954 Chr.8: 27604385-27604385 on Build GRCh38
rs766118113 Chr.8: 27604255-27604255 on Build GRCh38
rs139738246 Chr.8: 27604313-27604313 on Build GRCh38
rs770064654 Chr.8: 27604132-27604132 on Build GRCh38
rs758508760 Chr.8: 27604375-27604375 on Build GRCh38
rs778981507 Chr.8: 27604298-27604298 on Build GRCh38
rs202081077 Chr.8: 27604391-27604391 on Build GRCh38
rs9331918 Chr.8: 27604272-27604272 on Build GRCh38
rs80247028 Chr.8: 27604408-27604408 on Build GRCh38
rs777749323 Chr.8: 27604336-27604336 on Build GRCh38
rs947429738 Chr.8: 27604159-27604159 on Build GRCh38
rs111643424 Chr.8: 27604290-27604290 on Build GRCh38
rs192011917 Chr.8: 27604151-27604151 on Build GRCh38
rs761097626 Chr.8: 27604360-27604360 on Build GRCh38
rs759264374 Chr.8: 27604278-27604278 on Build GRCh38
rs529447494 Chr.8: 27604349-27604349 on Build GRCh38
rs764631028 Chr.8: 27604284-27604284 on Build GRCh38
rs757282082 Chr.8: 27604388-27604388 on Build GRCh38
rs745786940 Chr.8: 27604130-27604130 on Build GRCh38
rs757317713 Chr.8: 27604295-27604295 on Build GRCh38
rs3216167 Chr.8: 27604256-27604256 on Build GRCh38
rs200376447 Chr.8: 27604256-27604256 on Build GRCh38
rs143634423 Chr.8: 27604369-27604369 on Build GRCh38
rs909954958 Chr.8: 27604223-27604223 on Build GRCh38
rs750529144 Chr.8: 27604370-27604370 on Build GRCh38
rs988599663 Chr.8: 27604422-27604422 on Build GRCh38
rs140575752 Chr.8: 27604150-27604150 on Build GRCh38
rs139565837 Chr.8: 27604380-27604380 on Build GRCh38
rs572053967 Chr.8: 27604189-27604189 on Build GRCh38
rs954545609 Chr.8: 27604418-27604418 on Build GRCh38
rs755878741 Chr.8: 27604381-27604381 on Build GRCh38
rs561625939 Chr.8: 27604188-27604188 on Build GRCh38
rs764426787 Chr.8: 27604364-27604364 on Build GRCh38
rs767953371 Chr.8: 27604280-27604280 on Build GRCh38
rs753127553 Chr.8: 27604281-27604281 on Build GRCh38
rs372063497 Chr.8: 27604308-27604308 on Build GRCh38
rs543116935 Chr.8: 27604346-27604346 on Build GRCh38
rs200572774 Chr.8: 27604403-27604403 on Build GRCh38
rs905975676 Chr.8: 27604247-27604247 on Build GRCh38
rs372415096 Chr.8: 27604335-27604335 on Build GRCh38
rs374903220 Chr.8: 27604309-27604309 on Build GRCh38
rs540613919 Chr.8: 27604264-27604264 on Build GRCh38

View Full Product Details