Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144079852-144080349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC4
Entrez Gene ID
54512
Gene Name
exosome component 4
Gene Aliases
RRP41, RRP41A, Rrp41p, SKI6, Ski6p, hRrp41p, p12A
UniGene
Hs.632041
Gene Chromosome Location
Chr.8: 144064092-144080648 on Build GRCh38
Transcripts
NM_019037.2 XM_011517134.2 NR_106906.1

Gene Symbol
MIR6847
Entrez Gene ID
102465510
Gene Name
microRNA 6847
Gene Aliases
hsa-mir-6847
UniGene
Gene Chromosome Location
Chr.8: 144079874-144079942 on Build GRCh38
Transcripts
NM_019037.2 XM_011517134.2 NR_106906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1623764 c.200C>G p.P67R Chr.8: 144079971-144079971 on Build GRCh38
3898139 c.425C>T p.A142V Chr.8: 144080288-144080288 on Build GRCh38
2870733 c.201G>A p.P67P Chr.8: 144079972-144079972 on Build GRCh38
6657304 c.281G>A p.R94H Chr.8: 144080052-144080052 on Build GRCh38
1569145 c.311G>A p.R104H Chr.8: 144080082-144080082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200347210 Chr.8: 144080225-144080225 on Build GRCh38
rs781952585 Chr.8: 144080052-144080052 on Build GRCh38
rs782417114 Chr.8: 144080036-144080036 on Build GRCh38
rs924519410 Chr.8: 144079964-144079964 on Build GRCh38
rs782156888 Chr.8: 144080077-144080077 on Build GRCh38
rs782357099 Chr.8: 144080051-144080051 on Build GRCh38
rs11547777 Chr.8: 144080289-144080289 on Build GRCh38
rs782680861 Chr.8: 144079918-144079918 on Build GRCh38
rs200357319 Chr.8: 144080164-144080164 on Build GRCh38
rs781794594 Chr.8: 144080244-144080244 on Build GRCh38
rs782717037 Chr.8: 144080236-144080236 on Build GRCh38
rs887344804 Chr.8: 144080092-144080092 on Build GRCh38
rs782817228 Chr.8: 144080000-144080000 on Build GRCh38
rs782746867 Chr.8: 144079981-144079981 on Build GRCh38
rs201630396 Chr.8: 144080082-144080082 on Build GRCh38
rs375294871 Chr.8: 144080173-144080173 on Build GRCh38
rs781858259 Chr.8: 144080223-144080223 on Build GRCh38
rs782001397 Chr.8: 144079962-144079962 on Build GRCh38
rs200218337 Chr.8: 144080112-144080112 on Build GRCh38
rs782755725 Chr.8: 144079889-144079889 on Build GRCh38
rs782588361 Chr.8: 144079903-144079903 on Build GRCh38
rs782056134 Chr.8: 144079997-144079997 on Build GRCh38
rs987425240 Chr.8: 144080295-144080295 on Build GRCh38
rs782475794 Chr.8: 144079901-144079901 on Build GRCh38
rs782196587 Chr.8: 144080059-144080059 on Build GRCh38
rs782115039 Chr.8: 144080310-144080310 on Build GRCh38
rs782142047 Chr.8: 144080215-144080215 on Build GRCh38
rs782264866 Chr.8: 144080254-144080254 on Build GRCh38
rs914182774 Chr.8: 144079905-144079905 on Build GRCh38
rs201774377 Chr.8: 144080166-144080166 on Build GRCh38
rs782445743 Chr.8: 144080120-144080120 on Build GRCh38
rs782050719 Chr.8: 144080318-144080318 on Build GRCh38
rs184074068 Chr.8: 144080146-144080146 on Build GRCh38
rs367844132 Chr.8: 144080014-144080014 on Build GRCh38
rs373523635 Chr.8: 144080248-144080248 on Build GRCh38
rs782809439 Chr.8: 144080323-144080323 on Build GRCh38
rs11784223 Chr.8: 144080202-144080202 on Build GRCh38
rs781918119 Chr.8: 144079946-144079946 on Build GRCh38
rs144241636 Chr.8: 144080037-144080037 on Build GRCh38
rs376348792 Chr.8: 144080217-144080217 on Build GRCh38
rs782799057 Chr.8: 144080133-144080133 on Build GRCh38
rs782459069 Chr.8: 144080017-144080017 on Build GRCh38
rs782360735 Chr.8: 144080311-144080311 on Build GRCh38
rs781894428 Chr.8: 144080001-144080001 on Build GRCh38
rs782559077 Chr.8: 144079916-144079916 on Build GRCh38
rs782785182 Chr.8: 144080088-144080088 on Build GRCh38
rs782208338 Chr.8: 144079944-144079944 on Build GRCh38
rs782209049 Chr.8: 144080268-144080268 on Build GRCh38
rs782042026 Chr.8: 144080073-144080073 on Build GRCh38
rs151065132 Chr.8: 144080104-144080104 on Build GRCh38
rs781997916 Chr.8: 144080093-144080093 on Build GRCh38
rs201727380 Chr.8: 144080276-144080276 on Build GRCh38
rs782582326 Chr.8: 144080160-144080160 on Build GRCh38
rs782163750 Chr.8: 144079955-144079955 on Build GRCh38
rs782529066 Chr.8: 144080011-144080011 on Build GRCh38
rs782625031 Chr.8: 144080267-144080267 on Build GRCh38
rs372895890 Chr.8: 144079910-144079910 on Build GRCh38
rs782265338 Chr.8: 144079920-144079920 on Build GRCh38
rs782448527 Chr.8: 144080150-144080150 on Build GRCh38
rs116723461 Chr.8: 144080211-144080211 on Build GRCh38
rs371454670 Chr.8: 144079878-144079878 on Build GRCh38
rs782370363 Chr.8: 144079925-144079925 on Build GRCh38
rs939049113 Chr.8: 144080078-144080078 on Build GRCh38
rs782323580 Chr.8: 144080271-144080271 on Build GRCh38
rs201621803 Chr.8: 144080250-144080250 on Build GRCh38
rs910399768 Chr.8: 144080004-144080004 on Build GRCh38
rs375181992 Chr.8: 144079900-144079900 on Build GRCh38
rs201178982 Chr.8: 144080083-144080083 on Build GRCh38
rs139817759 Chr.8: 144080272-144080272 on Build GRCh38
rs1003158290 Chr.8: 144080110-144080110 on Build GRCh38
rs376040142 Chr.8: 144079911-144079911 on Build GRCh38
rs201948066 Chr.8: 144080207-144080207 on Build GRCh38
rs782310434 Chr.8: 144080071-144080071 on Build GRCh38
rs782214487 Chr.8: 144080148-144080148 on Build GRCh38
rs557258449 Chr.8: 144079956-144079956 on Build GRCh38
rs782502364 Chr.8: 144080028-144080028 on Build GRCh38
rs776416022 Chr.8: 144080194-144080194 on Build GRCh38
rs781888886 Chr.8: 144080136-144080136 on Build GRCh38
rs782321329 Chr.8: 144079945-144079945 on Build GRCh38
rs202168311 Chr.8: 144080050-144080050 on Build GRCh38
rs977119657 Chr.8: 144079922-144079922 on Build GRCh38
rs782178166 Chr.8: 144080033-144080033 on Build GRCh38
rs782061268 Chr.8: 144080208-144080208 on Build GRCh38
rs781934139 Chr.8: 144080315-144080315 on Build GRCh38
rs188660237 Chr.8: 144080213-144080213 on Build GRCh38
rs781832243 Chr.8: 144080016-144080016 on Build GRCh38
rs782256108 Chr.8: 144080185-144080185 on Build GRCh38
rs782351959 Chr.8: 144079912-144079912 on Build GRCh38
rs1039793967 Chr.8: 144080027-144080027 on Build GRCh38
rs782095288 Chr.8: 144079975-144079975 on Build GRCh38
rs528562530 Chr.8: 144080262-144080262 on Build GRCh38
rs782635100 Chr.8: 144080147-144080147 on Build GRCh38

View Full Product Details