Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144095755-144096245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYC1
Entrez Gene ID
1537
Gene Name
cytochrome c1
Gene Aliases
MC3DN6, UQCR4
UniGene
Hs.289271
Gene Chromosome Location
Chr.8: 144095035-144097527 on Build GRCh38
Transcripts
NM_001916.4 XM_017013102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6638786 c.219G>A p.G73G Chr.8: 144095922-144095922 on Build GRCh38
4655420 c.418T>C p.Y140H Chr.8: 144096215-144096215 on Build GRCh38
6638785 c.347_348delTA p.Y117fs*1 Chr.8: 144096144-144096145 on Build GRCh38
4675800 c.375C>T p.H125H Chr.8: 144096172-144096172 on Build GRCh38
4675799 c.144G>T p.S48S Chr.8: 144095847-144095847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375444687 Chr.8: 144096160-144096160 on Build GRCh38
rs745923518 Chr.8: 144095807-144095807 on Build GRCh38
rs750656085 Chr.8: 144095987-144095987 on Build GRCh38
rs587777041 Chr.8: 144095991-144095991 on Build GRCh38
rs775059723 Chr.8: 144096175-144096175 on Build GRCh38
rs773644422 Chr.8: 144096184-144096184 on Build GRCh38
rs764034811 Chr.8: 144095785-144095797 on Build GRCh38
rs1016550663 Chr.8: 144095836-144095836 on Build GRCh38
rs767309788 Chr.8: 144096221-144096221 on Build GRCh38
rs770580594 Chr.8: 144095958-144095958 on Build GRCh38
rs34079907 Chr.8: 144096163-144096163 on Build GRCh38
rs780077264 Chr.8: 144095979-144095980 on Build GRCh38
rs376311407 Chr.8: 144095816-144095816 on Build GRCh38
rs750485779 Chr.8: 144095931-144095931 on Build GRCh38
rs747902761 Chr.8: 144095834-144095834 on Build GRCh38
rs748108420 Chr.8: 144095942-144095942 on Build GRCh38
rs767640884 Chr.8: 144095986-144095986 on Build GRCh38
rs751483824 Chr.8: 144096130-144096130 on Build GRCh38
rs781009717 Chr.8: 144096156-144096156 on Build GRCh38
rs759668497 Chr.8: 144096200-144096200 on Build GRCh38
rs997489723 Chr.8: 144095805-144095805 on Build GRCh38
rs2230226 Chr.8: 144095962-144095962 on Build GRCh38
rs374210188 Chr.8: 144096036-144096036 on Build GRCh38
rs139470418 Chr.8: 144095918-144095918 on Build GRCh38
rs970947838 Chr.8: 144096155-144096155 on Build GRCh38
rs753816240 Chr.8: 144096006-144096006 on Build GRCh38
rs200190295 Chr.8: 144096114-144096114 on Build GRCh38
rs780764166 Chr.8: 144096052-144096052 on Build GRCh38
rs199600426 Chr.8: 144095790-144095790 on Build GRCh38
rs775910166 Chr.8: 144095964-144095964 on Build GRCh38
rs981713041 Chr.8: 144095913-144095913 on Build GRCh38
rs866342331 Chr.8: 144095912-144095912 on Build GRCh38
rs1041794561 Chr.8: 144095783-144095783 on Build GRCh38
rs757463304 Chr.8: 144095803-144095803 on Build GRCh38
rs1003226376 Chr.8: 144096192-144096192 on Build GRCh38
rs201814058 Chr.8: 144096223-144096223 on Build GRCh38
rs144257411 Chr.8: 144095930-144095930 on Build GRCh38
rs751437819 Chr.8: 144095844-144095846 on Build GRCh38
rs528015421 Chr.8: 144095967-144095967 on Build GRCh38
rs538330029 Chr.8: 144096113-144096113 on Build GRCh38
rs768280243 Chr.8: 144096166-144096166 on Build GRCh38
rs751871692 Chr.8: 144095928-144095928 on Build GRCh38
rs769301920 Chr.8: 144096075-144096075 on Build GRCh38
rs757050208 Chr.8: 144096145-144096145 on Build GRCh38
rs564652078 Chr.8: 144095922-144095922 on Build GRCh38
rs756155043 Chr.8: 144095935-144095935 on Build GRCh38
rs746809086 Chr.8: 144096018-144096018 on Build GRCh38
rs781390871 Chr.8: 144095804-144095804 on Build GRCh38
rs764532121 Chr.8: 144095927-144095927 on Build GRCh38
rs902075706 Chr.8: 144096117-144096117 on Build GRCh38
rs747220864 Chr.8: 144095792-144095792 on Build GRCh38
rs769227684 Chr.8: 144096058-144096058 on Build GRCh38
rs756210302 Chr.8: 144095993-144095993 on Build GRCh38
rs772910528 Chr.8: 144095850-144095850 on Build GRCh38
rs756254049 Chr.8: 144095972-144095973 on Build GRCh38
rs369673219 Chr.8: 144096087-144096087 on Build GRCh38
rs755814580 Chr.8: 144096161-144096161 on Build GRCh38
rs368107185 Chr.8: 144096057-144096057 on Build GRCh38
rs372663686 Chr.8: 144096224-144096224 on Build GRCh38
rs758350634 Chr.8: 144096009-144096009 on Build GRCh38
rs567693962 Chr.8: 144096096-144096096 on Build GRCh38
rs372218907 Chr.8: 144096070-144096070 on Build GRCh38
rs775097630 Chr.8: 144096060-144096060 on Build GRCh38
rs771853926 Chr.8: 144095843-144095843 on Build GRCh38
rs776054597 Chr.8: 144095938-144095938 on Build GRCh38
rs981802685 Chr.8: 144095970-144095970 on Build GRCh38
rs887935795 Chr.8: 144096177-144096177 on Build GRCh38
rs778208336 Chr.8: 144095791-144095791 on Build GRCh38
rs775233321 Chr.8: 144095871-144095871 on Build GRCh38
rs538272583 Chr.8: 144095949-144095949 on Build GRCh38
rs760285719 Chr.8: 144095853-144095853 on Build GRCh38
rs146030978 Chr.8: 144095892-144095892 on Build GRCh38
rs748482749 Chr.8: 144096037-144096040 on Build GRCh38
rs11541474 Chr.8: 144096023-144096023 on Build GRCh38
rs767914902 Chr.8: 144096094-144096094 on Build GRCh38
rs142370723 Chr.8: 144096129-144096129 on Build GRCh38
rs748813455 Chr.8: 144096164-144096164 on Build GRCh38
rs761104284 Chr.8: 144096208-144096208 on Build GRCh38
rs763522648 Chr.8: 144095965-144095965 on Build GRCh38
rs772367297 Chr.8: 144096181-144096181 on Build GRCh38
rs1007802953 Chr.8: 144095829-144095829 on Build GRCh38
rs371892448 Chr.8: 144096120-144096120 on Build GRCh38
rs993657678 Chr.8: 144096098-144096098 on Build GRCh38
rs77877372 Chr.8: 144095957-144095957 on Build GRCh38
rs748647625 Chr.8: 144096179-144096179 on Build GRCh38
rs751996245 Chr.8: 144095973-144095973 on Build GRCh38
rs771890509 Chr.8: 144095947-144095947 on Build GRCh38
rs145609083 Chr.8: 144096026-144096026 on Build GRCh38
rs375200496 Chr.8: 144096077-144096077 on Build GRCh38
rs766624873 Chr.8: 144096199-144096199 on Build GRCh38
rs754110346 Chr.8: 144096003-144096005 on Build GRCh38
rs780163811 Chr.8: 144096003-144096003 on Build GRCh38
rs7820984 Chr.8: 144095929-144095929 on Build GRCh38
rs200527190 Chr.8: 144095961-144095961 on Build GRCh38
rs903275893 Chr.8: 144095793-144095793 on Build GRCh38
rs754982364 Chr.8: 144096118-144096118 on Build GRCh38
rs768501982 Chr.8: 144095812-144095812 on Build GRCh38
rs757249899 Chr.8: 144096220-144096220 on Build GRCh38
rs1041034392 Chr.8: 144096101-144096101 on Build GRCh38
rs778408162 Chr.8: 144096113-144096114 on Build GRCh38
rs779179147 Chr.8: 144096036-144096037 on Build GRCh38
rs993592660 Chr.8: 144095867-144095867 on Build GRCh38
rs560816891 Chr.8: 144096036-144096039 on Build GRCh38
rs750664658 Chr.8: 144096100-144096100 on Build GRCh38
rs368429357 Chr.8: 144095857-144095857 on Build GRCh38
rs889103478 Chr.8: 144096051-144096051 on Build GRCh38
rs751537090 Chr.8: 144096209-144096209 on Build GRCh38
rs753465411 Chr.8: 144095936-144095936 on Build GRCh38
rs760887176 Chr.8: 144096198-144096198 on Build GRCh38
rs142635756 Chr.8: 144095886-144095886 on Build GRCh38

View Full Product Details