Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144392877-144393368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1 NM_013291.2 XM_006716548.2 XM_006716549.2 XM_006716550.3 XM_011516997.1 XM_011516998.1 XM_011516999.1

Gene Symbol
CPSF1
Entrez Gene ID
29894
Gene Name
cleavage and polyadenylation specific factor 1
Gene Aliases
CPSF160, HSU37012, P/cl.18
UniGene
Hs.493202
Gene Chromosome Location
Chr.8: 144393231-144409400 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1 NM_013291.2 XM_006716548.2 XM_006716549.2 XM_006716550.3 XM_011516997.1 XM_011516998.1 XM_011516999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420154 c.1664C>A p.T555N Chr.8: 144392995-144392995 on Build GRCh38
4150660 c.1638-9C>G p.? Chr.8: 144392960-144392960 on Build GRCh38
5345549 c.1730A>G p.Y577C Chr.8: 144393061-144393061 on Build GRCh38
3785910 c.1679G>A p.R560H Chr.8: 144393010-144393010 on Build GRCh38
1569141 c.1665C>T p.T555T Chr.8: 144392996-144392996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782596425 Chr.8: 144392918-144392918 on Build GRCh38
rs1052951 Chr.8: 144393094-144393094 on Build GRCh38
rs782798899 Chr.8: 144393116-144393116 on Build GRCh38
rs781793852 Chr.8: 144393293-144393293 on Build GRCh38
rs3817680 Chr.8: 144393222-144393222 on Build GRCh38
rs782471642 Chr.8: 144393213-144393213 on Build GRCh38
rs574946342 Chr.8: 144393085-144393085 on Build GRCh38
rs782151477 Chr.8: 144392925-144392925 on Build GRCh38
rs782730247 Chr.8: 144393289-144393289 on Build GRCh38
rs367710887 Chr.8: 144393114-144393114 on Build GRCh38
rs375914315 Chr.8: 144392905-144392905 on Build GRCh38
rs923055656 Chr.8: 144393170-144393170 on Build GRCh38
rs780902842 Chr.8: 144393280-144393280 on Build GRCh38
rs782015570 Chr.8: 144393121-144393121 on Build GRCh38
rs782659374 Chr.8: 144392907-144392907 on Build GRCh38
rs782722331 Chr.8: 144393075-144393075 on Build GRCh38
rs377619920 Chr.8: 144392952-144392952 on Build GRCh38
rs782052942 Chr.8: 144392960-144392960 on Build GRCh38
rs184127179 Chr.8: 144392921-144392921 on Build GRCh38
rs1052952 Chr.8: 144393097-144393097 on Build GRCh38
rs782230569 Chr.8: 144393107-144393107 on Build GRCh38
rs782448360 Chr.8: 144393096-144393096 on Build GRCh38
rs372703345 Chr.8: 144392909-144392909 on Build GRCh38
rs782568276 Chr.8: 144393298-144393298 on Build GRCh38
rs556789935 Chr.8: 144393083-144393083 on Build GRCh38
rs782077881 Chr.8: 144393074-144393074 on Build GRCh38
rs782601836 Chr.8: 144393327-144393327 on Build GRCh38
rs546616154 Chr.8: 144393343-144393343 on Build GRCh38
rs782038301 Chr.8: 144393047-144393047 on Build GRCh38
rs782407174 Chr.8: 144393099-144393099 on Build GRCh38
rs957059223 Chr.8: 144392935-144392935 on Build GRCh38
rs781978463 Chr.8: 144393100-144393100 on Build GRCh38
rs534948719 Chr.8: 144392981-144392981 on Build GRCh38
rs782479937 Chr.8: 144393284-144393284 on Build GRCh38
rs782146208 Chr.8: 144392912-144392913 on Build GRCh38
rs782753878 Chr.8: 144393056-144393056 on Build GRCh38
rs781954516 Chr.8: 144393035-144393035 on Build GRCh38
rs975831854 Chr.8: 144393153-144393153 on Build GRCh38
rs1038344790 Chr.8: 144393223-144393223 on Build GRCh38
rs782132943 Chr.8: 144393048-144393048 on Build GRCh38
rs201892829 Chr.8: 144393339-144393339 on Build GRCh38
rs535421070 Chr.8: 144393038-144393038 on Build GRCh38
rs1045792539 Chr.8: 144393257-144393257 on Build GRCh38
rs782195565 Chr.8: 144392920-144392920 on Build GRCh38
rs782244827 Chr.8: 144393318-144393318 on Build GRCh38
rs781876187 Chr.8: 144393087-144393087 on Build GRCh38
rs371037595 Chr.8: 144392993-144392993 on Build GRCh38
rs369068642 Chr.8: 144392906-144392906 on Build GRCh38
rs782359250 Chr.8: 144392913-144392913 on Build GRCh38
rs782140576 Chr.8: 144393122-144393122 on Build GRCh38
rs200275320 Chr.8: 144392939-144392940 on Build GRCh38
rs201933662 Chr.8: 144393098-144393098 on Build GRCh38
rs911460378 Chr.8: 144393216-144393216 on Build GRCh38
rs373822279 Chr.8: 144392951-144392951 on Build GRCh38
rs781990512 Chr.8: 144392945-144392945 on Build GRCh38
rs782291276 Chr.8: 144393021-144393021 on Build GRCh38
rs369037664 Chr.8: 144392946-144392946 on Build GRCh38
rs541353407 Chr.8: 144393125-144393125 on Build GRCh38
rs922773153 Chr.8: 144393004-144393004 on Build GRCh38
rs1042549844 Chr.8: 144392997-144392997 on Build GRCh38
rs781847185 Chr.8: 144393271-144393271 on Build GRCh38
rs139161523 Chr.8: 144392969-144392969 on Build GRCh38
rs530081948 Chr.8: 144393157-144393157 on Build GRCh38
rs201707698 Chr.8: 144393325-144393325 on Build GRCh38
rs781836012 Chr.8: 144393057-144393057 on Build GRCh38
rs370327410 Chr.8: 144393115-144393115 on Build GRCh38
rs782269383 Chr.8: 144393344-144393344 on Build GRCh38
rs782566896 Chr.8: 144393275-144393275 on Build GRCh38
rs570128839 Chr.8: 144392902-144392902 on Build GRCh38
rs782311286 Chr.8: 144393041-144393041 on Build GRCh38
rs373819051 Chr.8: 144393110-144393110 on Build GRCh38
rs934138752 Chr.8: 144393194-144393194 on Build GRCh38
rs563635544 Chr.8: 144393244-144393244 on Build GRCh38
rs782660919 Chr.8: 144393304-144393304 on Build GRCh38
rs373042703 Chr.8: 144392958-144392959 on Build GRCh38
rs897155935 Chr.8: 144393226-144393226 on Build GRCh38
rs200014598 Chr.8: 144393330-144393330 on Build GRCh38
rs782195803 Chr.8: 144393333-144393333 on Build GRCh38
rs374047184 Chr.8: 144393342-144393342 on Build GRCh38
rs375437355 Chr.8: 144392964-144392964 on Build GRCh38
rs782796597 Chr.8: 144393104-144393104 on Build GRCh38
rs781841554 Chr.8: 144393200-144393200 on Build GRCh38
rs553208368 Chr.8: 144393030-144393030 on Build GRCh38
rs530999703 Chr.8: 144393287-144393287 on Build GRCh38
rs782413945 Chr.8: 144393025-144393025 on Build GRCh38
rs782616643 Chr.8: 144393093-144393093 on Build GRCh38
rs1001364441 Chr.8: 144393113-144393113 on Build GRCh38
rs782394528 Chr.8: 144392927-144392927 on Build GRCh38
rs540871672 Chr.8: 144393141-144393141 on Build GRCh38
rs376127969 Chr.8: 144392922-144392922 on Build GRCh38
rs912961538 Chr.8: 144392961-144392961 on Build GRCh38

View Full Product Details