Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144393805-144394296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF1
Entrez Gene ID
29894
Gene Name
cleavage and polyadenylation specific factor 1
Gene Aliases
CPSF160, HSU37012, P/cl.18
UniGene
Hs.493202
Gene Chromosome Location
Chr.8: 144393231-144409400 on Build GRCh38
Transcripts
NM_013291.2 XM_006716548.2 XM_006716549.2 XM_006716550.3 XM_011516997.1 XM_011516998.1 XM_011516999.1 NR_030635.1

Gene Symbol
MIR939
Entrez Gene ID
100126351
Gene Name
microRNA 939
Gene Aliases
MIRN939, hsa-mir-939, mir-939
UniGene
Gene Chromosome Location
Chr.8: 144394149-144394230 on Build GRCh38
Transcripts
NM_013291.2 XM_006716548.2 XM_006716549.2 XM_006716550.3 XM_011516997.1 XM_011516998.1 XM_011516999.1 NR_030635.1

SNP Detail

SNP ID Chromosome Location
rs782713363 Chr.8: 144394133-144394133 on Build GRCh38
rs375489571 Chr.8: 144393943-144393943 on Build GRCh38
rs377739366 Chr.8: 144394102-144394102 on Build GRCh38
rs557683150 Chr.8: 144393982-144393982 on Build GRCh38
rs376683753 Chr.8: 144394207-144394207 on Build GRCh38
rs782520798 Chr.8: 144393871-144393871 on Build GRCh38
rs781966721 Chr.8: 144394127-144394127 on Build GRCh38
rs375621365 Chr.8: 144393877-144393877 on Build GRCh38
rs781946129 Chr.8: 144394196-144394196 on Build GRCh38
rs371862605 Chr.8: 144393937-144393937 on Build GRCh38
rs782233610 Chr.8: 144394189-144394189 on Build GRCh38
rs368508789 Chr.8: 144393917-144393917 on Build GRCh38
rs147891480 Chr.8: 144393896-144393896 on Build GRCh38
rs782602455 Chr.8: 144394268-144394268 on Build GRCh38
rs373457591 Chr.8: 144394093-144394093 on Build GRCh38
rs781809794 Chr.8: 144393958-144393959 on Build GRCh38
rs371302764 Chr.8: 144394210-144394210 on Build GRCh38
rs782559336 Chr.8: 144393942-144393943 on Build GRCh38
rs782112378 Chr.8: 144394224-144394224 on Build GRCh38
rs368926825 Chr.8: 144394047-144394047 on Build GRCh38
rs35486628 Chr.8: 144394162-144394163 on Build GRCh38
rs782490414 Chr.8: 144394264-144394264 on Build GRCh38
rs782525771 Chr.8: 144394254-144394254 on Build GRCh38
rs782801667 Chr.8: 144393948-144393949 on Build GRCh38
rs782315301 Chr.8: 144394005-144394005 on Build GRCh38
rs781859831 Chr.8: 144393920-144393920 on Build GRCh38
rs782251256 Chr.8: 144394012-144394012 on Build GRCh38
rs782136527 Chr.8: 144394118-144394118 on Build GRCh38
rs782089036 Chr.8: 144394128-144394128 on Build GRCh38
rs112257111 Chr.8: 144394190-144394190 on Build GRCh38
rs147063268 Chr.8: 144393953-144393953 on Build GRCh38
rs185630472 Chr.8: 144393916-144393916 on Build GRCh38
rs782453554 Chr.8: 144393978-144393978 on Build GRCh38
rs782669321 Chr.8: 144394011-144394011 on Build GRCh38
rs781996889 Chr.8: 144393968-144393968 on Build GRCh38
rs781941516 Chr.8: 144394242-144394242 on Build GRCh38
rs781901513 Chr.8: 144394253-144394253 on Build GRCh38
rs782110147 Chr.8: 144393969-144393969 on Build GRCh38
rs782627367 Chr.8: 144394140-144394140 on Build GRCh38
rs376066671 Chr.8: 144394275-144394275 on Build GRCh38
rs149434817 Chr.8: 144394150-144394150 on Build GRCh38
rs782420033 Chr.8: 144394112-144394112 on Build GRCh38
rs573258685 Chr.8: 144394080-144394080 on Build GRCh38
rs781806525 Chr.8: 144394151-144394151 on Build GRCh38
rs781796293 Chr.8: 144394136-144394136 on Build GRCh38
rs782285221 Chr.8: 144394166-144394166 on Build GRCh38
rs782776996 Chr.8: 144393929-144393929 on Build GRCh38
rs367644143 Chr.8: 144393890-144393890 on Build GRCh38
rs199796519 Chr.8: 144393949-144393949 on Build GRCh38
rs782544283 Chr.8: 144393926-144393926 on Build GRCh38
rs782436921 Chr.8: 144393942-144393942 on Build GRCh38
rs781856385 Chr.8: 144393995-144393995 on Build GRCh38
rs781960095 Chr.8: 144394024-144394024 on Build GRCh38
rs139055700 Chr.8: 144393914-144393914 on Build GRCh38
rs781914193 Chr.8: 144393846-144393847 on Build GRCh38
rs782680232 Chr.8: 144393864-144393864 on Build GRCh38
rs781857684 Chr.8: 144394263-144394263 on Build GRCh38
rs372193470 Chr.8: 144393891-144393891 on Build GRCh38
rs782203918 Chr.8: 144394002-144394002 on Build GRCh38
rs782495685 Chr.8: 144393998-144393998 on Build GRCh38
rs782380278 Chr.8: 144394120-144394120 on Build GRCh38
rs782433236 Chr.8: 144394082-144394082 on Build GRCh38
rs782279289 Chr.8: 144393867-144393867 on Build GRCh38
rs143985093 Chr.8: 144393989-144393989 on Build GRCh38
rs201105226 Chr.8: 144393980-144393980 on Build GRCh38
rs117646413 Chr.8: 144393918-144393918 on Build GRCh38
rs782636257 Chr.8: 144394187-144394187 on Build GRCh38
rs781886905 Chr.8: 144394092-144394092 on Build GRCh38
rs782111663 Chr.8: 144394073-144394073 on Build GRCh38
rs782217981 Chr.8: 144393885-144393885 on Build GRCh38
rs561854368 Chr.8: 144393834-144393834 on Build GRCh38
rs376361407 Chr.8: 144394094-144394094 on Build GRCh38
rs150679420 Chr.8: 144394000-144394000 on Build GRCh38
rs532776402 Chr.8: 144394227-144394227 on Build GRCh38
rs782688228 Chr.8: 144394251-144394251 on Build GRCh38
rs782171639 Chr.8: 144394140-144394140 on Build GRCh38
rs782211712 Chr.8: 144394179-144394179 on Build GRCh38
rs4436163 Chr.8: 144393965-144393965 on Build GRCh38
rs782482032 Chr.8: 144393934-144393934 on Build GRCh38
rs199548712 Chr.8: 144394078-144394078 on Build GRCh38
rs782327238 Chr.8: 144394033-144394033 on Build GRCh38
rs539044126 Chr.8: 144393913-144393913 on Build GRCh38
rs782170591 Chr.8: 144394051-144394051 on Build GRCh38
rs782187928 Chr.8: 144393940-144393940 on Build GRCh38
rs781853967 Chr.8: 144394145-144394145 on Build GRCh38
rs782782901 Chr.8: 144394260-144394260 on Build GRCh38
rs138444951 Chr.8: 144393977-144393977 on Build GRCh38
rs199576680 Chr.8: 144394029-144394029 on Build GRCh38
rs201604645 Chr.8: 144393848-144393848 on Build GRCh38
rs782065600 Chr.8: 144394243-144394243 on Build GRCh38
rs781826660 Chr.8: 144394079-144394079 on Build GRCh38
rs782593372 Chr.8: 144394101-144394101 on Build GRCh38
rs373063011 Chr.8: 144394100-144394100 on Build GRCh38
rs782513443 Chr.8: 144393833-144393833 on Build GRCh38
rs781873059 Chr.8: 144394072-144394072 on Build GRCh38
rs782219092 Chr.8: 144394096-144394096 on Build GRCh38
rs530381565 Chr.8: 144394198-144394198 on Build GRCh38
rs782428774 Chr.8: 144393860-144393860 on Build GRCh38
rs782425527 Chr.8: 144393903-144393903 on Build GRCh38
rs201455910 Chr.8: 144394099-144394099 on Build GRCh38
rs532165980 Chr.8: 144394274-144394274 on Build GRCh38
rs373395558 Chr.8: 144393976-144393976 on Build GRCh38
rs777198861 Chr.8: 144394146-144394146 on Build GRCh38
rs562433649 Chr.8: 144394171-144394171 on Build GRCh38
rs200181330 Chr.8: 144393985-144393986 on Build GRCh38
rs781855390 Chr.8: 144393933-144393933 on Build GRCh38
rs782448023 Chr.8: 144394152-144394152 on Build GRCh38
rs782372165 Chr.8: 144394015-144394015 on Build GRCh38
rs374825674 Chr.8: 144394197-144394197 on Build GRCh38
rs139967807 Chr.8: 144394117-144394117 on Build GRCh38
rs782788902 Chr.8: 144394057-144394057 on Build GRCh38
rs782232986 Chr.8: 144393948-144393948 on Build GRCh38
rs782383028 Chr.8: 144393954-144393954 on Build GRCh38
rs375904098 Chr.8: 144394025-144394025 on Build GRCh38
rs373936657 Chr.8: 144393846-144393846 on Build GRCh38
rs781982896 Chr.8: 144393947-144393947 on Build GRCh38
rs782573195 Chr.8: 144394089-144394089 on Build GRCh38
rs782397246 Chr.8: 144393944-144393944 on Build GRCh38
rs782548508 Chr.8: 144393988-144393988 on Build GRCh38

View Full Product Details