Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144450526-144451020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYHR1
Entrez Gene ID
50626
Gene Name
cysteine and histidine rich 1
Gene Aliases
CHRP
UniGene
Hs.459379
Gene Chromosome Location
Chr.8: 144449582-144465692 on Build GRCh38
Transcripts
NM_138496.1 XM_006716562.3 XM_011517051.1 XM_017013463.1 XM_017013464.1 XM_017013465.1 XM_017013466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5557137 c.939C>T p.I313I Chr.8: 144450529-144450529 on Build GRCh38
4510917 c.851C>T p.S284F Chr.8: 144450617-144450617 on Build GRCh38
5743311 c.841C>T p.R281C Chr.8: 144450627-144450627 on Build GRCh38
5988139 c.710G>A p.R237Q Chr.8: 144450758-144450758 on Build GRCh38
4512864 c.919C>T p.P307S Chr.8: 144450549-144450549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868267233 Chr.8: 144450824-144450824 on Build GRCh38
rs774652958 Chr.8: 144450777-144450780 on Build GRCh38
rs941548865 Chr.8: 144450685-144450685 on Build GRCh38
rs376408801 Chr.8: 144450623-144450623 on Build GRCh38
rs796354767 Chr.8: 144450585-144450585 on Build GRCh38
rs964506337 Chr.8: 144450869-144450869 on Build GRCh38
rs1032720788 Chr.8: 144450655-144450655 on Build GRCh38
rs577942508 Chr.8: 144450895-144450895 on Build GRCh38
rs771879806 Chr.8: 144450720-144450720 on Build GRCh38
rs974279204 Chr.8: 144450923-144450923 on Build GRCh38
rs1042741186 Chr.8: 144450977-144450977 on Build GRCh38
rs949902524 Chr.8: 144450864-144450864 on Build GRCh38
rs770295065 Chr.8: 144450742-144450742 on Build GRCh38
rs556846797 Chr.8: 144450888-144450888 on Build GRCh38
rs938158582 Chr.8: 144450939-144450939 on Build GRCh38
rs549353291 Chr.8: 144450583-144450583 on Build GRCh38
rs1009927431 Chr.8: 144450950-144450950 on Build GRCh38
rs770912811 Chr.8: 144450784-144450784 on Build GRCh38
rs538277145 Chr.8: 144450880-144450880 on Build GRCh38
rs747702632 Chr.8: 144450712-144450712 on Build GRCh38
rs1044655877 Chr.8: 144450865-144450865 on Build GRCh38
rs966220052 Chr.8: 144450993-144450993 on Build GRCh38
rs1030185058 Chr.8: 144450887-144450887 on Build GRCh38
rs922974303 Chr.8: 144450644-144450644 on Build GRCh38
rs566965865 Chr.8: 144450709-144450709 on Build GRCh38
rs1009999504 Chr.8: 144450664-144450664 on Build GRCh38
rs746781715 Chr.8: 144450768-144450768 on Build GRCh38
rs908940347 Chr.8: 144450678-144450678 on Build GRCh38
rs905447693 Chr.8: 144450757-144450757 on Build GRCh38
rs1054090977 Chr.8: 144450892-144450892 on Build GRCh38
rs549701114 Chr.8: 144450807-144450807 on Build GRCh38
rs1018691414 Chr.8: 144450963-144450963 on Build GRCh38
rs143645257 Chr.8: 144450800-144450800 on Build GRCh38
rs910487553 Chr.8: 144450589-144450589 on Build GRCh38
rs983492730 Chr.8: 144450893-144450893 on Build GRCh38
rs776430893 Chr.8: 144450786-144450786 on Build GRCh38
rs895664866 Chr.8: 144450580-144450580 on Build GRCh38
rs908981873 Chr.8: 144450894-144450894 on Build GRCh38
rs148080310 Chr.8: 144450942-144450942 on Build GRCh38
rs759526224 Chr.8: 144450791-144450791 on Build GRCh38
rs1056756042 Chr.8: 144450945-144450945 on Build GRCh38
rs762110820 Chr.8: 144450553-144450553 on Build GRCh38
rs547439110 Chr.8: 144450622-144450622 on Build GRCh38
rs372126009 Chr.8: 144450660-144450660 on Build GRCh38
rs778498105 Chr.8: 144450686-144450686 on Build GRCh38
rs917345596 Chr.8: 144450851-144450851 on Build GRCh38
rs945410969 Chr.8: 144450970-144450970 on Build GRCh38
rs905779871 Chr.8: 144450866-144450866 on Build GRCh38
rs540144156 Chr.8: 144450754-144450754 on Build GRCh38
rs905804939 Chr.8: 144450596-144450596 on Build GRCh38
rs367873167 Chr.8: 144450586-144450586 on Build GRCh38
rs909894076 Chr.8: 144450832-144450832 on Build GRCh38
rs532085784 Chr.8: 144450587-144450587 on Build GRCh38
rs766834948 Chr.8: 144450924-144450924 on Build GRCh38
rs774513270 Chr.8: 144450819-144450819 on Build GRCh38
rs748653253 Chr.8: 144450556-144450556 on Build GRCh38
rs554199126 Chr.8: 144450987-144450987 on Build GRCh38
rs1017357362 Chr.8: 144450867-144450867 on Build GRCh38
rs556754246 Chr.8: 144450669-144450669 on Build GRCh38
rs34576495 Chr.8: 144450604-144450604 on Build GRCh38
rs1047496400 Chr.8: 144450661-144450661 on Build GRCh38
rs75215796 Chr.8: 144450844-144450844 on Build GRCh38
rs896728750 Chr.8: 144450967-144450967 on Build GRCh38
rs572152374 Chr.8: 144450994-144450994 on Build GRCh38
rs975727952 Chr.8: 144450626-144450626 on Build GRCh38
rs1002425442 Chr.8: 144450792-144450792 on Build GRCh38
rs773453591 Chr.8: 144450850-144450850 on Build GRCh38
rs921516679 Chr.8: 144450802-144450802 on Build GRCh38

View Full Product Details