Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144841985-144842487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD5
Entrez Gene ID
28991
Gene Name
COMM domain containing 5
Gene Aliases
HCARG, HT002
UniGene
Hs.493218
Gene Chromosome Location
Chr.8: 144837978-144853578 on Build GRCh38
Transcripts
NM_001282795.1 NM_001282796.1 NM_001282797.1 NM_003416.3 XM_006716654.2 XM_006716656.3 XM_011517291.2 XM_011517292.2 XM_011517293.2 XM_011517294.2 XM_011517295.2 XM_011517296.2 XM_011517297.2 XM_017013813.1 XM_017013814.1 XM_017013815.1 XM_017013816.1 XM_017013817.1

Gene Symbol
ZNF7
Entrez Gene ID
7553
Gene Name
zinc finger protein 7
Gene Aliases
HF.16, KOX4, zf30
UniGene
Hs.493218
Gene Chromosome Location
Chr.8: 144827500-144848070 on Build GRCh38
Transcripts
NM_001282795.1 NM_001282796.1 NM_001282797.1 NM_003416.3 XM_006716654.2 XM_006716656.3 XM_011517291.2 XM_011517292.2 XM_011517293.2 XM_011517294.2 XM_011517295.2 XM_011517296.2 XM_011517297.2 XM_017013813.1 XM_017013814.1 XM_017013815.1 XM_017013816.1 XM_017013817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1097327 c.1081C>A p.P361T Chr.8: 144842188-144842188 on Build GRCh38
6832528 c.1221C>T p.H407H Chr.8: 144842328-144842328 on Build GRCh38
258510 c.977G>T p.R326I Chr.8: 144842084-144842084 on Build GRCh38
3834321 c.909G>A p.E303E Chr.8: 144842016-144842016 on Build GRCh38
5559221 c.879T>C p.L293L Chr.8: 144841986-144841986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912958894 Chr.8: 144842389-144842389 on Build GRCh38
rs2954677 Chr.8: 144842217-144842217 on Build GRCh38
rs902225883 Chr.8: 144842037-144842037 on Build GRCh38
rs376229584 Chr.8: 144842101-144842101 on Build GRCh38
rs769607840 Chr.8: 144842148-144842148 on Build GRCh38
rs778958401 Chr.8: 144842334-144842334 on Build GRCh38
rs777841858 Chr.8: 144842453-144842453 on Build GRCh38
rs945780826 Chr.8: 144842399-144842399 on Build GRCh38
rs891469798 Chr.8: 144842089-144842089 on Build GRCh38
rs904179337 Chr.8: 144842425-144842425 on Build GRCh38
rs767028266 Chr.8: 144842387-144842387 on Build GRCh38
rs146760497 Chr.8: 144842328-144842328 on Build GRCh38
rs769550912 Chr.8: 144842087-144842087 on Build GRCh38
rs769827022 Chr.8: 144842026-144842035 on Build GRCh38
rs756778603 Chr.8: 144842295-144842295 on Build GRCh38
rs765527846 Chr.8: 144842228-144842228 on Build GRCh38
rs950710083 Chr.8: 144842224-144842224 on Build GRCh38
rs927110422 Chr.8: 144842025-144842025 on Build GRCh38
rs754083127 Chr.8: 144842107-144842107 on Build GRCh38
rs779557831 Chr.8: 144842324-144842324 on Build GRCh38
rs746650017 Chr.8: 144842024-144842024 on Build GRCh38
rs767497965 Chr.8: 144842094-144842094 on Build GRCh38
rs755443407 Chr.8: 144842108-144842108 on Build GRCh38
rs773442279 Chr.8: 144842462-144842464 on Build GRCh38
rs749991522 Chr.8: 144842393-144842393 on Build GRCh38
rs557408119 Chr.8: 144842367-144842367 on Build GRCh38
rs140218924 Chr.8: 144842412-144842412 on Build GRCh38
rs999283743 Chr.8: 144842050-144842050 on Build GRCh38
rs370671853 Chr.8: 144842105-144842105 on Build GRCh38
rs772081975 Chr.8: 144842452-144842452 on Build GRCh38
rs575420887 Chr.8: 144842047-144842047 on Build GRCh38
rs963357397 Chr.8: 144842304-144842304 on Build GRCh38
rs770515213 Chr.8: 144842040-144842040 on Build GRCh38
rs773588606 Chr.8: 144842039-144842039 on Build GRCh38
rs776788773 Chr.8: 144842349-144842349 on Build GRCh38
rs1001512186 Chr.8: 144842106-144842106 on Build GRCh38
rs760879577 Chr.8: 144842196-144842196 on Build GRCh38
rs778811579 Chr.8: 144842449-144842449 on Build GRCh38
rs748264027 Chr.8: 144842298-144842298 on Build GRCh38
rs372564530 Chr.8: 144842381-144842381 on Build GRCh38
rs267601831 Chr.8: 144842144-144842144 on Build GRCh38
rs753411479 Chr.8: 144842424-144842424 on Build GRCh38
rs752036728 Chr.8: 144842277-144842277 on Build GRCh38
rs143516383 Chr.8: 144842097-144842097 on Build GRCh38
rs754823897 Chr.8: 144842442-144842442 on Build GRCh38
rs139568707 Chr.8: 144842353-144842353 on Build GRCh38
rs777016015 Chr.8: 144842211-144842211 on Build GRCh38
rs142872514 Chr.8: 144842356-144842356 on Build GRCh38
rs764312595 Chr.8: 144842380-144842380 on Build GRCh38
rs775541945 Chr.8: 144842359-144842359 on Build GRCh38
rs748050885 Chr.8: 144842340-144842340 on Build GRCh38
rs146102880 Chr.8: 144842382-144842382 on Build GRCh38
rs983429838 Chr.8: 144842231-144842231 on Build GRCh38
rs756726658 Chr.8: 144842121-144842121 on Build GRCh38
rs150826392 Chr.8: 144842273-144842273 on Build GRCh38
rs374571392 Chr.8: 144842118-144842118 on Build GRCh38
rs771170422 Chr.8: 144842346-144842346 on Build GRCh38
rs997605344 Chr.8: 144842167-144842167 on Build GRCh38
rs777147974 Chr.8: 144842375-144842375 on Build GRCh38
rs144589431 Chr.8: 144842091-144842091 on Build GRCh38
rs774152794 Chr.8: 144842194-144842194 on Build GRCh38
rs748104153 Chr.8: 144842450-144842450 on Build GRCh38
rs768695739 Chr.8: 144842178-144842178 on Build GRCh38
rs780636592 Chr.8: 144842305-144842305 on Build GRCh38
rs371582136 Chr.8: 144842116-144842116 on Build GRCh38
rs763216060 Chr.8: 144842259-144842259 on Build GRCh38
rs775288270 Chr.8: 144842161-144842161 on Build GRCh38
rs759794931 Chr.8: 144842212-144842212 on Build GRCh38
rs2228180 Chr.8: 144842147-144842147 on Build GRCh38
rs370205689 Chr.8: 144842401-144842401 on Build GRCh38
rs1011195285 Chr.8: 144842013-144842013 on Build GRCh38
rs749194864 Chr.8: 144842177-144842177 on Build GRCh38
rs753050056 Chr.8: 144842244-144842244 on Build GRCh38
rs568443862 Chr.8: 144842321-144842321 on Build GRCh38
rs142823355 Chr.8: 144842067-144842067 on Build GRCh38
rs376385370 Chr.8: 144842317-144842317 on Build GRCh38
rs575469319 Chr.8: 144842386-144842386 on Build GRCh38
rs909190101 Chr.8: 144842293-144842293 on Build GRCh38
rs766545378 Chr.8: 144842206-144842206 on Build GRCh38
rs780869114 Chr.8: 144842133-144842133 on Build GRCh38
rs776259525 Chr.8: 144842056-144842056 on Build GRCh38
rs140119157 Chr.8: 144842417-144842417 on Build GRCh38
rs141409063 Chr.8: 144842117-144842117 on Build GRCh38
rs374019253 Chr.8: 144842302-144842302 on Build GRCh38
rs200004251 Chr.8: 144842070-144842070 on Build GRCh38
rs1054963579 Chr.8: 144842421-144842421 on Build GRCh38
rs769081391 Chr.8: 144842010-144842010 on Build GRCh38
rs779797260 Chr.8: 144842179-144842179 on Build GRCh38
rs768307745 Chr.8: 144842329-144842329 on Build GRCh38
rs938472139 Chr.8: 144842046-144842046 on Build GRCh38
rs772095101 Chr.8: 144842348-144842356 on Build GRCh38
rs762906749 Chr.8: 144842093-144842093 on Build GRCh38
rs376519102 Chr.8: 144842122-144842122 on Build GRCh38

View Full Product Details