Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 3826645-3827182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS3
Entrez Gene ID
169792
Gene Name
GLIS family zinc finger 3
Gene Aliases
NDH, ZNF515
UniGene
Hs.162125
Gene Chromosome Location
Chr.9: 3824127-4310694 on Build GRCh38
Transcripts
NM_001042413.1 NM_152629.3 XM_005251386.4 XM_005251387.4 XM_005251388.4 XM_011517763.2 XM_011517764.2 XM_011517766.2 XM_011517767.2 XM_017014361.1

SNP Detail

SNP ID Chromosome Location
rs948998761 Chr.9: 3826700-3826700 on Build GRCh38
rs997727009 Chr.9: 3826697-3826697 on Build GRCh38
rs998396308 Chr.9: 3826786-3826786 on Build GRCh38
rs1015939090 Chr.9: 3826979-3826979 on Build GRCh38
rs910157090 Chr.9: 3827001-3827001 on Build GRCh38
rs923839341 Chr.9: 3826823-3826823 on Build GRCh38
rs527698212 Chr.9: 3826839-3826839 on Build GRCh38
rs889614072 Chr.9: 3827085-3827085 on Build GRCh38
rs561183159 Chr.9: 3826710-3826711 on Build GRCh38
rs1057334347 Chr.9: 3826682-3826682 on Build GRCh38
rs552052956 Chr.9: 3826715-3826715 on Build GRCh38
rs371779404 Chr.9: 3826673-3826673 on Build GRCh38
rs947503809 Chr.9: 3826887-3826887 on Build GRCh38
rs774303824 Chr.9: 3826693-3826693 on Build GRCh38
rs916418957 Chr.9: 3826785-3826785 on Build GRCh38
rs927689315 Chr.9: 3826946-3826946 on Build GRCh38
rs1012053719 Chr.9: 3826677-3826677 on Build GRCh38
rs923747964 Chr.9: 3827083-3827083 on Build GRCh38
rs950421879 Chr.9: 3826707-3826707 on Build GRCh38
rs956665249 Chr.9: 3827090-3827090 on Build GRCh38
rs993366400 Chr.9: 3826981-3826981 on Build GRCh38
rs377113657 Chr.9: 3826896-3826896 on Build GRCh38
rs960710438 Chr.9: 3827143-3827143 on Build GRCh38
rs1331260 Chr.9: 3827120-3827120 on Build GRCh38
rs188662841 Chr.9: 3826706-3826706 on Build GRCh38
rs902464941 Chr.9: 3827104-3827104 on Build GRCh38
rs1004539311 Chr.9: 3826709-3826709 on Build GRCh38
rs569524976 Chr.9: 3827063-3827063 on Build GRCh38
rs886710399 Chr.9: 3826948-3826948 on Build GRCh38
rs997632125 Chr.9: 3826923-3826923 on Build GRCh38
rs41278331 Chr.9: 3826814-3826814 on Build GRCh38
rs900719781 Chr.9: 3826905-3826905 on Build GRCh38
rs1037003491 Chr.9: 3827053-3827053 on Build GRCh38
rs982057070 Chr.9: 3826821-3826821 on Build GRCh38
rs34919057 Chr.9: 3827087-3827088 on Build GRCh38
rs1039217926 Chr.9: 3826687-3826687 on Build GRCh38
rs12376878 Chr.9: 3826710-3826710 on Build GRCh38
rs114506692 Chr.9: 3827040-3827040 on Build GRCh38
rs80030970 Chr.9: 3826670-3826670 on Build GRCh38
rs760508729 Chr.9: 3826862-3826862 on Build GRCh38
rs150411858 Chr.9: 3827156-3827156 on Build GRCh38
rs775321147 Chr.9: 3826856-3826856 on Build GRCh38
rs947373688 Chr.9: 3827111-3827111 on Build GRCh38
rs10973690 Chr.9: 3827075-3827075 on Build GRCh38
rs922007897 Chr.9: 3827152-3827152 on Build GRCh38
rs115457453 Chr.9: 3826986-3826986 on Build GRCh38
rs989771561 Chr.9: 3827102-3827102 on Build GRCh38
rs547511467 Chr.9: 3826876-3826876 on Build GRCh38
rs1051995930 Chr.9: 3826944-3826944 on Build GRCh38
rs1037077105 Chr.9: 3826766-3826766 on Build GRCh38
rs1005516650 Chr.9: 3826974-3826974 on Build GRCh38
rs1039491146 Chr.9: 3826897-3826897 on Build GRCh38
rs970575821 Chr.9: 3827064-3827064 on Build GRCh38
rs536397203 Chr.9: 3826987-3826987 on Build GRCh38
rs371759528 Chr.9: 3827106-3827106 on Build GRCh38
rs1022217562 Chr.9: 3827100-3827100 on Build GRCh38
rs938845496 Chr.9: 3826679-3826679 on Build GRCh38
rs762860564 Chr.9: 3827139-3827140 on Build GRCh38
rs900646283 Chr.9: 3826694-3826694 on Build GRCh38
rs901292983 Chr.9: 3826777-3826777 on Build GRCh38
rs763895455 Chr.9: 3826867-3826867 on Build GRCh38
rs954075641 Chr.9: 3826847-3826847 on Build GRCh38
rs934669458 Chr.9: 3826829-3826829 on Build GRCh38
rs1048612959 Chr.9: 3826989-3826989 on Build GRCh38
rs966349280 Chr.9: 3826912-3826912 on Build GRCh38
rs990940683 Chr.9: 3826754-3826754 on Build GRCh38

View Full Product Details