Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5450418-5450926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD274
Entrez Gene ID
29126
Gene Name
CD274 molecule
Gene Aliases
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1
UniGene
Hs.521989
Gene Chromosome Location
Chr.9: 5450503-5470567 on Build GRCh38
Transcripts
NM_001267706.1 NM_001314029.1 NM_014143.3 NR_052005.1

SNP Detail

SNP ID Chromosome Location
rs955981483 Chr.9: 5450487-5450487 on Build GRCh38
rs140476284 Chr.9: 5450635-5450635 on Build GRCh38
rs989420962 Chr.9: 5450845-5450845 on Build GRCh38
rs368909847 Chr.9: 5450899-5450899 on Build GRCh38
rs905982602 Chr.9: 5450567-5450567 on Build GRCh38
rs563552297 Chr.9: 5450491-5450491 on Build GRCh38
rs1038695633 Chr.9: 5450712-5450712 on Build GRCh38
rs955903612 Chr.9: 5450590-5450590 on Build GRCh38
rs751188837 Chr.9: 5450663-5450663 on Build GRCh38
rs763863722 Chr.9: 5450550-5450550 on Build GRCh38
rs927341864 Chr.9: 5450736-5450736 on Build GRCh38
rs1011367357 Chr.9: 5450751-5450751 on Build GRCh38
rs1045849457 Chr.9: 5450562-5450562 on Build GRCh38
rs374792232 Chr.9: 5450620-5450620 on Build GRCh38
rs946265558 Chr.9: 5450741-5450756 on Build GRCh38
rs1034007712 Chr.9: 5450441-5450441 on Build GRCh38
rs985680077 Chr.9: 5450499-5450499 on Build GRCh38
rs1005437658 Chr.9: 5450604-5450604 on Build GRCh38
rs557147437 Chr.9: 5450835-5450835 on Build GRCh38
rs1053794156 Chr.9: 5450531-5450531 on Build GRCh38
rs990050475 Chr.9: 5450490-5450490 on Build GRCh38
rs898025726 Chr.9: 5450533-5450534 on Build GRCh38
rs144429445 Chr.9: 5450744-5450744 on Build GRCh38
rs749389442 Chr.9: 5450767-5450767 on Build GRCh38
rs889463930 Chr.9: 5450802-5450802 on Build GRCh38
rs536109304 Chr.9: 5450446-5450446 on Build GRCh38
rs534744904 Chr.9: 5450772-5450772 on Build GRCh38
rs115818896 Chr.9: 5450803-5450803 on Build GRCh38
rs1005610090 Chr.9: 5450455-5450455 on Build GRCh38
rs888294079 Chr.9: 5450602-5450602 on Build GRCh38
rs1017740835 Chr.9: 5450547-5450547 on Build GRCh38
rs1007237402 Chr.9: 5450546-5450546 on Build GRCh38
rs992224687 Chr.9: 5450889-5450889 on Build GRCh38
rs961039923 Chr.9: 5450476-5450476 on Build GRCh38
rs1054989395 Chr.9: 5450595-5450595 on Build GRCh38
rs980798176 Chr.9: 5450511-5450511 on Build GRCh38
rs7852887 Chr.9: 5450750-5450750 on Build GRCh38
rs989041207 Chr.9: 5450682-5450682 on Build GRCh38
rs1033665703 Chr.9: 5450829-5450829 on Build GRCh38
rs543328343 Chr.9: 5450508-5450508 on Build GRCh38
rs559530117 Chr.9: 5450584-5450584 on Build GRCh38
rs754149006 Chr.9: 5450494-5450495 on Build GRCh38
rs929525191 Chr.9: 5450539-5450539 on Build GRCh38
rs1002434995 Chr.9: 5450576-5450576 on Build GRCh38
rs950036371 Chr.9: 5450561-5450561 on Build GRCh38
rs575810788 Chr.9: 5450886-5450886 on Build GRCh38
rs943638660 Chr.9: 5450708-5450708 on Build GRCh38
rs779114769 Chr.9: 5450777-5450777 on Build GRCh38
rs957560683 Chr.9: 5450815-5450815 on Build GRCh38
rs981471216 Chr.9: 5450731-5450731 on Build GRCh38
rs7866740 Chr.9: 5450529-5450529 on Build GRCh38
rs757423880 Chr.9: 5450684-5450684 on Build GRCh38
rs755970653 Chr.9: 5450693-5450693 on Build GRCh38
rs186936117 Chr.9: 5450691-5450691 on Build GRCh38
rs10815225 Chr.9: 5450497-5450497 on Build GRCh38
rs773427222 Chr.9: 5450872-5450872 on Build GRCh38
rs1049623005 Chr.9: 5450742-5450742 on Build GRCh38
rs1015931983 Chr.9: 5450614-5450614 on Build GRCh38
rs879779721 Chr.9: 5450830-5450830 on Build GRCh38
rs577710774 Chr.9: 5450603-5450603 on Build GRCh38
rs561390500 Chr.9: 5450524-5450524 on Build GRCh38
rs1020537200 Chr.9: 5450856-5450856 on Build GRCh38
rs62559142 Chr.9: 5450541-5450541 on Build GRCh38
rs751419649 Chr.9: 5450666-5450666 on Build GRCh38
rs975950688 Chr.9: 5450459-5450459 on Build GRCh38
rs971664801 Chr.9: 5450862-5450862 on Build GRCh38
rs957780220 Chr.9: 5450669-5450669 on Build GRCh38
rs926649962 Chr.9: 5450659-5450659 on Build GRCh38
rs953820558 Chr.9: 5450654-5450654 on Build GRCh38
rs569851352 Chr.9: 5450683-5450683 on Build GRCh38
rs768666866 Chr.9: 5450774-5450774 on Build GRCh38
rs12337507 Chr.9: 5450453-5450453 on Build GRCh38
rs937416272 Chr.9: 5450738-5450738 on Build GRCh38
rs534870056 Chr.9: 5450513-5450513 on Build GRCh38
rs35608565 Chr.9: 5450633-5450634 on Build GRCh38
rs954208053 Chr.9: 5450498-5450498 on Build GRCh38
rs764174484 Chr.9: 5450592-5450592 on Build GRCh38
rs980803504 Chr.9: 5450650-5450650 on Build GRCh38
rs989996544 Chr.9: 5450605-5450605 on Build GRCh38
rs901530606 Chr.9: 5450726-5450726 on Build GRCh38
rs142555137 Chr.9: 5450474-5450474 on Build GRCh38
rs867347252 Chr.9: 5450800-5450800 on Build GRCh38
rs117829760 Chr.9: 5450790-5450790 on Build GRCh38
rs12342381 Chr.9: 5450810-5450810 on Build GRCh38
rs997846281 Chr.9: 5450628-5450628 on Build GRCh38
rs192414020 Chr.9: 5450876-5450876 on Build GRCh38
rs749699815 Chr.9: 5450892-5450892 on Build GRCh38

View Full Product Details