Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6532372-6532870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

SNP Detail

SNP ID Chromosome Location
rs770385648 Chr.9: 6532408-6532408 on Build GRCh38
rs976174465 Chr.9: 6532827-6532827 on Build GRCh38
rs916291048 Chr.9: 6532395-6532395 on Build GRCh38
rs1037232301 Chr.9: 6532466-6532466 on Build GRCh38
rs772987594 Chr.9: 6532473-6532473 on Build GRCh38
rs1037248523 Chr.9: 6532656-6532656 on Build GRCh38
rs577665662 Chr.9: 6532829-6532829 on Build GRCh38
rs901195780 Chr.9: 6532699-6532700 on Build GRCh38
rs7848919 Chr.9: 6532477-6532477 on Build GRCh38
rs73639311 Chr.9: 6532460-6532460 on Build GRCh38
rs544133044 Chr.9: 6532480-6532480 on Build GRCh38
rs1030929444 Chr.9: 6532730-6532730 on Build GRCh38
rs1061407 Chr.9: 6532544-6532544 on Build GRCh38
rs1049111024 Chr.9: 6532828-6532828 on Build GRCh38
rs969296973 Chr.9: 6532559-6532559 on Build GRCh38
rs553188892 Chr.9: 6532816-6532816 on Build GRCh38
rs969613210 Chr.9: 6532783-6532783 on Build GRCh38
rs904055435 Chr.9: 6532510-6532510 on Build GRCh38
rs774139467 Chr.9: 6532729-6532729 on Build GRCh38
rs528027921 Chr.9: 6532442-6532442 on Build GRCh38
rs553855999 Chr.9: 6532450-6532450 on Build GRCh38
rs931978912 Chr.9: 6532681-6532681 on Build GRCh38
rs1031194384 Chr.9: 6532525-6532525 on Build GRCh38
rs947867766 Chr.9: 6532565-6532565 on Build GRCh38
rs944777799 Chr.9: 6532659-6532659 on Build GRCh38
rs982616567 Chr.9: 6532629-6532629 on Build GRCh38
rs749941145 Chr.9: 6532782-6532782 on Build GRCh38
rs111326356 Chr.9: 6532760-6532760 on Build GRCh38
rs757228471 Chr.9: 6532511-6532511 on Build GRCh38
rs958013726 Chr.9: 6532741-6532741 on Build GRCh38
rs555435648 Chr.9: 6532486-6532486 on Build GRCh38
rs1031141758 Chr.9: 6532703-6532703 on Build GRCh38
rs768890473 Chr.9: 6532471-6532471 on Build GRCh38
rs750981994 Chr.9: 6532512-6532512 on Build GRCh38
rs900469251 Chr.9: 6532679-6532679 on Build GRCh38
rs947718083 Chr.9: 6532419-6532419 on Build GRCh38
rs537707294 Chr.9: 6532781-6532781 on Build GRCh38
rs3215922 Chr.9: 6532523-6532523 on Build GRCh38
rs796199691 Chr.9: 6532431-6532431 on Build GRCh38
rs547641214 Chr.9: 6532470-6532470 on Build GRCh38
rs141035393 Chr.9: 6532701-6532701 on Build GRCh38
rs145481724 Chr.9: 6532511-6532512 on Build GRCh38
rs973951078 Chr.9: 6532587-6532587 on Build GRCh38
rs148861074 Chr.9: 6532421-6532421 on Build GRCh38
rs1023313280 Chr.9: 6532752-6532752 on Build GRCh38
rs540075890 Chr.9: 6532469-6532469 on Build GRCh38
rs540036394 Chr.9: 6532704-6532704 on Build GRCh38
rs999698910 Chr.9: 6532521-6532521 on Build GRCh38
rs547163879 Chr.9: 6532691-6532691 on Build GRCh38
rs912371579 Chr.9: 6532808-6532808 on Build GRCh38
rs965360965 Chr.9: 6532517-6532517 on Build GRCh38
rs940369337 Chr.9: 6532798-6532798 on Build GRCh38
rs951314581 Chr.9: 6532790-6532790 on Build GRCh38
rs1043995999 Chr.9: 6532685-6532685 on Build GRCh38
rs886522578 Chr.9: 6532428-6532428 on Build GRCh38
rs563940351 Chr.9: 6532558-6532558 on Build GRCh38
rs191365831 Chr.9: 6532495-6532495 on Build GRCh38
rs931550046 Chr.9: 6532668-6532668 on Build GRCh38
rs940332248 Chr.9: 6532655-6532655 on Build GRCh38
rs192055239 Chr.9: 6532670-6532670 on Build GRCh38
rs900773890 Chr.9: 6532467-6532467 on Build GRCh38
rs570274404 Chr.9: 6532767-6532767 on Build GRCh38
rs573934284 Chr.9: 6532801-6532801 on Build GRCh38
rs187792305 Chr.9: 6532663-6532663 on Build GRCh38
rs541456045 Chr.9: 6532804-6532804 on Build GRCh38
rs531460735 Chr.9: 6532577-6532577 on Build GRCh38
rs35834773 Chr.9: 6532607-6532607 on Build GRCh38
rs3211505 Chr.9: 6532553-6532553 on Build GRCh38
rs897347476 Chr.9: 6532469-6532469 on Build GRCh38
rs541142490 Chr.9: 6532504-6532504 on Build GRCh38
rs559832099 Chr.9: 6532529-6532529 on Build GRCh38
rs919833288 Chr.9: 6532598-6532598 on Build GRCh38
rs927662234 Chr.9: 6532650-6532650 on Build GRCh38
rs889934324 Chr.9: 6532677-6532677 on Build GRCh38
rs545105557 Chr.9: 6532835-6532835 on Build GRCh38
rs971738825 Chr.9: 6532809-6532809 on Build GRCh38
rs183063125 Chr.9: 6532548-6532548 on Build GRCh38
rs184498089 Chr.9: 6532788-6532788 on Build GRCh38
rs907978914 Chr.9: 6532639-6532639 on Build GRCh38

View Full Product Details