Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173799729-173800278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPL
Entrez Gene ID
91687
Gene Name
centromere protein L
Gene Aliases
C1orf155, CENP-L, dJ383J4.3
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173799550-173824639 on Build GRCh38
Transcripts
NM_001127181.2 NM_001171182.1 NM_033319.3

SNP Detail

SNP ID Chromosome Location
rs1045712997 Chr.1: 173800250-173800250 on Build GRCh38
rs887691463 Chr.1: 173799900-173799900 on Build GRCh38
rs910027195 Chr.1: 173800045-173800045 on Build GRCh38
rs542139910 Chr.1: 173799935-173799935 on Build GRCh38
rs770471189 Chr.1: 173800160-173800161 on Build GRCh38
rs988152463 Chr.1: 173799787-173799787 on Build GRCh38
rs747213450 Chr.1: 173800240-173800240 on Build GRCh38
rs868250604 Chr.1: 173799896-173799896 on Build GRCh38
rs551084960 Chr.1: 173800148-173800148 on Build GRCh38
rs561911586 Chr.1: 173800022-173800022 on Build GRCh38
rs775602177 Chr.1: 173800165-173800165 on Build GRCh38
rs897515749 Chr.1: 173800223-173800223 on Build GRCh38
rs941963615 Chr.1: 173800194-173800194 on Build GRCh38
rs386636884 Chr.1: 173800022-173800023 on Build GRCh38
rs773825040 Chr.1: 173800161-173800162 on Build GRCh38
rs1033042119 Chr.1: 173800048-173800048 on Build GRCh38
rs950094170 Chr.1: 173799835-173799835 on Build GRCh38
rs192931239 Chr.1: 173800023-173800023 on Build GRCh38
rs577569364 Chr.1: 173799828-173799828 on Build GRCh38
rs35055956 Chr.1: 173799894-173799895 on Build GRCh38
rs1056145613 Chr.1: 173799887-173799887 on Build GRCh38
rs545596806 Chr.1: 173799895-173799895 on Build GRCh38
rs556682097 Chr.1: 173799844-173799844 on Build GRCh38
rs982183733 Chr.1: 173800120-173800120 on Build GRCh38
rs892859709 Chr.1: 173799910-173799910 on Build GRCh38
rs772188109 Chr.1: 173800099-173800099 on Build GRCh38
rs988760390 Chr.1: 173800071-173800071 on Build GRCh38
rs991531933 Chr.1: 173800121-173800121 on Build GRCh38
rs941454193 Chr.1: 173799759-173799759 on Build GRCh38
rs568861306 Chr.1: 173799965-173799965 on Build GRCh38
rs975751939 Chr.1: 173800016-173800016 on Build GRCh38
rs748234543 Chr.1: 173800097-173800097 on Build GRCh38
rs111576505 Chr.1: 173799921-173799921 on Build GRCh38
rs1045616990 Chr.1: 173799836-173799836 on Build GRCh38
rs997281165 Chr.1: 173800000-173800000 on Build GRCh38
rs1020919854 Chr.1: 173800083-173800083 on Build GRCh38
rs1055836047 Chr.1: 173799774-173799774 on Build GRCh38
rs543150744 Chr.1: 173799843-173799843 on Build GRCh38
rs1036658425 Chr.1: 173800196-173800196 on Build GRCh38
rs9425742 Chr.1: 173799847-173799847 on Build GRCh38
rs189432261 Chr.1: 173799933-173799933 on Build GRCh38
rs777900676 Chr.1: 173800166-173800166 on Build GRCh38
rs965692320 Chr.1: 173799971-173799971 on Build GRCh38
rs982941526 Chr.1: 173800034-173800034 on Build GRCh38
rs928488780 Chr.1: 173799846-173799846 on Build GRCh38
rs958312680 Chr.1: 173800055-173800055 on Build GRCh38
rs937205466 Chr.1: 173800159-173800160 on Build GRCh38

View Full Product Details