Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173811095-173811645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPL
Entrez Gene ID
91687
Gene Name
centromere protein L
Gene Aliases
C1orf155, CENP-L, dJ383J4.3
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173799550-173824639 on Build GRCh38
Transcripts
NM_001127181.2 NM_001171182.1 NM_033319.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4902558 c.149C>T p.P50L Chr.1: 173811151-173811151 on Build GRCh38
4948414 c.89G>A p.R30Q Chr.1: 173811211-173811211 on Build GRCh38
3802902 c.8C>T p.S3F Chr.1: 173811292-173811292 on Build GRCh38
4025220 c.158C>T p.S53L Chr.1: 173811142-173811142 on Build GRCh38
899789 c.111G>T p.Q37H Chr.1: 173811189-173811189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753838147 Chr.1: 173811261-173811261 on Build GRCh38
rs544972061 Chr.1: 173811516-173811516 on Build GRCh38
rs776986729 Chr.1: 173811141-173811141 on Build GRCh38
rs1046329356 Chr.1: 173811564-173811564 on Build GRCh38
rs952181147 Chr.1: 173811236-173811236 on Build GRCh38
rs75337814 Chr.1: 173811353-173811353 on Build GRCh38
rs750186632 Chr.1: 173811164-173811164 on Build GRCh38
rs898034642 Chr.1: 173811479-173811479 on Build GRCh38
rs768818748 Chr.1: 173811265-173811265 on Build GRCh38
rs766067591 Chr.1: 173811256-173811256 on Build GRCh38
rs774150923 Chr.1: 173811523-173811523 on Build GRCh38
rs905129572 Chr.1: 173811614-173811614 on Build GRCh38
rs767450186 Chr.1: 173811163-173811163 on Build GRCh38
rs143227824 Chr.1: 173811288-173811288 on Build GRCh38
rs994018186 Chr.1: 173811212-173811212 on Build GRCh38
rs760560286 Chr.1: 173811252-173811252 on Build GRCh38
rs200103202 Chr.1: 173811283-173811283 on Build GRCh38
rs932318413 Chr.1: 173811351-173811351 on Build GRCh38
rs771407346 Chr.1: 173811138-173811138 on Build GRCh38
rs753688759 Chr.1: 173811176-173811176 on Build GRCh38
rs767846769 Chr.1: 173811390-173811390 on Build GRCh38
rs531870769 Chr.1: 173811476-173811476 on Build GRCh38
rs960594921 Chr.1: 173811537-173811537 on Build GRCh38
rs895811289 Chr.1: 173811522-173811522 on Build GRCh38
rs908204549 Chr.1: 173811421-173811421 on Build GRCh38
rs778918147 Chr.1: 173811186-173811186 on Build GRCh38
rs999235861 Chr.1: 173811226-173811226 on Build GRCh38
rs139831401 Chr.1: 173811224-173811224 on Build GRCh38
rs747479891 Chr.1: 173811135-173811135 on Build GRCh38
rs111496356 Chr.1: 173811349-173811349 on Build GRCh38
rs921545471 Chr.1: 173811452-173811452 on Build GRCh38
rs145812754 Chr.1: 173811279-173811279 on Build GRCh38
rs147657848 Chr.1: 173811200-173811200 on Build GRCh38
rs751697791 Chr.1: 173811325-173811325 on Build GRCh38
rs1050672502 Chr.1: 173811377-173811377 on Build GRCh38
rs1002062309 Chr.1: 173811327-173811327 on Build GRCh38
rs1033570347 Chr.1: 173811531-173811531 on Build GRCh38
rs982821155 Chr.1: 173811408-173811408 on Build GRCh38
rs773732756 Chr.1: 173811438-173811438 on Build GRCh38
rs761507334 Chr.1: 173811159-173811159 on Build GRCh38
rs993595615 Chr.1: 173811494-173811494 on Build GRCh38
rs758553316 Chr.1: 173811187-173811187 on Build GRCh38
rs924935355 Chr.1: 173811335-173811335 on Build GRCh38
rs573193895 Chr.1: 173811496-173811496 on Build GRCh38
rs151044818 Chr.1: 173811237-173811237 on Build GRCh38
rs755847882 Chr.1: 173811165-173811165 on Build GRCh38
rs759355150 Chr.1: 173811275-173811275 on Build GRCh38
rs746337503 Chr.1: 173811213-173811213 on Build GRCh38
rs772451925 Chr.1: 173811133-173811133 on Build GRCh38
rs762122544 Chr.1: 173811333-173811333 on Build GRCh38
rs748728967 Chr.1: 173811409-173811409 on Build GRCh38
rs373147233 Chr.1: 173811166-173811166 on Build GRCh38
rs778033095 Chr.1: 173811289-173811289 on Build GRCh38
rs201830798 Chr.1: 173811211-173811211 on Build GRCh38
rs747460617 Chr.1: 173811201-173811201 on Build GRCh38
rs762589173 Chr.1: 173811142-173811142 on Build GRCh38
rs755137479 Chr.1: 173811183-173811183 on Build GRCh38
rs945093328 Chr.1: 173811328-173811328 on Build GRCh38
rs1040507418 Chr.1: 173811455-173811455 on Build GRCh38
rs562810769 Chr.1: 173811446-173811446 on Build GRCh38
rs773958233 Chr.1: 173811231-173811231 on Build GRCh38
rs965052396 Chr.1: 173811434-173811434 on Build GRCh38
rs139971891 Chr.1: 173811250-173811250 on Build GRCh38
rs1053654080 Chr.1: 173811518-173811518 on Build GRCh38
rs376379173 Chr.1: 173811245-173811245 on Build GRCh38
rs66554404 Chr.1: 173811349-173811349 on Build GRCh38
rs377503187 Chr.1: 173811246-173811246 on Build GRCh38
rs774194739 Chr.1: 173811151-173811151 on Build GRCh38

View Full Product Details