Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175006667-175007179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
UniGene
Hs.508524
Gene Chromosome Location
Chr.1: 174999435-175012027 on Build GRCh38
Transcripts
NM_001007214.1 NM_014412.2 XM_017001046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4025295 c.274A>G p.T92A Chr.1: 175006783-175006783 on Build GRCh38
1336409 c.379A>G p.M127V Chr.1: 175007144-175007144 on Build GRCh38
677040 c.381G>A p.M127I Chr.1: 175007146-175007146 on Build GRCh38
1626637 c.344T>G p.L115R Chr.1: 175007109-175007109 on Build GRCh38
7195725 c.333-1G>A p.? Chr.1: 175007097-175007097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753358841 Chr.1: 175006776-175006776 on Build GRCh38
rs368385861 Chr.1: 175006736-175006736 on Build GRCh38
rs16847689 Chr.1: 175007048-175007048 on Build GRCh38
rs147412966 Chr.1: 175006810-175006810 on Build GRCh38
rs760286586 Chr.1: 175006762-175006762 on Build GRCh38
rs746196232 Chr.1: 175006888-175006888 on Build GRCh38
rs147232439 Chr.1: 175006821-175006821 on Build GRCh38
rs202104286 Chr.1: 175006729-175006729 on Build GRCh38
rs372155098 Chr.1: 175006984-175006984 on Build GRCh38
rs754636756 Chr.1: 175006779-175006779 on Build GRCh38
rs145239568 Chr.1: 175006893-175006893 on Build GRCh38
rs751407379 Chr.1: 175006892-175006905 on Build GRCh38
rs936689171 Chr.1: 175007147-175007147 on Build GRCh38
rs749935580 Chr.1: 175006734-175006736 on Build GRCh38
rs150909890 Chr.1: 175007128-175007128 on Build GRCh38
rs569818004 Chr.1: 175007137-175007137 on Build GRCh38
rs116135783 Chr.1: 175006975-175006975 on Build GRCh38
rs888935260 Chr.1: 175006967-175006967 on Build GRCh38
rs750475722 Chr.1: 175007139-175007139 on Build GRCh38
rs376303272 Chr.1: 175006719-175006719 on Build GRCh38
rs746113040 Chr.1: 175006733-175006733 on Build GRCh38
rs374135102 Chr.1: 175006852-175006854 on Build GRCh38
rs376434858 Chr.1: 175006811-175006811 on Build GRCh38
rs992048172 Chr.1: 175007134-175007134 on Build GRCh38
rs780786686 Chr.1: 175007082-175007084 on Build GRCh38
rs745950373 Chr.1: 175006819-175006819 on Build GRCh38
rs1018831998 Chr.1: 175006974-175006974 on Build GRCh38
rs775719446 Chr.1: 175006825-175006825 on Build GRCh38
rs945607640 Chr.1: 175007052-175007052 on Build GRCh38
rs752874784 Chr.1: 175006951-175006951 on Build GRCh38
rs751330585 Chr.1: 175007070-175007070 on Build GRCh38
rs1042694422 Chr.1: 175007101-175007101 on Build GRCh38
rs369204691 Chr.1: 175007051-175007051 on Build GRCh38
rs994786155 Chr.1: 175006932-175006932 on Build GRCh38
rs967728753 Chr.1: 175007002-175007002 on Build GRCh38
rs757240575 Chr.1: 175007116-175007116 on Build GRCh38
rs548172319 Chr.1: 175007019-175007020 on Build GRCh38
rs201198662 Chr.1: 175006737-175006737 on Build GRCh38
rs1802325 Chr.1: 175006836-175006836 on Build GRCh38
rs761697071 Chr.1: 175007020-175007020 on Build GRCh38
rs769093535 Chr.1: 175006847-175006847 on Build GRCh38
rs780175150 Chr.1: 175006892-175006896 on Build GRCh38
rs754833803 Chr.1: 175006696-175006696 on Build GRCh38
rs777707220 Chr.1: 175006807-175006807 on Build GRCh38
rs200258548 Chr.1: 175007118-175007118 on Build GRCh38
rs369081704 Chr.1: 175006849-175006849 on Build GRCh38
rs747961374 Chr.1: 175006710-175006710 on Build GRCh38
rs758131688 Chr.1: 175006803-175006803 on Build GRCh38
rs374753179 Chr.1: 175006815-175006815 on Build GRCh38
rs1056701075 Chr.1: 175006702-175006702 on Build GRCh38
rs754817774 Chr.1: 175007071-175007073 on Build GRCh38
rs896284522 Chr.1: 175006852-175006852 on Build GRCh38
rs189421840 Chr.1: 175006908-175006908 on Build GRCh38
rs752436527 Chr.1: 175006792-175006792 on Build GRCh38
rs774945166 Chr.1: 175006755-175006755 on Build GRCh38
rs747103218 Chr.1: 175006724-175006724 on Build GRCh38
rs539831859 Chr.1: 175007111-175007111 on Build GRCh38
rs778614243 Chr.1: 175006699-175006699 on Build GRCh38
rs762764567 Chr.1: 175007064-175007064 on Build GRCh38
rs776069773 Chr.1: 175006892-175006892 on Build GRCh38
rs763911991 Chr.1: 175007067-175007067 on Build GRCh38
rs938170632 Chr.1: 175007027-175007027 on Build GRCh38
rs764937689 Chr.1: 175006791-175006791 on Build GRCh38
rs138568219 Chr.1: 175006745-175006745 on Build GRCh38
rs1005087433 Chr.1: 175006969-175006969 on Build GRCh38
rs984009041 Chr.1: 175006721-175006721 on Build GRCh38
rs372208487 Chr.1: 175006885-175006885 on Build GRCh38
rs371245913 Chr.1: 175006718-175006718 on Build GRCh38
rs571802566 Chr.1: 175006962-175006962 on Build GRCh38
rs181474450 Chr.1: 175007013-175007013 on Build GRCh38
rs781361346 Chr.1: 175006756-175006756 on Build GRCh38
rs764904365 Chr.1: 175006710-175006713 on Build GRCh38
rs377076757 Chr.1: 175006854-175006856 on Build GRCh38
rs1007822080 Chr.1: 175006970-175006970 on Build GRCh38
rs1025935645 Chr.1: 175006979-175006979 on Build GRCh38
rs776867638 Chr.1: 175006732-175006732 on Build GRCh38
rs765949671 Chr.1: 175006764-175006764 on Build GRCh38
rs971668246 Chr.1: 175006978-175006978 on Build GRCh38
rs992749826 Chr.1: 175007031-175007031 on Build GRCh38
rs6677924 Chr.1: 175006798-175006798 on Build GRCh38
rs75796638 Chr.1: 175006994-175006994 on Build GRCh38

View Full Product Details