Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175010861-175011366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
UniGene
Hs.508524
Gene Chromosome Location
Chr.1: 174999435-175012027 on Build GRCh38
Transcripts
NM_001007214.1 NM_014412.2

SNP Detail

SNP ID Chromosome Location
rs938385803 Chr.1: 175011291-175011291 on Build GRCh38
rs942878437 Chr.1: 175010947-175010947 on Build GRCh38
rs907971911 Chr.1: 175011067-175011067 on Build GRCh38
rs568752762 Chr.1: 175011070-175011070 on Build GRCh38
rs1035459388 Chr.1: 175010895-175010895 on Build GRCh38
rs558602646 Chr.1: 175011107-175011114 on Build GRCh38
rs918615770 Chr.1: 175010912-175010913 on Build GRCh38
rs758182425 Chr.1: 175011107-175011115 on Build GRCh38
rs754386242 Chr.1: 175010912-175010912 on Build GRCh38
rs1026128827 Chr.1: 175011181-175011182 on Build GRCh38
rs972594013 Chr.1: 175011130-175011130 on Build GRCh38
rs548573392 Chr.1: 175011065-175011065 on Build GRCh38
rs891934407 Chr.1: 175010905-175010905 on Build GRCh38
rs186390272 Chr.1: 175011248-175011248 on Build GRCh38
rs912813573 Chr.1: 175010941-175010942 on Build GRCh38
rs1022262783 Chr.1: 175010949-175010949 on Build GRCh38
rs535813233 Chr.1: 175011120-175011127 on Build GRCh38
rs894109021 Chr.1: 175011299-175011299 on Build GRCh38
rs765592493 Chr.1: 175010963-175010963 on Build GRCh38
rs575290642 Chr.1: 175011086-175011087 on Build GRCh38
rs370010497 Chr.1: 175011081-175011081 on Build GRCh38
rs528090675 Chr.1: 175011041-175011041 on Build GRCh38
rs987498359 Chr.1: 175010991-175010991 on Build GRCh38
rs191295452 Chr.1: 175011270-175011270 on Build GRCh38
rs759854119 Chr.1: 175011106-175011107 on Build GRCh38
rs368806487 Chr.1: 175010933-175010934 on Build GRCh38
rs1043230034 Chr.1: 175011309-175011309 on Build GRCh38
rs1056734461 Chr.1: 175011296-175011298 on Build GRCh38
rs570006878 Chr.1: 175010934-175010934 on Build GRCh38
rs549005025 Chr.1: 175011178-175011179 on Build GRCh38
rs558685649 Chr.1: 175011254-175011254 on Build GRCh38
rs1002476896 Chr.1: 175011066-175011066 on Build GRCh38
rs939821890 Chr.1: 175011095-175011095 on Build GRCh38
rs919792659 Chr.1: 175011131-175011131 on Build GRCh38
rs1041830823 Chr.1: 175011004-175011004 on Build GRCh38
rs554507921 Chr.1: 175011308-175011309 on Build GRCh38
rs747170461 Chr.1: 175011108-175011115 on Build GRCh38
rs987072408 Chr.1: 175010938-175010939 on Build GRCh38
rs1057018250 Chr.1: 175011085-175011086 on Build GRCh38
rs138660037 Chr.1: 175011216-175011217 on Build GRCh38
rs914759464 Chr.1: 175011264-175011264 on Build GRCh38
rs984381 Chr.1: 175010962-175010962 on Build GRCh38
rs188774185 Chr.1: 175011143-175011143 on Build GRCh38
rs772508287 Chr.1: 175011326-175011327 on Build GRCh38
rs1041491713 Chr.1: 175011275-175011275 on Build GRCh38
rs565648903 Chr.1: 175011260-175011260 on Build GRCh38
rs984380 Chr.1: 175011039-175011039 on Build GRCh38
rs931238482 Chr.1: 175011139-175011139 on Build GRCh38
rs902877528 Chr.1: 175011048-175011048 on Build GRCh38
rs12565944 Chr.1: 175010985-175010985 on Build GRCh38
rs963523089 Chr.1: 175011137-175011138 on Build GRCh38
rs368560590 Chr.1: 175011224-175011224 on Build GRCh38
rs964290532 Chr.1: 175011252-175011253 on Build GRCh38
rs954739596 Chr.1: 175010989-175010989 on Build GRCh38
rs181765977 Chr.1: 175011233-175011233 on Build GRCh38
rs1016102912 Chr.1: 175011133-175011134 on Build GRCh38
rs891550386 Chr.1: 175011258-175011258 on Build GRCh38
rs11444655 Chr.1: 175011107-175011108 on Build GRCh38
rs528470129 Chr.1: 175011010-175011010 on Build GRCh38
rs372540376 Chr.1: 175011220-175011221 on Build GRCh38
rs924361108 Chr.1: 175011287-175011287 on Build GRCh38
rs755513403 Chr.1: 175010957-175010957 on Build GRCh38
rs1034462 Chr.1: 175011187-175011187 on Build GRCh38
rs926925734 Chr.1: 175011162-175011162 on Build GRCh38

View Full Product Details