Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179814899-179815408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM163A
Entrez Gene ID
148753
Gene Name
family with sequence similarity 163 member A
Gene Aliases
C1orf76, NDSP
UniGene
Hs.400696
Gene Chromosome Location
Chr.1: 179727763-179816198 on Build GRCh38
Transcripts
NM_173509.2 XM_005244913.2 XM_006711174.2 XM_006711175.2 XM_006711176.2 XM_006711177.1 XM_011509217.1 XM_011509221.1 XM_011509226.1 XM_011509228.1 XM_017000386.1 XM_017000387.1 XM_017000388.1 XM_017000389.1 XM_017000390.1 XM_017000391.1 XM_017000392.1 XM_017000393.1

SNP Detail

SNP ID Chromosome Location
rs890293594 Chr.1: 179815381-179815381 on Build GRCh38
rs747780803 Chr.1: 179814945-179814945 on Build GRCh38
rs397834372 Chr.1: 179815116-179815116 on Build GRCh38
rs774118754 Chr.1: 179815105-179815109 on Build GRCh38
rs938450308 Chr.1: 179814944-179814944 on Build GRCh38
rs1051659175 Chr.1: 179815308-179815308 on Build GRCh38
rs538460339 Chr.1: 179815298-179815298 on Build GRCh38
rs141413813 Chr.1: 179815107-179815107 on Build GRCh38
rs574383042 Chr.1: 179814997-179814997 on Build GRCh38
rs980378693 Chr.1: 179815094-179815095 on Build GRCh38
rs889700240 Chr.1: 179815161-179815161 on Build GRCh38
rs760828964 Chr.1: 179815107-179815109 on Build GRCh38
rs5779040 Chr.1: 179815148-179815151 on Build GRCh38
rs991650470 Chr.1: 179815224-179815224 on Build GRCh38
rs547387195 Chr.1: 179815152-179815152 on Build GRCh38
rs111251859 Chr.1: 179815150-179815150 on Build GRCh38
rs773702948 Chr.1: 179815092-179815097 on Build GRCh38
rs61258070 Chr.1: 179814936-179814936 on Build GRCh38
rs71571091 Chr.1: 179815105-179815118 on Build GRCh38
rs869135444 Chr.1: 179815114-179815118 on Build GRCh38
rs1028185784 Chr.1: 179814939-179814939 on Build GRCh38
rs375678554 Chr.1: 179815107-179815114 on Build GRCh38
rs142871374 Chr.1: 179815105-179815114 on Build GRCh38
rs4058502 Chr.1: 179815109-179815109 on Build GRCh38
rs543454422 Chr.1: 179815000-179815000 on Build GRCh38
rs869177921 Chr.1: 179815107-179815110 on Build GRCh38
rs866131108 Chr.1: 179815105-179815105 on Build GRCh38
rs140673030 Chr.1: 179815096-179815097 on Build GRCh38
rs3897287 Chr.1: 179815111-179815111 on Build GRCh38
rs928811355 Chr.1: 179815160-179815160 on Build GRCh38
rs543369337 Chr.1: 179815003-179815003 on Build GRCh38
rs568544092 Chr.1: 179815096-179815099 on Build GRCh38
rs373394477 Chr.1: 179815105-179815110 on Build GRCh38
rs867980406 Chr.1: 179815119-179815119 on Build GRCh38
rs373344360 Chr.1: 179814922-179814922 on Build GRCh38
rs536127882 Chr.1: 179815233-179815233 on Build GRCh38
rs3080146 Chr.1: 179815101-179815106 on Build GRCh38
rs551887362 Chr.1: 179814935-179814935 on Build GRCh38
rs565715864 Chr.1: 179815113-179815114 on Build GRCh38
rs57473297 Chr.1: 179815105-179815126 on Build GRCh38
rs531826543 Chr.1: 179815102-179815102 on Build GRCh38
rs531595874 Chr.1: 179814979-179814979 on Build GRCh38
rs4060112 Chr.1: 179815113-179815113 on Build GRCh38
rs1018324114 Chr.1: 179815117-179815117 on Build GRCh38
rs1055521304 Chr.1: 179814956-179814956 on Build GRCh38
rs1022113916 Chr.1: 179815333-179815333 on Build GRCh38
rs563081848 Chr.1: 179815046-179815046 on Build GRCh38
rs750238590 Chr.1: 179815106-179815129 on Build GRCh38
rs898196922 Chr.1: 179815151-179815151 on Build GRCh38
rs1008739885 Chr.1: 179815115-179815115 on Build GRCh38
rs915661211 Chr.1: 179815241-179815241 on Build GRCh38
rs977622416 Chr.1: 179815159-179815159 on Build GRCh38
rs143408946 Chr.1: 179815033-179815033 on Build GRCh38
rs145938450 Chr.1: 179815288-179815288 on Build GRCh38
rs370076995 Chr.1: 179815382-179815382 on Build GRCh38
rs71744420 Chr.1: 179815096-179815101 on Build GRCh38
rs2008599 Chr.1: 179815118-179815118 on Build GRCh38
rs1011529269 Chr.1: 179815140-179815140 on Build GRCh38
rs775531242 Chr.1: 179815093-179815094 on Build GRCh38
rs767690817 Chr.1: 179815107-179815112 on Build GRCh38
rs756448545 Chr.1: 179815346-179815346 on Build GRCh38
rs993358760 Chr.1: 179815052-179815052 on Build GRCh38
rs968687072 Chr.1: 179815092-179815092 on Build GRCh38
rs141649832 Chr.1: 179815261-179815261 on Build GRCh38
rs3075114 Chr.1: 179815104-179815105 on Build GRCh38
rs953933029 Chr.1: 179814972-179814972 on Build GRCh38
rs116756246 Chr.1: 179815188-179815188 on Build GRCh38
rs930699520 Chr.1: 179815102-179815115 on Build GRCh38
rs111748247 Chr.1: 179815114-179815114 on Build GRCh38
rs914617424 Chr.1: 179815078-179815078 on Build GRCh38
rs765621413 Chr.1: 179815108-179815109 on Build GRCh38
rs929225691 Chr.1: 179815299-179815299 on Build GRCh38
rs67057183 Chr.1: 179815115-179815118 on Build GRCh38
rs1037662130 Chr.1: 179815036-179815036 on Build GRCh38
rs560905771 Chr.1: 179814991-179814991 on Build GRCh38

View Full Product Details