Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180011814-180012327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP350
Entrez Gene ID
9857
Gene Name
centrosomal protein 350
Gene Aliases
CAP350, GM133
UniGene
Hs.413045
Gene Chromosome Location
Chr.1: 179954773-180114880 on Build GRCh38
Transcripts
NM_014810.4 XM_011510191.2 XM_011510198.2 XM_017002929.1 XM_017002930.1 XM_017002931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
900355 c.1280G>A p.R427Q Chr.1: 180011962-180011962 on Build GRCh38
6059870 c.1267A>G p.T423A Chr.1: 180011949-180011949 on Build GRCh38
3479373 c.1321C>T p.P441S Chr.1: 180012003-180012003 on Build GRCh38
1689325 c.1319C>T p.A440V Chr.1: 180012001-180012001 on Build GRCh38
6843059 c.1250G>C p.S417T Chr.1: 180011932-180011932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005341442 Chr.1: 180012216-180012216 on Build GRCh38
rs994124259 Chr.1: 180012264-180012264 on Build GRCh38
rs754097377 Chr.1: 180012102-180012102 on Build GRCh38
rs937376807 Chr.1: 180012248-180012248 on Build GRCh38
rs746615088 Chr.1: 180011990-180011990 on Build GRCh38
rs750258467 Chr.1: 180011919-180011920 on Build GRCh38
rs761965679 Chr.1: 180011937-180011937 on Build GRCh38
rs993847409 Chr.1: 180011979-180011979 on Build GRCh38
rs558117823 Chr.1: 180012075-180012075 on Build GRCh38
rs767449053 Chr.1: 180011946-180011946 on Build GRCh38
rs774313501 Chr.1: 180012123-180012123 on Build GRCh38
rs968066947 Chr.1: 180012205-180012205 on Build GRCh38
rs202134877 Chr.1: 180011952-180011952 on Build GRCh38
rs1037873853 Chr.1: 180012228-180012228 on Build GRCh38
rs1043488962 Chr.1: 180011896-180011896 on Build GRCh38
rs945255011 Chr.1: 180012069-180012069 on Build GRCh38
rs551475317 Chr.1: 180012159-180012159 on Build GRCh38
rs913481287 Chr.1: 180012190-180012190 on Build GRCh38
rs1017998996 Chr.1: 180012147-180012147 on Build GRCh38
rs958142042 Chr.1: 180011961-180011961 on Build GRCh38
rs756963483 Chr.1: 180012000-180012000 on Build GRCh38
rs937449279 Chr.1: 180011911-180011911 on Build GRCh38
rs745398810 Chr.1: 180012012-180012012 on Build GRCh38
rs554144833 Chr.1: 180012258-180012258 on Build GRCh38
rs531346168 Chr.1: 180012137-180012137 on Build GRCh38
rs754193891 Chr.1: 180011966-180011966 on Build GRCh38
rs755372829 Chr.1: 180011987-180011987 on Build GRCh38
rs1046310972 Chr.1: 180011951-180011951 on Build GRCh38
rs946027835 Chr.1: 180011844-180011844 on Build GRCh38
rs371238889 Chr.1: 180011950-180011950 on Build GRCh38
rs769606273 Chr.1: 180012015-180012015 on Build GRCh38
rs768625333 Chr.1: 180012106-180012106 on Build GRCh38
rs777380878 Chr.1: 180011988-180011988 on Build GRCh38
rs762899529 Chr.1: 180011885-180011885 on Build GRCh38
rs143415064 Chr.1: 180012052-180012052 on Build GRCh38
rs534487127 Chr.1: 180012224-180012224 on Build GRCh38
rs569363586 Chr.1: 180011962-180011962 on Build GRCh38
rs989265126 Chr.1: 180011976-180011976 on Build GRCh38
rs888184176 Chr.1: 180012171-180012171 on Build GRCh38
rs780502621 Chr.1: 180012004-180012004 on Build GRCh38
rs61371401 Chr.1: 180011919-180011919 on Build GRCh38
rs978769445 Chr.1: 180012223-180012223 on Build GRCh38
rs755192173 Chr.1: 180012135-180012135 on Build GRCh38
rs529730072 Chr.1: 180012086-180012086 on Build GRCh38
rs937712181 Chr.1: 180012121-180012121 on Build GRCh38
rs764407592 Chr.1: 180011913-180011913 on Build GRCh38
rs191553263 Chr.1: 180012029-180012029 on Build GRCh38
rs1030598815 Chr.1: 180012286-180012286 on Build GRCh38
rs571978909 Chr.1: 180012209-180012209 on Build GRCh38
rs926049265 Chr.1: 180011898-180011898 on Build GRCh38
rs1055858625 Chr.1: 180012288-180012288 on Build GRCh38

View Full Product Details