Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180199328-180199832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ23867
Entrez Gene ID
200058
Gene Name
uncharacterized protein FLJ23867
Gene Aliases
-
UniGene
Hs.719176
Gene Chromosome Location
Chr.1: 180198009-180200725 on Build GRCh38
Transcripts
NR_026900.1

SNP Detail

SNP ID Chromosome Location
rs28524589 Chr.1: 180199800-180199800 on Build GRCh38
rs1030119863 Chr.1: 180199699-180199699 on Build GRCh38
rs192729786 Chr.1: 180199588-180199588 on Build GRCh38
rs148761737 Chr.1: 180199525-180199525 on Build GRCh38
rs372295840 Chr.1: 180199493-180199493 on Build GRCh38
rs924975985 Chr.1: 180199762-180199762 on Build GRCh38
rs144403733 Chr.1: 180199492-180199492 on Build GRCh38
rs765129410 Chr.1: 180199584-180199584 on Build GRCh38
rs56211231 Chr.1: 180199792-180199792 on Build GRCh38
rs1020346990 Chr.1: 180199495-180199495 on Build GRCh38
rs776399567 Chr.1: 180199467-180199467 on Build GRCh38
rs34012178 Chr.1: 180199655-180199655 on Build GRCh38
rs576042600 Chr.1: 180199766-180199766 on Build GRCh38
rs56203087 Chr.1: 180199657-180199657 on Build GRCh38
rs1025249608 Chr.1: 180199398-180199398 on Build GRCh38
rs942822327 Chr.1: 180199637-180199637 on Build GRCh38
rs1016559786 Chr.1: 180199582-180199582 on Build GRCh38
rs867381890 Chr.1: 180199712-180199712 on Build GRCh38
rs933082605 Chr.1: 180199597-180199597 on Build GRCh38
rs183538780 Chr.1: 180199618-180199618 on Build GRCh38
rs766093435 Chr.1: 180199413-180199413 on Build GRCh38
rs538065052 Chr.1: 180199811-180199811 on Build GRCh38
rs544453442 Chr.1: 180199591-180199591 on Build GRCh38
rs934165067 Chr.1: 180199353-180199353 on Build GRCh38
rs897412757 Chr.1: 180199604-180199604 on Build GRCh38
rs971959154 Chr.1: 180199784-180199784 on Build GRCh38
rs550494150 Chr.1: 180199414-180199414 on Build GRCh38
rs966634644 Chr.1: 180199724-180199724 on Build GRCh38
rs916341659 Chr.1: 180199801-180199801 on Build GRCh38
rs893145974 Chr.1: 180199436-180199436 on Build GRCh38
rs188489516 Chr.1: 180199683-180199683 on Build GRCh38
rs1055112833 Chr.1: 180199427-180199427 on Build GRCh38
rs962293111 Chr.1: 180199750-180199750 on Build GRCh38
rs1041731703 Chr.1: 180199665-180199665 on Build GRCh38
rs957745814 Chr.1: 180199768-180199768 on Build GRCh38
rs958783596 Chr.1: 180199543-180199543 on Build GRCh38
rs977967383 Chr.1: 180199748-180199748 on Build GRCh38
rs16855498 Chr.1: 180199682-180199682 on Build GRCh38
rs540238444 Chr.1: 180199368-180199368 on Build GRCh38
rs1051287463 Chr.1: 180199387-180199387 on Build GRCh38
rs985867904 Chr.1: 180199602-180199602 on Build GRCh38
rs984728168 Chr.1: 180199563-180199563 on Build GRCh38
rs564623624 Chr.1: 180199616-180199616 on Build GRCh38
rs562357835 Chr.1: 180199533-180199533 on Build GRCh38
rs11414836 Chr.1: 180199795-180199796 on Build GRCh38
rs1002953397 Chr.1: 180199678-180199678 on Build GRCh38
rs893357739 Chr.1: 180199656-180199656 on Build GRCh38
rs952810718 Chr.1: 180199417-180199420 on Build GRCh38
rs375245870 Chr.1: 180199515-180199517 on Build GRCh38
rs1034180361 Chr.1: 180199534-180199534 on Build GRCh38
rs954356328 Chr.1: 180199806-180199806 on Build GRCh38
rs748574521 Chr.1: 180199587-180199587 on Build GRCh38
rs1002899662 Chr.1: 180199528-180199528 on Build GRCh38
rs756288228 Chr.1: 180199684-180199684 on Build GRCh38
rs1048459940 Chr.1: 180199634-180199634 on Build GRCh38
rs897383332 Chr.1: 180199399-180199399 on Build GRCh38
rs977768284 Chr.1: 180199555-180199555 on Build GRCh38
rs1046569200 Chr.1: 180199593-180199593 on Build GRCh38
rs1016199363 Chr.1: 180199725-180199725 on Build GRCh38
rs907204776 Chr.1: 180199675-180199675 on Build GRCh38
rs1010627047 Chr.1: 180199470-180199470 on Build GRCh38
rs553773310 Chr.1: 180199581-180199581 on Build GRCh38
rs1014277935 Chr.1: 180199384-180199384 on Build GRCh38

View Full Product Details